Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 71719071-71719559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARTPT
Entrez Gene ID
9607
Gene Name
CART prepropeptide
Gene Aliases
CART
UniGene
Hs.1707
Gene Chromosome Location
Chr.5: 71719163-71721048 on Build GRCh38
Transcripts
NM_004291.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390239 c.87G>A p.E29E Chr.5: 71719380-71719380 on Build GRCh38
4643013 c.134A>G p.D45G Chr.5: 71719427-71719427 on Build GRCh38
4585984 c.101A>G p.Q34R Chr.5: 71719394-71719394 on Build GRCh38
4669312 c.144C>A p.S48S Chr.5: 71719437-71719437 on Build GRCh38
4669311 c.131T>C p.V44A Chr.5: 71719424-71719424 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745647871 Chr.5: 71719384-71719384 on Build GRCh38
rs780919254 Chr.5: 71719374-71719374 on Build GRCh38
rs776593655 Chr.5: 71719225-71719225 on Build GRCh38
rs756619320 Chr.5: 71719346-71719346 on Build GRCh38
rs953280005 Chr.5: 71719111-71719111 on Build GRCh38
rs57596319 Chr.5: 71719175-71719175 on Build GRCh38
rs773496902 Chr.5: 71719452-71719452 on Build GRCh38
rs964903985 Chr.5: 71719536-71719536 on Build GRCh38
rs766823448 Chr.5: 71719460-71719460 on Build GRCh38
rs373471408 Chr.5: 71719487-71719487 on Build GRCh38
rs571261536 Chr.5: 71719108-71719108 on Build GRCh38
rs759699724 Chr.5: 71719471-71719471 on Build GRCh38
rs77879475 Chr.5: 71719387-71719387 on Build GRCh38
rs749355194 Chr.5: 71719245-71719245 on Build GRCh38
rs766886436 Chr.5: 71719350-71719350 on Build GRCh38
rs748320595 Chr.5: 71719368-71719368 on Build GRCh38
rs369192763 Chr.5: 71719482-71719482 on Build GRCh38
rs200591900 Chr.5: 71719493-71719493 on Build GRCh38
rs963713699 Chr.5: 71719148-71719148 on Build GRCh38
rs372413454 Chr.5: 71719421-71719421 on Build GRCh38
rs948081550 Chr.5: 71719477-71719477 on Build GRCh38
rs142927190 Chr.5: 71719324-71719324 on Build GRCh38
rs948302107 Chr.5: 71719191-71719191 on Build GRCh38
rs367802978 Chr.5: 71719307-71719307 on Build GRCh38
rs779224343 Chr.5: 71719270-71719270 on Build GRCh38
rs763557886 Chr.5: 71719330-71719332 on Build GRCh38
rs763758600 Chr.5: 71719320-71719320 on Build GRCh38
rs546329153 Chr.5: 71719092-71719092 on Build GRCh38
rs79443105 Chr.5: 71719282-71719282 on Build GRCh38
rs767124345 Chr.5: 71719441-71719441 on Build GRCh38
rs376776917 Chr.5: 71719369-71719369 on Build GRCh38
rs780062993 Chr.5: 71719243-71719243 on Build GRCh38
rs774906197 Chr.5: 71719300-71719300 on Build GRCh38
rs987247958 Chr.5: 71719215-71719215 on Build GRCh38
rs779279699 Chr.5: 71719367-71719367 on Build GRCh38
rs773955353 Chr.5: 71719321-71719321 on Build GRCh38
rs770780074 Chr.5: 71719277-71719277 on Build GRCh38
rs367870712 Chr.5: 71719248-71719248 on Build GRCh38
rs1002735857 Chr.5: 71719126-71719126 on Build GRCh38
rs1033652799 Chr.5: 71719147-71719147 on Build GRCh38
rs74779288 Chr.5: 71719354-71719354 on Build GRCh38
rs35862863 Chr.5: 71719119-71719119 on Build GRCh38
rs201676548 Chr.5: 71719458-71719458 on Build GRCh38
rs61730984 Chr.5: 71719401-71719401 on Build GRCh38
rs748828591 Chr.5: 71719418-71719418 on Build GRCh38
rs776002245 Chr.5: 71719314-71719340 on Build GRCh38
rs11544174 Chr.5: 71719422-71719422 on Build GRCh38
rs924060365 Chr.5: 71719131-71719131 on Build GRCh38
rs11544173 Chr.5: 71719375-71719375 on Build GRCh38
rs776647375 Chr.5: 71719278-71719278 on Build GRCh38
rs761518367 Chr.5: 71719434-71719434 on Build GRCh38
rs578035951 Chr.5: 71719462-71719462 on Build GRCh38
rs750208256 Chr.5: 71719490-71719490 on Build GRCh38
rs766852629 Chr.5: 71719380-71719380 on Build GRCh38
rs766124224 Chr.5: 71719505-71719505 on Build GRCh38
rs369909141 Chr.5: 71719101-71719101 on Build GRCh38
rs202033874 Chr.5: 71719232-71719232 on Build GRCh38
rs1008801965 Chr.5: 71719527-71719527 on Build GRCh38
rs1045321671 Chr.5: 71719241-71719241 on Build GRCh38
rs752823978 Chr.5: 71719480-71719480 on Build GRCh38
rs13178795 Chr.5: 71719511-71719511 on Build GRCh38
rs1018698438 Chr.5: 71719528-71719528 on Build GRCh38
rs769770010 Chr.5: 71719294-71719294 on Build GRCh38
rs376714327 Chr.5: 71719474-71719474 on Build GRCh38
rs563727160 Chr.5: 71719144-71719144 on Build GRCh38
rs1026298284 Chr.5: 71719507-71719507 on Build GRCh38
rs372232321 Chr.5: 71719342-71719342 on Build GRCh38
rs956790167 Chr.5: 71719439-71719439 on Build GRCh38
rs746828689 Chr.5: 71719274-71719274 on Build GRCh38
rs950666396 Chr.5: 71719188-71719188 on Build GRCh38
rs541530843 Chr.5: 71719431-71719431 on Build GRCh38
rs968568228 Chr.5: 71719123-71719123 on Build GRCh38
rs147424966 Chr.5: 71719356-71719356 on Build GRCh38
rs994743213 Chr.5: 71719173-71719173 on Build GRCh38
rs556896590 Chr.5: 71719167-71719167 on Build GRCh38
rs968537044 Chr.5: 71719394-71719394 on Build GRCh38
rs866218536 Chr.5: 71719213-71719213 on Build GRCh38
rs906595249 Chr.5: 71719122-71719122 on Build GRCh38
rs1046595093 Chr.5: 71719400-71719400 on Build GRCh38
rs1036064458 Chr.5: 71719293-71719293 on Build GRCh38

View Full Product Details