Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73448147-73448640 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXD1
Entrez Gene ID
2297
Gene Name
forkhead box D1
Gene Aliases
FKHL8, FREAC-4, FREAC4
UniGene
Hs.519385
Gene Chromosome Location
Chr.5: 73446256-73448527 on Build GRCh38
Transcripts
NM_004472.2

SNP Detail

SNP ID Chromosome Location
rs575327005 Chr.5: 73448340-73448340 on Build GRCh38
rs925503876 Chr.5: 73448290-73448290 on Build GRCh38
rs981601831 Chr.5: 73448590-73448590 on Build GRCh38
rs764007375 Chr.5: 73448233-73448233 on Build GRCh38
rs749900517 Chr.5: 73448251-73448251 on Build GRCh38
rs922722261 Chr.5: 73448602-73448602 on Build GRCh38
rs758488181 Chr.5: 73448192-73448193 on Build GRCh38
rs775281482 Chr.5: 73448199-73448199 on Build GRCh38
rs751490284 Chr.5: 73448243-73448243 on Build GRCh38
rs933961968 Chr.5: 73448610-73448610 on Build GRCh38
rs755845023 Chr.5: 73448252-73448252 on Build GRCh38
rs372004843 Chr.5: 73448372-73448372 on Build GRCh38
rs114980107 Chr.5: 73448294-73448294 on Build GRCh38
rs996705961 Chr.5: 73448407-73448407 on Build GRCh38
rs762746502 Chr.5: 73448229-73448229 on Build GRCh38
rs761266002 Chr.5: 73448244-73448244 on Build GRCh38
rs886783605 Chr.5: 73448338-73448338 on Build GRCh38
rs768455084 Chr.5: 73448371-73448372 on Build GRCh38
rs939664945 Chr.5: 73448358-73448358 on Build GRCh38
rs748658647 Chr.5: 73448293-73448293 on Build GRCh38
rs767163725 Chr.5: 73448248-73448248 on Build GRCh38
rs898388360 Chr.5: 73448404-73448404 on Build GRCh38
rs754320174 Chr.5: 73448279-73448279 on Build GRCh38
rs967115187 Chr.5: 73448476-73448476 on Build GRCh38
rs750578392 Chr.5: 73448193-73448195 on Build GRCh38
rs919574113 Chr.5: 73448600-73448600 on Build GRCh38
rs986883179 Chr.5: 73448187-73448187 on Build GRCh38
rs966725692 Chr.5: 73448249-73448249 on Build GRCh38
rs780101710 Chr.5: 73448259-73448261 on Build GRCh38
rs747037560 Chr.5: 73448360-73448360 on Build GRCh38
rs771097469 Chr.5: 73448182-73448182 on Build GRCh38
rs779785902 Chr.5: 73448276-73448276 on Build GRCh38
rs890957044 Chr.5: 73448433-73448433 on Build GRCh38
rs959637179 Chr.5: 73448519-73448519 on Build GRCh38
rs1021002651 Chr.5: 73448461-73448461 on Build GRCh38
rs918075860 Chr.5: 73448321-73448321 on Build GRCh38
rs991069800 Chr.5: 73448535-73448535 on Build GRCh38
rs747005522 Chr.5: 73448372-73448374 on Build GRCh38
rs939702722 Chr.5: 73448593-73448593 on Build GRCh38
rs970887037 Chr.5: 73448579-73448581 on Build GRCh38
rs543653188 Chr.5: 73448480-73448480 on Build GRCh38
rs1008761197 Chr.5: 73448458-73448458 on Build GRCh38
rs1028136718 Chr.5: 73448429-73448429 on Build GRCh38
rs1032333106 Chr.5: 73448488-73448488 on Build GRCh38
rs1038028709 Chr.5: 73448598-73448598 on Build GRCh38
rs932607471 Chr.5: 73448611-73448611 on Build GRCh38
rs1049787211 Chr.5: 73448618-73448618 on Build GRCh38
rs374981506 Chr.5: 73448288-73448288 on Build GRCh38
rs776832475 Chr.5: 73448410-73448410 on Build GRCh38
rs777852551 Chr.5: 73448333-73448333 on Build GRCh38
rs949878128 Chr.5: 73448327-73448327 on Build GRCh38
rs760919186 Chr.5: 73448180-73448180 on Build GRCh38
rs116044273 Chr.5: 73448548-73448548 on Build GRCh38

View Full Product Details