Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74629291-74629791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENC1
Entrez Gene ID
8507
Gene Name
ectodermal-neural cortex 1
Gene Aliases
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
UniGene
Hs.104925
Gene Chromosome Location
Chr.5: 74627406-74641424 on Build GRCh38
Transcripts
NM_001256574.1 NM_001256575.1 NM_001256576.1 NM_003633.3 NR_046318.1 XM_011543696.2 XM_011543697.2

SNP Detail

SNP ID Chromosome Location
rs995351137 Chr.5: 74629743-74629743 on Build GRCh38
rs913513141 Chr.5: 74629618-74629618 on Build GRCh38
rs907273623 Chr.5: 74629672-74629672 on Build GRCh38
rs1034161892 Chr.5: 74629345-74629345 on Build GRCh38
rs549129202 Chr.5: 74629560-74629560 on Build GRCh38
rs1055891044 Chr.5: 74629711-74629711 on Build GRCh38
rs898306791 Chr.5: 74629731-74629731 on Build GRCh38
rs551386315 Chr.5: 74629446-74629446 on Build GRCh38
rs936192170 Chr.5: 74629570-74629570 on Build GRCh38
rs1001584565 Chr.5: 74629679-74629679 on Build GRCh38
rs528128925 Chr.5: 74629676-74629676 on Build GRCh38
rs11557038 Chr.5: 74629466-74629466 on Build GRCh38
rs1025584175 Chr.5: 74629370-74629370 on Build GRCh38
rs138822207 Chr.5: 74629438-74629438 on Build GRCh38
rs1026526136 Chr.5: 74629423-74629423 on Build GRCh38
rs111318749 Chr.5: 74629433-74629433 on Build GRCh38
rs140549113 Chr.5: 74629480-74629480 on Build GRCh38
rs946020755 Chr.5: 74629653-74629653 on Build GRCh38
rs1014137431 Chr.5: 74629365-74629365 on Build GRCh38
rs1015911283 Chr.5: 74629467-74629467 on Build GRCh38
rs530605402 Chr.5: 74629555-74629555 on Build GRCh38
rs957833759 Chr.5: 74629500-74629500 on Build GRCh38
rs947258873 Chr.5: 74629641-74629641 on Build GRCh38
rs913172026 Chr.5: 74629614-74629614 on Build GRCh38
rs964326666 Chr.5: 74629473-74629473 on Build GRCh38
rs768656008 Chr.5: 74629485-74629485 on Build GRCh38
rs1042982729 Chr.5: 74629706-74629706 on Build GRCh38
rs779050913 Chr.5: 74629583-74629583 on Build GRCh38
rs1045769483 Chr.5: 74629660-74629660 on Build GRCh38
rs142002192 Chr.5: 74629354-74629355 on Build GRCh38
rs567045010 Chr.5: 74629611-74629611 on Build GRCh38
rs922066716 Chr.5: 74629506-74629506 on Build GRCh38
rs950944210 Chr.5: 74629425-74629425 on Build GRCh38
rs145628273 Chr.5: 74629754-74629754 on Build GRCh38
rs755850817 Chr.5: 74629315-74629315 on Build GRCh38
rs951087211 Chr.5: 74629385-74629385 on Build GRCh38
rs995382100 Chr.5: 74629358-74629358 on Build GRCh38
rs986509048 Chr.5: 74629430-74629430 on Build GRCh38
rs779667368 Chr.5: 74629355-74629355 on Build GRCh38

View Full Product Details