Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74634988-74635496 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENC1
Entrez Gene ID
8507
Gene Name
ectodermal-neural cortex 1
Gene Aliases
CCL28, ENC-1, KLHL35, KLHL37, NRPB, PIG10, TP53I10
UniGene
Hs.104925
Gene Chromosome Location
Chr.5: 74627406-74641424 on Build GRCh38
Transcripts
NM_001256574.1 NM_001256575.1 NM_001256576.1 NM_003633.3 NR_046318.1 XM_011543696.2 XM_011543697.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3073153 c.994G>T p.E332* Chr.5: 74635492-74635492 on Build GRCh38
3856261 c.1051G>C p.E351Q Chr.5: 74635435-74635435 on Build GRCh38
74511 c.1039G>C p.G347R Chr.5: 74635447-74635447 on Build GRCh38
6513106 c.1058G>T p.G353V Chr.5: 74635428-74635428 on Build GRCh38
6546321 c.1129G>A p.A377T Chr.5: 74635357-74635357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764826517 Chr.5: 74635373-74635373 on Build GRCh38
rs755213400 Chr.5: 74635472-74635472 on Build GRCh38
rs765020924 Chr.5: 74635265-74635265 on Build GRCh38
rs907154987 Chr.5: 74635275-74635275 on Build GRCh38
rs781529529 Chr.5: 74635291-74635291 on Build GRCh38
rs757531813 Chr.5: 74635417-74635417 on Build GRCh38
rs772935271 Chr.5: 74635205-74635205 on Build GRCh38
rs760662583 Chr.5: 74635224-74635224 on Build GRCh38
rs201897189 Chr.5: 74635064-74635064 on Build GRCh38
rs983624018 Chr.5: 74635461-74635461 on Build GRCh38
rs775659610 Chr.5: 74635187-74635187 on Build GRCh38
rs781365084 Chr.5: 74635177-74635177 on Build GRCh38
rs778363964 Chr.5: 74635195-74635195 on Build GRCh38
rs148438771 Chr.5: 74635148-74635148 on Build GRCh38
rs753407500 Chr.5: 74635230-74635230 on Build GRCh38
rs144525141 Chr.5: 74635435-74635435 on Build GRCh38
rs770103291 Chr.5: 74635181-74635181 on Build GRCh38
rs770945434 Chr.5: 74635330-74635330 on Build GRCh38
rs751832690 Chr.5: 74635285-74635285 on Build GRCh38
rs369592581 Chr.5: 74635343-74635343 on Build GRCh38
rs760336240 Chr.5: 74635327-74635327 on Build GRCh38
rs1010099136 Chr.5: 74635183-74635183 on Build GRCh38
rs759810274 Chr.5: 74635090-74635090 on Build GRCh38
rs746631875 Chr.5: 74635091-74635091 on Build GRCh38
rs771845512 Chr.5: 74635202-74635202 on Build GRCh38
rs776773829 Chr.5: 74635081-74635081 on Build GRCh38
rs145105480 Chr.5: 74635174-74635174 on Build GRCh38
rs757806641 Chr.5: 74635286-74635286 on Build GRCh38
rs749528223 Chr.5: 74635301-74635301 on Build GRCh38
rs973817940 Chr.5: 74635379-74635379 on Build GRCh38
rs758804267 Chr.5: 74635169-74635169 on Build GRCh38
rs769864224 Chr.5: 74635295-74635295 on Build GRCh38
rs752006109 Chr.5: 74635412-74635412 on Build GRCh38
rs776398012 Chr.5: 74635336-74635336 on Build GRCh38
rs768803538 Chr.5: 74635302-74635302 on Build GRCh38
rs780317395 Chr.5: 74635296-74635296 on Build GRCh38
rs770932086 Chr.5: 74635300-74635301 on Build GRCh38
rs780114303 Chr.5: 74635440-74635440 on Build GRCh38
rs1024228789 Chr.5: 74635262-74635262 on Build GRCh38
rs750885340 Chr.5: 74635426-74635426 on Build GRCh38
rs765928883 Chr.5: 74635143-74635143 on Build GRCh38
rs763791861 Chr.5: 74635398-74635398 on Build GRCh38
rs988443396 Chr.5: 74635273-74635273 on Build GRCh38
rs749330219 Chr.5: 74635445-74635445 on Build GRCh38
rs765338534 Chr.5: 74635106-74635106 on Build GRCh38
rs888489840 Chr.5: 74635061-74635061 on Build GRCh38
rs759151780 Chr.5: 74635371-74635371 on Build GRCh38
rs778294516 Chr.5: 74635279-74635279 on Build GRCh38
rs919131141 Chr.5: 74635460-74635460 on Build GRCh38
rs546313916 Chr.5: 74635390-74635390 on Build GRCh38
rs778490596 Chr.5: 74635172-74635172 on Build GRCh38
rs572526118 Chr.5: 74635292-74635292 on Build GRCh38
rs754688646 Chr.5: 74635154-74635154 on Build GRCh38
rs144437936 Chr.5: 74635139-74635139 on Build GRCh38
rs762698517 Chr.5: 74635391-74635391 on Build GRCh38
rs11747630 Chr.5: 74635068-74635068 on Build GRCh38
rs771283848 Chr.5: 74635175-74635175 on Build GRCh38
rs147620275 Chr.5: 74635178-74635178 on Build GRCh38
rs761573677 Chr.5: 74635120-74635120 on Build GRCh38
rs771037231 Chr.5: 74635066-74635066 on Build GRCh38
rs748568186 Chr.5: 74635161-74635161 on Build GRCh38
rs767486474 Chr.5: 74635121-74635121 on Build GRCh38
rs761667313 Chr.5: 74635194-74635194 on Build GRCh38
rs758889299 Chr.5: 74635274-74635274 on Build GRCh38
rs442425 Chr.5: 74635421-74635421 on Build GRCh38
rs773833247 Chr.5: 74635028-74635028 on Build GRCh38
rs766184349 Chr.5: 74635228-74635228 on Build GRCh38
rs758943823 Chr.5: 74635244-74635244 on Build GRCh38
rs950866641 Chr.5: 74635439-74635439 on Build GRCh38
rs778656065 Chr.5: 74635160-74635160 on Build GRCh38
rs956854151 Chr.5: 74635214-74635214 on Build GRCh38
rs368405991 Chr.5: 74635138-74635138 on Build GRCh38
rs773018893 Chr.5: 74635316-74635316 on Build GRCh38

View Full Product Details