Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74781208-74781721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169A
Entrez Gene ID
26049
Gene Name
family with sequence similarity 169 member A
Gene Aliases
SLAP75
UniGene
Hs.91662
Gene Chromosome Location
Chr.5: 74777574-74867994 on Build GRCh38
Transcripts
NM_015566.2 NR_046462.1 XM_005248480.2 XM_011543306.2 XM_011543307.2 XM_017009332.1 XM_017009333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5047092 c.1888G>A p.E630K Chr.5: 74781585-74781585 on Build GRCh38
4142048 c.1856C>G p.S619C Chr.5: 74781617-74781617 on Build GRCh38
421190 c.1900G>T p.D634Y Chr.5: 74781573-74781573 on Build GRCh38
1070033 c.1896T>C p.A632A Chr.5: 74781577-74781577 on Build GRCh38
4533445 c.1982G>A p.G661E Chr.5: 74781491-74781491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032887544 Chr.5: 74781395-74781395 on Build GRCh38
rs375344386 Chr.5: 74781512-74781512 on Build GRCh38
rs772962791 Chr.5: 74781458-74781458 on Build GRCh38
rs756399674 Chr.5: 74781589-74781589 on Build GRCh38
rs764687081 Chr.5: 74781549-74781549 on Build GRCh38
rs897300234 Chr.5: 74781616-74781616 on Build GRCh38
rs572085553 Chr.5: 74781639-74781639 on Build GRCh38
rs920641409 Chr.5: 74781613-74781613 on Build GRCh38
rs972311739 Chr.5: 74781234-74781234 on Build GRCh38
rs772139541 Chr.5: 74781454-74781454 on Build GRCh38
rs1056360942 Chr.5: 74781321-74781321 on Build GRCh38
rs746054700 Chr.5: 74781430-74781430 on Build GRCh38
rs917810987 Chr.5: 74781329-74781329 on Build GRCh38
rs771713405 Chr.5: 74781612-74781612 on Build GRCh38
rs753068782 Chr.5: 74781525-74781525 on Build GRCh38
rs770532392 Chr.5: 74781635-74781635 on Build GRCh38
rs961984268 Chr.5: 74781537-74781537 on Build GRCh38
rs369250258 Chr.5: 74781515-74781515 on Build GRCh38
rs866010178 Chr.5: 74781575-74781575 on Build GRCh38
rs546239796 Chr.5: 74781336-74781336 on Build GRCh38
rs968102060 Chr.5: 74781301-74781301 on Build GRCh38
rs369288865 Chr.5: 74781588-74781588 on Build GRCh38
rs778872207 Chr.5: 74781476-74781478 on Build GRCh38
rs747968921 Chr.5: 74781601-74781601 on Build GRCh38
rs768617020 Chr.5: 74781440-74781440 on Build GRCh38
rs558675325 Chr.5: 74781371-74781371 on Build GRCh38
rs887566264 Chr.5: 74781391-74781391 on Build GRCh38
rs760253599 Chr.5: 74781461-74781461 on Build GRCh38
rs781504252 Chr.5: 74781424-74781424 on Build GRCh38
rs555077937 Chr.5: 74781355-74781355 on Build GRCh38
rs377242596 Chr.5: 74781574-74781574 on Build GRCh38
rs776396916 Chr.5: 74781505-74781505 on Build GRCh38
rs1038556966 Chr.5: 74781593-74781593 on Build GRCh38
rs574608618 Chr.5: 74781630-74781630 on Build GRCh38
rs553702150 Chr.5: 74781678-74781678 on Build GRCh38
rs746899812 Chr.5: 74781622-74781622 on Build GRCh38
rs761280706 Chr.5: 74781408-74781410 on Build GRCh38
rs373494393 Chr.5: 74781444-74781444 on Build GRCh38
rs893038953 Chr.5: 74781425-74781425 on Build GRCh38
rs923740474 Chr.5: 74781303-74781303 on Build GRCh38
rs764887237 Chr.5: 74781670-74781670 on Build GRCh38
rs781105582 Chr.5: 74781580-74781580 on Build GRCh38
rs754231250 Chr.5: 74781304-74781304 on Build GRCh38
rs200386622 Chr.5: 74781553-74781553 on Build GRCh38
rs372575886 Chr.5: 74781520-74781520 on Build GRCh38
rs377658529 Chr.5: 74781659-74781659 on Build GRCh38
rs777501979 Chr.5: 74781485-74781485 on Build GRCh38
rs373307378 Chr.5: 74781565-74781565 on Build GRCh38
rs979234589 Chr.5: 74781302-74781302 on Build GRCh38
rs201025077 Chr.5: 74781615-74781615 on Build GRCh38
rs920851753 Chr.5: 74781253-74781253 on Build GRCh38
rs774270869 Chr.5: 74781442-74781442 on Build GRCh38
rs757208180 Chr.5: 74781439-74781441 on Build GRCh38
rs34996582 Chr.5: 74781605-74781606 on Build GRCh38
rs1023204491 Chr.5: 74781511-74781511 on Build GRCh38
rs955805278 Chr.5: 74781667-74781667 on Build GRCh38
rs16872269 Chr.5: 74781649-74781649 on Build GRCh38
rs773243071 Chr.5: 74781618-74781618 on Build GRCh38
rs1012867069 Chr.5: 74781439-74781439 on Build GRCh38
rs751968067 Chr.5: 74781550-74781550 on Build GRCh38
rs757928736 Chr.5: 74781384-74781384 on Build GRCh38
rs113748692 Chr.5: 74781307-74781307 on Build GRCh38
rs554768931 Chr.5: 74781590-74781590 on Build GRCh38
rs199744243 Chr.5: 74781330-74781332 on Build GRCh38
rs189973366 Chr.5: 74781417-74781417 on Build GRCh38
rs139333387 Chr.5: 74781331-74781334 on Build GRCh38
rs758708052 Chr.5: 74781534-74781534 on Build GRCh38
rs764331056 Chr.5: 74781692-74781692 on Build GRCh38
rs763342531 Chr.5: 74781679-74781679 on Build GRCh38
rs747128003 Chr.5: 74781411-74781411 on Build GRCh38
rs566478941 Chr.5: 74781338-74781338 on Build GRCh38

View Full Product Details