Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 74782853-74783367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM169A
Entrez Gene ID
26049
Gene Name
family with sequence similarity 169 member A
Gene Aliases
SLAP75
UniGene
Hs.91662
Gene Chromosome Location
Chr.5: 74777574-74867994 on Build GRCh38
Transcripts
NM_015566.2 NR_046462.1 XM_005248480.2 XM_011543306.2 XM_011543307.2 XM_017009332.1 XM_017009333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7367135 c.1273G>C p.E425Q Chr.5: 74783122-74783122 on Build GRCh38
1438548 c.1359T>C p.D453D Chr.5: 74783036-74783036 on Build GRCh38
1438549 c.1299C>T p.D433D Chr.5: 74783096-74783096 on Build GRCh38
6297538 c.1375C>A p.Q459K Chr.5: 74783020-74783020 on Build GRCh38
4411019 c.1411G>T p.V471F Chr.5: 74782984-74782984 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377572506 Chr.5: 74783148-74783148 on Build GRCh38
rs777151105 Chr.5: 74782883-74782883 on Build GRCh38
rs1013691718 Chr.5: 74783260-74783260 on Build GRCh38
rs765333034 Chr.5: 74782904-74782904 on Build GRCh38
rs753768291 Chr.5: 74783073-74783073 on Build GRCh38
rs918514426 Chr.5: 74782898-74782898 on Build GRCh38
rs775627956 Chr.5: 74782906-74782906 on Build GRCh38
rs777512485 Chr.5: 74782948-74782948 on Build GRCh38
rs542312668 Chr.5: 74783100-74783100 on Build GRCh38
rs576419179 Chr.5: 74783056-74783056 on Build GRCh38
rs746368964 Chr.5: 74782972-74782972 on Build GRCh38
rs889644241 Chr.5: 74782923-74782923 on Build GRCh38
rs1039153377 Chr.5: 74783011-74783011 on Build GRCh38
rs754939341 Chr.5: 74783074-74783074 on Build GRCh38
rs755399119 Chr.5: 74783270-74783270 on Build GRCh38
rs747401145 Chr.5: 74782949-74782949 on Build GRCh38
rs201356825 Chr.5: 74783155-74783155 on Build GRCh38
rs776491459 Chr.5: 74783173-74783173 on Build GRCh38
rs747823067 Chr.5: 74782942-74782942 on Build GRCh38
rs757934117 Chr.5: 74783108-74783108 on Build GRCh38
rs761016690 Chr.5: 74782922-74782922 on Build GRCh38
rs775631632 Chr.5: 74782992-74782992 on Build GRCh38
rs971723644 Chr.5: 74783002-74783002 on Build GRCh38
rs931265470 Chr.5: 74782905-74782906 on Build GRCh38
rs377550660 Chr.5: 74782932-74782932 on Build GRCh38
rs775065396 Chr.5: 74783180-74783181 on Build GRCh38
rs765017697 Chr.5: 74783093-74783093 on Build GRCh38
rs1012275203 Chr.5: 74783192-74783192 on Build GRCh38
rs561439137 Chr.5: 74782905-74782905 on Build GRCh38
rs541678472 Chr.5: 74783288-74783288 on Build GRCh38
rs764310860 Chr.5: 74783030-74783030 on Build GRCh38
rs752150913 Chr.5: 74783095-74783095 on Build GRCh38
rs777046319 Chr.5: 74782961-74782961 on Build GRCh38
rs905269148 Chr.5: 74783343-74783343 on Build GRCh38
rs777602574 Chr.5: 74783115-74783115 on Build GRCh38
rs771385257 Chr.5: 74782958-74782958 on Build GRCh38
rs771844817 Chr.5: 74783147-74783148 on Build GRCh38
rs556154499 Chr.5: 74782945-74782945 on Build GRCh38
rs746425335 Chr.5: 74783136-74783136 on Build GRCh38
rs533689307 Chr.5: 74783306-74783306 on Build GRCh38
rs774594308 Chr.5: 74783154-74783154 on Build GRCh38
rs769825795 Chr.5: 74782975-74782975 on Build GRCh38
rs774506364 Chr.5: 74783038-74783038 on Build GRCh38
rs759194459 Chr.5: 74783183-74783183 on Build GRCh38
rs1039841234 Chr.5: 74783184-74783184 on Build GRCh38
rs778463540 Chr.5: 74782936-74782936 on Build GRCh38
rs755843386 Chr.5: 74782920-74782920 on Build GRCh38
rs918284378 Chr.5: 74783291-74783291 on Build GRCh38
rs750221072 Chr.5: 74782919-74782919 on Build GRCh38
rs371105140 Chr.5: 74783150-74783150 on Build GRCh38
rs199789721 Chr.5: 74782928-74782928 on Build GRCh38
rs374114319 Chr.5: 74783144-74783144 on Build GRCh38
rs780365421 Chr.5: 74783147-74783147 on Build GRCh38
rs979793943 Chr.5: 74783203-74783203 on Build GRCh38
rs766166456 Chr.5: 74783060-74783060 on Build GRCh38
rs931230510 Chr.5: 74783302-74783302 on Build GRCh38
rs763145750 Chr.5: 74782911-74782911 on Build GRCh38
rs571256628 Chr.5: 74783257-74783257 on Build GRCh38
rs762951169 Chr.5: 74782995-74782995 on Build GRCh38
rs377517351 Chr.5: 74782891-74782891 on Build GRCh38
rs754102976 Chr.5: 74783194-74783194 on Build GRCh38
rs770689107 Chr.5: 74783161-74783161 on Build GRCh38
rs369687897 Chr.5: 74783122-74783122 on Build GRCh38
rs1021777047 Chr.5: 74783196-74783196 on Build GRCh38
rs781610478 Chr.5: 74783127-74783127 on Build GRCh38
rs766324727 Chr.5: 74782926-74782926 on Build GRCh38
rs892435165 Chr.5: 74783251-74783251 on Build GRCh38
rs189191625 Chr.5: 74783310-74783310 on Build GRCh38
rs925578099 Chr.5: 74783186-74783186 on Build GRCh38
rs972371632 Chr.5: 74783287-74783287 on Build GRCh38
rs767363387 Chr.5: 74782912-74782912 on Build GRCh38
rs1035410938 Chr.5: 74783235-74783235 on Build GRCh38
rs751227953 Chr.5: 74783120-74783120 on Build GRCh38
rs564746665 Chr.5: 74783289-74783289 on Build GRCh38
rs572589660 Chr.5: 74783280-74783280 on Build GRCh38
rs1056822700 Chr.5: 74783202-74783202 on Build GRCh38
rs943413050 Chr.5: 74782946-74782946 on Build GRCh38

View Full Product Details