Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75080218-75080745 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD31
Entrez Gene ID
256006
Gene Name
ankyrin repeat domain 31
Gene Aliases
-
UniGene
Hs.482533
Gene Chromosome Location
Chr.5: 75068297-75236897 on Build GRCh38
Transcripts
NM_001164443.1 XM_011543298.2 XM_011543299.1 XM_011543300.2 XM_011543301.2 XM_011543302.2 XM_017009317.1 XM_017009318.1 XM_017009319.1 XM_017009320.1 XM_017009321.1

SNP Detail

SNP ID Chromosome Location
rs953953116 Chr.5: 75080489-75080489 on Build GRCh38
rs965233550 Chr.5: 75080535-75080535 on Build GRCh38
rs751896148 Chr.5: 75080422-75080423 on Build GRCh38
rs139536292 Chr.5: 75080382-75080382 on Build GRCh38
rs769990481 Chr.5: 75080529-75080529 on Build GRCh38
rs565925931 Chr.5: 75080345-75080345 on Build GRCh38
rs571943744 Chr.5: 75080404-75080404 on Build GRCh38
rs889431842 Chr.5: 75080710-75080710 on Build GRCh38
rs564139060 Chr.5: 75080718-75080718 on Build GRCh38
rs778587832 Chr.5: 75080532-75080532 on Build GRCh38
rs907451574 Chr.5: 75080633-75080633 on Build GRCh38
rs116960235 Chr.5: 75080307-75080307 on Build GRCh38
rs760315290 Chr.5: 75080602-75080602 on Build GRCh38
rs533156718 Chr.5: 75080267-75080267 on Build GRCh38
rs541935925 Chr.5: 75080648-75080648 on Build GRCh38
rs372149308 Chr.5: 75080657-75080657 on Build GRCh38
rs896474271 Chr.5: 75080632-75080632 on Build GRCh38
rs1044650858 Chr.5: 75080619-75080619 on Build GRCh38
rs961940288 Chr.5: 75080714-75080714 on Build GRCh38
rs985764326 Chr.5: 75080513-75080513 on Build GRCh38
rs150662203 Chr.5: 75080377-75080377 on Build GRCh38
rs771935848 Chr.5: 75080548-75080548 on Build GRCh38
rs962085653 Chr.5: 75080436-75080436 on Build GRCh38
rs1027080682 Chr.5: 75080700-75080700 on Build GRCh38
rs550117358 Chr.5: 75080278-75080278 on Build GRCh38
rs749365181 Chr.5: 75080277-75080277 on Build GRCh38
rs907355129 Chr.5: 75080341-75080341 on Build GRCh38
rs541252698 Chr.5: 75080716-75080716 on Build GRCh38
rs1015160608 Chr.5: 75080712-75080712 on Build GRCh38
rs202150644 Chr.5: 75080329-75080329 on Build GRCh38
rs113073861 Chr.5: 75080674-75080674 on Build GRCh38
rs924726409 Chr.5: 75080564-75080564 on Build GRCh38
rs1012214076 Chr.5: 75080642-75080642 on Build GRCh38
rs922922840 Chr.5: 75080497-75080497 on Build GRCh38
rs775855882 Chr.5: 75080555-75080556 on Build GRCh38
rs760055411 Chr.5: 75080530-75080531 on Build GRCh38
rs924703350 Chr.5: 75080260-75080260 on Build GRCh38
rs1012160215 Chr.5: 75080344-75080344 on Build GRCh38
rs551743942 Chr.5: 75080568-75080568 on Build GRCh38
rs201792584 Chr.5: 75080556-75080556 on Build GRCh38
rs565619358 Chr.5: 75080302-75080302 on Build GRCh38
rs759062435 Chr.5: 75080677-75080677 on Build GRCh38
rs763841898 Chr.5: 75080590-75080590 on Build GRCh38
rs747928004 Chr.5: 75080539-75080539 on Build GRCh38
rs4447963 Chr.5: 75080484-75080484 on Build GRCh38
rs961145541 Chr.5: 75080414-75080414 on Build GRCh38
rs149690398 Chr.5: 75080571-75080571 on Build GRCh38
rs375874932 Chr.5: 75080658-75080658 on Build GRCh38
rs1014779520 Chr.5: 75080418-75080418 on Build GRCh38
rs772499843 Chr.5: 75080572-75080572 on Build GRCh38
rs977940760 Chr.5: 75080541-75080541 on Build GRCh38
rs549077231 Chr.5: 75080327-75080327 on Build GRCh38
rs766694933 Chr.5: 75080292-75080292 on Build GRCh38
rs73127353 Chr.5: 75080509-75080509 on Build GRCh38
rs572244018 Chr.5: 75080687-75080687 on Build GRCh38
rs999633523 Chr.5: 75080305-75080305 on Build GRCh38
rs887672584 Chr.5: 75080670-75080670 on Build GRCh38
rs912419138 Chr.5: 75080518-75080518 on Build GRCh38
rs1056584546 Chr.5: 75080608-75080608 on Build GRCh38

View Full Product Details