Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 75371019-75371527 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COL4A3BP
Entrez Gene ID
10087
Gene Name
collagen type IV alpha 3 binding protein
Gene Aliases
CERT, CERTL, GPBP, MRD34, STARD11
UniGene
Hs.270437
Gene Chromosome Location
Chr.5: 75368486-75511981 on Build GRCh38
Transcripts
NM_001130105.1 NM_005713.2 NM_031361.2 XM_006714513.2 XM_011543090.2 XM_011543091.2 XM_017008919.1

SNP Detail

SNP ID Chromosome Location
rs575919463 Chr.5: 75371292-75371292 on Build GRCh38
rs147434171 Chr.5: 75371067-75371067 on Build GRCh38
rs535269375 Chr.5: 75371062-75371062 on Build GRCh38
rs764670863 Chr.5: 75371443-75371443 on Build GRCh38
rs1041683726 Chr.5: 75371059-75371059 on Build GRCh38
rs987223465 Chr.5: 75371311-75371311 on Build GRCh38
rs1052612856 Chr.5: 75371412-75371412 on Build GRCh38
rs1000243151 Chr.5: 75371063-75371063 on Build GRCh38
rs1011177079 Chr.5: 75371481-75371481 on Build GRCh38
rs963354846 Chr.5: 75371219-75371219 on Build GRCh38
rs945938059 Chr.5: 75371337-75371337 on Build GRCh38
rs544873041 Chr.5: 75371215-75371215 on Build GRCh38
rs535189896 Chr.5: 75371324-75371324 on Build GRCh38
rs930578625 Chr.5: 75371300-75371300 on Build GRCh38
rs931749768 Chr.5: 75371281-75371281 on Build GRCh38
rs894542176 Chr.5: 75371493-75371493 on Build GRCh38
rs774710862 Chr.5: 75371225-75371225 on Build GRCh38
rs771271158 Chr.5: 75371226-75371228 on Build GRCh38
rs921656051 Chr.5: 75371236-75371236 on Build GRCh38
rs1034805374 Chr.5: 75371192-75371192 on Build GRCh38
rs1042026347 Chr.5: 75371353-75371353 on Build GRCh38
rs774921445 Chr.5: 75371254-75371255 on Build GRCh38
rs1014403248 Chr.5: 75371100-75371100 on Build GRCh38
rs563280580 Chr.5: 75371441-75371441 on Build GRCh38
rs181114252 Chr.5: 75371197-75371197 on Build GRCh38
rs377385070 Chr.5: 75371071-75371071 on Build GRCh38
rs537304011 Chr.5: 75371144-75371144 on Build GRCh38
rs904464943 Chr.5: 75371366-75371366 on Build GRCh38
rs1024498655 Chr.5: 75371113-75371113 on Build GRCh38
rs1011291849 Chr.5: 75371098-75371098 on Build GRCh38
rs529713131 Chr.5: 75371101-75371101 on Build GRCh38
rs60625896 Chr.5: 75371427-75371427 on Build GRCh38
rs982177043 Chr.5: 75371169-75371169 on Build GRCh38
rs1040171304 Chr.5: 75371341-75371341 on Build GRCh38
rs185660652 Chr.5: 75371407-75371407 on Build GRCh38
rs973120692 Chr.5: 75371222-75371222 on Build GRCh38
rs3180743 Chr.5: 75371104-75371104 on Build GRCh38
rs983430686 Chr.5: 75371153-75371153 on Build GRCh38
rs760020805 Chr.5: 75371312-75371312 on Build GRCh38
rs1017225928 Chr.5: 75371161-75371161 on Build GRCh38

View Full Product Details