Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76616546-76617081 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
UniGene
Hs.42502
Gene Chromosome Location
Chr.5: 76615482-76623434 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

SNP Detail

SNP ID Chromosome Location
rs547273908 Chr.5: 76616695-76616695 on Build GRCh38
rs536537852 Chr.5: 76616636-76616636 on Build GRCh38
rs2069689 Chr.5: 76616576-76616576 on Build GRCh38
rs879564631 Chr.5: 76616688-76616688 on Build GRCh38
rs973446030 Chr.5: 76616597-76616597 on Build GRCh38
rs977626684 Chr.5: 76616693-76616694 on Build GRCh38
rs141376959 Chr.5: 76616700-76616700 on Build GRCh38
rs993822293 Chr.5: 76616805-76616805 on Build GRCh38
rs988182741 Chr.5: 76616993-76616993 on Build GRCh38
rs575678994 Chr.5: 76616965-76616965 on Build GRCh38
rs956675626 Chr.5: 76616973-76616973 on Build GRCh38
rs34421679 Chr.5: 76616857-76616857 on Build GRCh38
rs947296414 Chr.5: 76617002-76617002 on Build GRCh38
rs2069704 Chr.5: 76616615-76616615 on Build GRCh38
rs570686103 Chr.5: 76616917-76616917 on Build GRCh38
rs977257954 Chr.5: 76616934-76616934 on Build GRCh38
rs749097235 Chr.5: 76616727-76616727 on Build GRCh38
rs150370480 Chr.5: 76616839-76616839 on Build GRCh38
rs558749688 Chr.5: 76616840-76616840 on Build GRCh38
rs964120584 Chr.5: 76616931-76616931 on Build GRCh38
rs1018864510 Chr.5: 76616907-76616907 on Build GRCh38
rs768498680 Chr.5: 76616886-76616886 on Build GRCh38
rs1027635107 Chr.5: 76616599-76616599 on Build GRCh38
rs551159409 Chr.5: 76616584-76616584 on Build GRCh38
rs947709592 Chr.5: 76616859-76616859 on Build GRCh38
rs961536 Chr.5: 76617018-76617018 on Build GRCh38
rs759571869 Chr.5: 76616834-76616834 on Build GRCh38
rs544747148 Chr.5: 76617009-76617009 on Build GRCh38
rs947202991 Chr.5: 76616635-76616635 on Build GRCh38
rs567566627 Chr.5: 76616622-76616622 on Build GRCh38
rs571332772 Chr.5: 76616587-76616587 on Build GRCh38
rs927139463 Chr.5: 76617038-76617038 on Build GRCh38
rs976744367 Chr.5: 76616577-76616577 on Build GRCh38
rs922594150 Chr.5: 76616578-76616578 on Build GRCh38
rs536782543 Chr.5: 76616591-76616591 on Build GRCh38
rs905578728 Chr.5: 76616683-76616683 on Build GRCh38
rs935098521 Chr.5: 76616785-76616785 on Build GRCh38
rs2069688 Chr.5: 76616705-76616705 on Build GRCh38
rs774013674 Chr.5: 76616955-76616955 on Build GRCh38
rs1025834078 Chr.5: 76616835-76616835 on Build GRCh38
rs895499507 Chr.5: 76616716-76616716 on Build GRCh38
rs913145916 Chr.5: 76616998-76616998 on Build GRCh38

View Full Product Details