Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76616946-76617472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL2
Entrez Gene ID
2151
Gene Name
coagulation factor II thrombin receptor like 2
Gene Aliases
PAR-3, PAR3
UniGene
Hs.42502
Gene Chromosome Location
Chr.5: 76615482-76623434 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

Gene Symbol
IQGAP2
Entrez Gene ID
10788
Gene Name
IQ motif containing GTPase activating protein 2
Gene Aliases
-
UniGene
Hs.291030
Gene Chromosome Location
Chr.5: 76403255-76708132 on Build GRCh38
Transcripts
NM_001256566.1 NM_004101.3 NM_001285460.1 NM_001285461.1 NM_001285462.1 NM_006633.3 XM_005248410.2 XM_005248414.1 XM_011543108.1 XM_017008960.1

SNP Detail

SNP ID Chromosome Location
rs2069703 Chr.5: 76617115-76617115 on Build GRCh38
rs916989729 Chr.5: 76617340-76617340 on Build GRCh38
rs570072931 Chr.5: 76617234-76617234 on Build GRCh38
rs1040345188 Chr.5: 76617443-76617443 on Build GRCh38
rs551078614 Chr.5: 76617391-76617391 on Build GRCh38
rs992713069 Chr.5: 76617284-76617284 on Build GRCh38
rs888663492 Chr.5: 76617113-76617113 on Build GRCh38
rs968600230 Chr.5: 76617252-76617252 on Build GRCh38
rs181385728 Chr.5: 76617178-76617178 on Build GRCh38
rs988182741 Chr.5: 76616993-76616993 on Build GRCh38
rs372139148 Chr.5: 76617428-76617428 on Build GRCh38
rs956675626 Chr.5: 76616973-76616973 on Build GRCh38
rs937077994 Chr.5: 76617075-76617075 on Build GRCh38
rs188551970 Chr.5: 76617158-76617158 on Build GRCh38
rs929641843 Chr.5: 76617094-76617094 on Build GRCh38
rs947296414 Chr.5: 76617002-76617002 on Build GRCh38
rs550301042 Chr.5: 76617344-76617344 on Build GRCh38
rs956812462 Chr.5: 76617198-76617198 on Build GRCh38
rs753600254 Chr.5: 76617274-76617274 on Build GRCh38
rs573925844 Chr.5: 76617104-76617104 on Build GRCh38
rs947790955 Chr.5: 76617347-76617347 on Build GRCh38
rs2069687 Chr.5: 76617167-76617167 on Build GRCh38
rs1046748816 Chr.5: 76617112-76617112 on Build GRCh38
rs772808862 Chr.5: 76617141-76617141 on Build GRCh38
rs998390675 Chr.5: 76617170-76617170 on Build GRCh38
rs530516675 Chr.5: 76617332-76617332 on Build GRCh38
rs1046672204 Chr.5: 76617390-76617390 on Build GRCh38
rs961536 Chr.5: 76617018-76617018 on Build GRCh38
rs544747148 Chr.5: 76617009-76617009 on Build GRCh38
rs759254142 Chr.5: 76617400-76617400 on Build GRCh38
rs567479800 Chr.5: 76617363-76617363 on Build GRCh38
rs994899810 Chr.5: 76617066-76617066 on Build GRCh38
rs757460096 Chr.5: 76617420-76617420 on Build GRCh38
rs766140776 Chr.5: 76617237-76617237 on Build GRCh38
rs979101098 Chr.5: 76617270-76617270 on Build GRCh38
rs927139463 Chr.5: 76617038-76617038 on Build GRCh38
rs897644207 Chr.5: 76617061-76617061 on Build GRCh38
rs368607249 Chr.5: 76617366-76617366 on Build GRCh38
rs927262691 Chr.5: 76617273-76617273 on Build GRCh38
rs895592683 Chr.5: 76617093-76617093 on Build GRCh38
rs1022308877 Chr.5: 76617228-76617228 on Build GRCh38
rs929735134 Chr.5: 76617362-76617362 on Build GRCh38
rs1027667151 Chr.5: 76617086-76617086 on Build GRCh38
rs760829267 Chr.5: 76617149-76617149 on Build GRCh38
rs186008904 Chr.5: 76617204-76617204 on Build GRCh38
rs989244760 Chr.5: 76617255-76617255 on Build GRCh38
rs1057270896 Chr.5: 76617092-76617092 on Build GRCh38
rs913145916 Chr.5: 76616998-76616998 on Build GRCh38

View Full Product Details