Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76834839-76835358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F2RL1
Entrez Gene ID
2150
Gene Name
F2R like trypsin receptor 1
Gene Aliases
GPR11, PAR2
UniGene
Hs.744181
Gene Chromosome Location
Chr.5: 76819008-76835315 on Build GRCh38
Transcripts
XM_017009223.1

SNP Detail

SNP ID Chromosome Location
rs941047990 Chr.5: 76835298-76835298 on Build GRCh38
rs900000981 Chr.5: 76834876-76834876 on Build GRCh38
rs1005244021 Chr.5: 76835051-76835051 on Build GRCh38
rs376364152 Chr.5: 76834957-76834957 on Build GRCh38
rs1009635373 Chr.5: 76834922-76834922 on Build GRCh38
rs951183123 Chr.5: 76835330-76835330 on Build GRCh38
rs1051191508 Chr.5: 76834917-76834917 on Build GRCh38
rs998330727 Chr.5: 76834894-76834894 on Build GRCh38
rs988822660 Chr.5: 76835154-76835154 on Build GRCh38
rs951158657 Chr.5: 76835149-76835149 on Build GRCh38
rs762023152 Chr.5: 76834941-76834941 on Build GRCh38
rs573164812 Chr.5: 76835239-76835239 on Build GRCh38
rs926698396 Chr.5: 76835237-76835237 on Build GRCh38
rs1040378343 Chr.5: 76835236-76835236 on Build GRCh38
rs892634309 Chr.5: 76835260-76835260 on Build GRCh38
rs940996240 Chr.5: 76835223-76835223 on Build GRCh38
rs143476361 Chr.5: 76835308-76835308 on Build GRCh38
rs982550989 Chr.5: 76835175-76835175 on Build GRCh38
rs147763831 Chr.5: 76835071-76835071 on Build GRCh38
rs909545379 Chr.5: 76835193-76835193 on Build GRCh38
rs886797068 Chr.5: 76835031-76835034 on Build GRCh38
rs575690460 Chr.5: 76835299-76835299 on Build GRCh38
rs1002188816 Chr.5: 76835287-76835287 on Build GRCh38
rs113368839 Chr.5: 76835118-76835118 on Build GRCh38
rs146194119 Chr.5: 76834956-76834956 on Build GRCh38
rs955642682 Chr.5: 76835146-76835146 on Build GRCh38
rs945478904 Chr.5: 76835273-76835273 on Build GRCh38
rs937209932 Chr.5: 76834965-76834965 on Build GRCh38
rs1034233222 Chr.5: 76834959-76834959 on Build GRCh38
rs558997126 Chr.5: 76835280-76835280 on Build GRCh38
rs752294684 Chr.5: 76835283-76835283 on Build GRCh38
rs534930455 Chr.5: 76834925-76834925 on Build GRCh38
rs899974780 Chr.5: 76835089-76835089 on Build GRCh38
rs187241307 Chr.5: 76835329-76835329 on Build GRCh38
rs1056079982 Chr.5: 76834974-76834974 on Build GRCh38
rs982940802 Chr.5: 76835332-76835332 on Build GRCh38
rs754913009 Chr.5: 76835005-76835005 on Build GRCh38
rs534549684 Chr.5: 76834875-76834878 on Build GRCh38
rs1009794724 Chr.5: 76835335-76835335 on Build GRCh38
rs968054502 Chr.5: 76835186-76835186 on Build GRCh38
rs1034693299 Chr.5: 76835295-76835295 on Build GRCh38
rs766193566 Chr.5: 76834942-76834942 on Build GRCh38
rs545102761 Chr.5: 76835243-76835243 on Build GRCh38
rs997026403 Chr.5: 76835090-76835090 on Build GRCh38
rs958553196 Chr.5: 76835014-76835014 on Build GRCh38
rs1051287508 Chr.5: 76835246-76835246 on Build GRCh38
rs1046709569 Chr.5: 76835011-76835011 on Build GRCh38
rs567614928 Chr.5: 76835048-76835048 on Build GRCh38
rs892581528 Chr.5: 76834921-76834921 on Build GRCh38
rs979540907 Chr.5: 76834931-76834931 on Build GRCh38

View Full Product Details