Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 76958649-76959161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRHBP
Entrez Gene ID
1393
Gene Name
corticotropin releasing hormone binding protein
Gene Aliases
CRF-BP, CRFBP
UniGene
Hs.115617
Gene Chromosome Location
Chr.5: 76952713-76981143 on Build GRCh38
Transcripts
NM_001882.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1695884 c.577A>T p.K193* Chr.5: 76958773-76958773 on Build GRCh38
1620560 c.615C>A p.C205* Chr.5: 76958811-76958811 on Build GRCh38
5459697 c.598C>G p.H200D Chr.5: 76958794-76958794 on Build GRCh38
6230904 c.658C>A p.L220I Chr.5: 76958854-76958854 on Build GRCh38
1438663 c.608G>A p.R203Q Chr.5: 76958804-76958804 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777836843 Chr.5: 76958722-76958722 on Build GRCh38
rs889280452 Chr.5: 76958805-76958805 on Build GRCh38
rs771236353 Chr.5: 76959057-76959057 on Build GRCh38
rs940394481 Chr.5: 76959069-76959069 on Build GRCh38
rs770775409 Chr.5: 76958727-76958727 on Build GRCh38
rs775127860 Chr.5: 76958868-76958868 on Build GRCh38
rs767954351 Chr.5: 76958912-76958912 on Build GRCh38
rs571544189 Chr.5: 76958946-76958946 on Build GRCh38
rs144298478 Chr.5: 76958808-76958808 on Build GRCh38
rs202024228 Chr.5: 76958715-76958715 on Build GRCh38
rs918299040 Chr.5: 76958723-76958723 on Build GRCh38
rs776100344 Chr.5: 76958728-76958728 on Build GRCh38
rs759218687 Chr.5: 76958733-76958733 on Build GRCh38
rs563306152 Chr.5: 76958725-76958725 on Build GRCh38
rs904380054 Chr.5: 76959020-76959020 on Build GRCh38
rs769431244 Chr.5: 76958861-76958861 on Build GRCh38
rs780485429 Chr.5: 76958788-76958788 on Build GRCh38
rs548963823 Chr.5: 76958838-76958838 on Build GRCh38
rs1052442111 Chr.5: 76958942-76958942 on Build GRCh38
rs762348146 Chr.5: 76958911-76958911 on Build GRCh38
rs753968029 Chr.5: 76958798-76958798 on Build GRCh38
rs778039856 Chr.5: 76959039-76959039 on Build GRCh38
rs879146853 Chr.5: 76958702-76958702 on Build GRCh38
rs199529245 Chr.5: 76958714-76958714 on Build GRCh38
rs201530535 Chr.5: 76958804-76958804 on Build GRCh38
rs752119854 Chr.5: 76958764-76958764 on Build GRCh38
rs571043986 Chr.5: 76958987-76958987 on Build GRCh38
rs146589192 Chr.5: 76958849-76958849 on Build GRCh38
rs542787214 Chr.5: 76958994-76958994 on Build GRCh38
rs747352315 Chr.5: 76959040-76959040 on Build GRCh38
rs765392092 Chr.5: 76958950-76958950 on Build GRCh38
rs76328154 Chr.5: 76959075-76959075 on Build GRCh38
rs370676315 Chr.5: 76958915-76958915 on Build GRCh38
rs200414648 Chr.5: 76958751-76958751 on Build GRCh38
rs550937164 Chr.5: 76958931-76958931 on Build GRCh38
rs982316447 Chr.5: 76958995-76958995 on Build GRCh38
rs1018988028 Chr.5: 76958832-76958832 on Build GRCh38
rs780246392 Chr.5: 76958713-76958713 on Build GRCh38
rs140991323 Chr.5: 76958988-76958988 on Build GRCh38
rs529210191 Chr.5: 76958767-76958767 on Build GRCh38
rs374135210 Chr.5: 76958921-76958921 on Build GRCh38
rs768233885 Chr.5: 76958875-76958875 on Build GRCh38
rs1011008963 Chr.5: 76958983-76958983 on Build GRCh38
rs745712246 Chr.5: 76958860-76958860 on Build GRCh38
rs773694603 Chr.5: 76958914-76958914 on Build GRCh38
rs989400928 Chr.5: 76958923-76958923 on Build GRCh38
rs748736594 Chr.5: 76958869-76958869 on Build GRCh38
rs910062264 Chr.5: 76958841-76958841 on Build GRCh38
rs370602224 Chr.5: 76958699-76958699 on Build GRCh38
rs754964897 Chr.5: 76958930-76958930 on Build GRCh38
rs764439387 Chr.5: 76958752-76958752 on Build GRCh38
rs376822831 Chr.5: 76958902-76958902 on Build GRCh38
rs746306127 Chr.5: 76959097-76959097 on Build GRCh38
rs756379046 Chr.5: 76958782-76958782 on Build GRCh38
rs555831463 Chr.5: 76958916-76958916 on Build GRCh38

View Full Product Details