Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78779509-78780002 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
UniGene
Hs.149103
Gene Chromosome Location
Chr.5: 78777209-78986534 on Build GRCh38
Transcripts
NM_000046.3 XM_011543390.1

SNP Detail

SNP ID Chromosome Location
rs1009076221 Chr.5: 78779622-78779622 on Build GRCh38
rs973438440 Chr.5: 78779846-78779846 on Build GRCh38
rs940643881 Chr.5: 78779809-78779809 on Build GRCh38
rs569587593 Chr.5: 78779811-78779811 on Build GRCh38
rs962569360 Chr.5: 78779836-78779836 on Build GRCh38
rs1012424576 Chr.5: 78779553-78779553 on Build GRCh38
rs907903168 Chr.5: 78779574-78779574 on Build GRCh38
rs749372784 Chr.5: 78779813-78779813 on Build GRCh38
rs1012669209 Chr.5: 78779778-78779778 on Build GRCh38
rs773033700 Chr.5: 78779962-78779962 on Build GRCh38
rs1041918987 Chr.5: 78779935-78779935 on Build GRCh38
rs866877062 Chr.5: 78779830-78779830 on Build GRCh38
rs757857543 Chr.5: 78779779-78779779 on Build GRCh38
rs920646570 Chr.5: 78779833-78779833 on Build GRCh38
rs560652209 Chr.5: 78779555-78779555 on Build GRCh38
rs899235523 Chr.5: 78779593-78779593 on Build GRCh38
rs1020507735 Chr.5: 78779645-78779645 on Build GRCh38
rs534480571 Chr.5: 78779762-78779762 on Build GRCh38
rs890544960 Chr.5: 78779892-78779892 on Build GRCh38
rs1079397 Chr.5: 78779901-78779901 on Build GRCh38
rs996301793 Chr.5: 78779597-78779597 on Build GRCh38
rs898209203 Chr.5: 78779768-78779768 on Build GRCh38
rs151197584 Chr.5: 78779942-78779942 on Build GRCh38
rs971005935 Chr.5: 78779563-78779563 on Build GRCh38
rs1000473258 Chr.5: 78779943-78779943 on Build GRCh38
rs977184196 Chr.5: 78779971-78779971 on Build GRCh38
rs999866691 Chr.5: 78779719-78779719 on Build GRCh38
rs970496566 Chr.5: 78779763-78779763 on Build GRCh38
rs532783256 Chr.5: 78779570-78779570 on Build GRCh38
rs886060796 Chr.5: 78779789-78779789 on Build GRCh38
rs868866414 Chr.5: 78779852-78779852 on Build GRCh38
rs535536121 Chr.5: 78779589-78779589 on Build GRCh38
rs982340985 Chr.5: 78779568-78779568 on Build GRCh38
rs1041604176 Chr.5: 78779663-78779663 on Build GRCh38
rs907768933 Chr.5: 78779794-78779794 on Build GRCh38
rs1050497091 Chr.5: 78779857-78779857 on Build GRCh38
rs777160780 Chr.5: 78779587-78779587 on Build GRCh38
rs886060794 Chr.5: 78779604-78779604 on Build GRCh38
rs894258651 Chr.5: 78779757-78779757 on Build GRCh38
rs1032035805 Chr.5: 78779722-78779722 on Build GRCh38
rs886060795 Chr.5: 78779778-78779778 on Build GRCh38
rs186035080 Chr.5: 78779609-78779609 on Build GRCh38
rs953530086 Chr.5: 78779902-78779902 on Build GRCh38
rs967638908 Chr.5: 78779673-78779673 on Build GRCh38
rs1051032311 Chr.5: 78779606-78779606 on Build GRCh38
rs565551823 Chr.5: 78779611-78779611 on Build GRCh38
rs932052233 Chr.5: 78779845-78779845 on Build GRCh38
rs552876911 Chr.5: 78779750-78779750 on Build GRCh38
rs771930799 Chr.5: 78779677-78779677 on Build GRCh38
rs958008157 Chr.5: 78779749-78779749 on Build GRCh38
rs1009043206 Chr.5: 78779930-78779930 on Build GRCh38

View Full Product Details