Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 78781763-78782262 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSB
Entrez Gene ID
411
Gene Name
arylsulfatase B
Gene Aliases
ASB, G4S, MPS6
UniGene
Hs.149103
Gene Chromosome Location
Chr.5: 78777209-78986534 on Build GRCh38
Transcripts
NM_000046.3 XM_011543390.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618021 c.1302A>G p.R434R Chr.5: 78781886-78781886 on Build GRCh38
3856414 c.1291G>A p.A431T Chr.5: 78781897-78781897 on Build GRCh38
5709753 c.1285G>A p.V429I Chr.5: 78781903-78781903 on Build GRCh38
3715269 c.1326G>A p.T442T Chr.5: 78781862-78781862 on Build GRCh38
5474843 c.1263A>C p.E421D Chr.5: 78781925-78781925 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370490378 Chr.5: 78781933-78781933 on Build GRCh38
rs780926335 Chr.5: 78781941-78781941 on Build GRCh38
rs753899382 Chr.5: 78781880-78781880 on Build GRCh38
rs1044786199 Chr.5: 78781938-78781938 on Build GRCh38
rs985765448 Chr.5: 78782103-78782103 on Build GRCh38
rs772536800 Chr.5: 78782147-78782148 on Build GRCh38
rs763353963 Chr.5: 78781836-78781836 on Build GRCh38
rs746743704 Chr.5: 78781805-78781805 on Build GRCh38
rs764623204 Chr.5: 78781827-78781827 on Build GRCh38
rs573179858 Chr.5: 78782170-78782170 on Build GRCh38
rs574407307 Chr.5: 78782072-78782072 on Build GRCh38
rs755037325 Chr.5: 78781991-78781991 on Build GRCh38
rs770690800 Chr.5: 78781906-78781906 on Build GRCh38
rs900835890 Chr.5: 78782206-78782206 on Build GRCh38
rs914447734 Chr.5: 78782230-78782230 on Build GRCh38
rs774947204 Chr.5: 78781960-78781960 on Build GRCh38
rs759237377 Chr.5: 78781821-78781821 on Build GRCh38
rs551775820 Chr.5: 78781798-78781798 on Build GRCh38
rs769394809 Chr.5: 78782028-78782028 on Build GRCh38
rs751469522 Chr.5: 78782019-78782019 on Build GRCh38
rs544640945 Chr.5: 78782224-78782224 on Build GRCh38
rs1030946663 Chr.5: 78781859-78781859 on Build GRCh38
rs1017886963 Chr.5: 78781822-78781822 on Build GRCh38
rs965138684 Chr.5: 78781834-78781834 on Build GRCh38
rs554258996 Chr.5: 78781956-78781956 on Build GRCh38
rs745504153 Chr.5: 78781917-78781917 on Build GRCh38
rs918640798 Chr.5: 78782046-78782046 on Build GRCh38
rs370678227 Chr.5: 78781818-78781818 on Build GRCh38
rs1048669157 Chr.5: 78782104-78782104 on Build GRCh38
rs376233975 Chr.5: 78781851-78781851 on Build GRCh38
rs368770141 Chr.5: 78781986-78781986 on Build GRCh38
rs748129076 Chr.5: 78781845-78781845 on Build GRCh38
rs773151566 Chr.5: 78781803-78781803 on Build GRCh38
rs553223495 Chr.5: 78782140-78782140 on Build GRCh38
rs927277945 Chr.5: 78782005-78782005 on Build GRCh38
rs536564629 Chr.5: 78782099-78782099 on Build GRCh38
rs975766357 Chr.5: 78782178-78782178 on Build GRCh38
rs752116762 Chr.5: 78781840-78781840 on Build GRCh38
rs7721866 Chr.5: 78782006-78782006 on Build GRCh38
rs147288610 Chr.5: 78782229-78782229 on Build GRCh38
rs914833400 Chr.5: 78781942-78781942 on Build GRCh38
rs762225092 Chr.5: 78781848-78781848 on Build GRCh38
rs767959488 Chr.5: 78781972-78781972 on Build GRCh38
rs886060797 Chr.5: 78781957-78781957 on Build GRCh38
rs537543103 Chr.5: 78781862-78781862 on Build GRCh38
rs756933794 Chr.5: 78781897-78781897 on Build GRCh38
rs761082363 Chr.5: 78782177-78782177 on Build GRCh38
rs755021848 Chr.5: 78781882-78781882 on Build GRCh38
rs994714299 Chr.5: 78782062-78782062 on Build GRCh38
rs781006512 Chr.5: 78781909-78781909 on Build GRCh38
rs775132181 Chr.5: 78781828-78781828 on Build GRCh38
rs953074616 Chr.5: 78782075-78782075 on Build GRCh38
rs764604137 Chr.5: 78781837-78781837 on Build GRCh38
rs760962960 Chr.5: 78781978-78781978 on Build GRCh38
rs768012515 Chr.5: 78781850-78781850 on Build GRCh38
rs118203941 Chr.5: 78781974-78781974 on Build GRCh38
rs746972113 Chr.5: 78781903-78781903 on Build GRCh38
rs777651865 Chr.5: 78781884-78781884 on Build GRCh38
rs897262383 Chr.5: 78782044-78782044 on Build GRCh38
rs766561160 Chr.5: 78781982-78781982 on Build GRCh38
rs1002837181 Chr.5: 78781996-78781996 on Build GRCh38
rs964336934 Chr.5: 78782171-78782171 on Build GRCh38
rs202130108 Chr.5: 78781819-78781819 on Build GRCh38
rs955846473 Chr.5: 78782225-78782225 on Build GRCh38
rs762454192 Chr.5: 78781964-78781964 on Build GRCh38
rs762458435 Chr.5: 78781935-78781935 on Build GRCh38
rs956246041 Chr.5: 78781868-78781868 on Build GRCh38
rs769542268 Chr.5: 78781930-78781930 on Build GRCh38

View Full Product Details