Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79029701-79030210 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMGDH
Entrez Gene ID
29958
Gene Name
dimethylglycine dehydrogenase
Gene Aliases
DMGDHD, ME2GLYDH
UniGene
Hs.655653
Gene Chromosome Location
Chr.5: 78997564-79069674 on Build GRCh38
Transcripts
NM_013391.3 NR_104002.1 NR_104003.1 XM_006714597.2 XM_011543354.2 XM_011543355.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1250253 c.1707C>G p.H569Q Chr.5: 79030011-79030011 on Build GRCh38
3618029 c.1164G>A p.G388G Chr.5: 79029951-79029951 on Build GRCh38
6372002 c.1714A>G p.T572A Chr.5: 79030004-79030004 on Build GRCh38
6104717 c.1084G>T p.G362C Chr.5: 79030031-79030031 on Build GRCh38
3429669 c.1171C>A p.L391I Chr.5: 79029944-79029944 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760774513 Chr.5: 79029939-79029939 on Build GRCh38
rs560813383 Chr.5: 79030041-79030041 on Build GRCh38
rs960590999 Chr.5: 79029909-79029909 on Build GRCh38
rs763002672 Chr.5: 79030063-79030063 on Build GRCh38
rs1034957959 Chr.5: 79029900-79029900 on Build GRCh38
rs751155711 Chr.5: 79029971-79029971 on Build GRCh38
rs748526327 Chr.5: 79030024-79030024 on Build GRCh38
rs189581496 Chr.5: 79029986-79029986 on Build GRCh38
rs749569950 Chr.5: 79029913-79029913 on Build GRCh38
rs892247705 Chr.5: 79029823-79029823 on Build GRCh38
rs568535560 Chr.5: 79030144-79030144 on Build GRCh38
rs41272268 Chr.5: 79030056-79030056 on Build GRCh38
rs762091133 Chr.5: 79029935-79029935 on Build GRCh38
rs952165813 Chr.5: 79029950-79029950 on Build GRCh38
rs774408032 Chr.5: 79029916-79029916 on Build GRCh38
rs566725933 Chr.5: 79029755-79029755 on Build GRCh38
rs538929652 Chr.5: 79029772-79029772 on Build GRCh38
rs762171009 Chr.5: 79029778-79029778 on Build GRCh38
rs902063673 Chr.5: 79029736-79029736 on Build GRCh38
rs755295841 Chr.5: 79030016-79030016 on Build GRCh38
rs868573192 Chr.5: 79029979-79029979 on Build GRCh38
rs763948105 Chr.5: 79030070-79030070 on Build GRCh38
rs780645948 Chr.5: 79029899-79029899 on Build GRCh38
rs537081787 Chr.5: 79029817-79029817 on Build GRCh38
rs1031480658 Chr.5: 79029748-79029748 on Build GRCh38
rs946325261 Chr.5: 79030176-79030176 on Build GRCh38
rs780406415 Chr.5: 79029992-79029992 on Build GRCh38
rs773444465 Chr.5: 79030037-79030037 on Build GRCh38
rs764014496 Chr.5: 79029960-79029960 on Build GRCh38
rs747068071 Chr.5: 79030039-79030039 on Build GRCh38
rs752861815 Chr.5: 79029930-79029930 on Build GRCh38
rs939057131 Chr.5: 79029761-79029761 on Build GRCh38
rs774219092 Chr.5: 79030071-79030071 on Build GRCh38
rs868321745 Chr.5: 79029951-79029951 on Build GRCh38
rs777266909 Chr.5: 79029855-79029855 on Build GRCh38
rs971704483 Chr.5: 79029801-79029801 on Build GRCh38
rs753784796 Chr.5: 79030071-79030072 on Build GRCh38
rs969281459 Chr.5: 79030166-79030166 on Build GRCh38
rs780904418 Chr.5: 79029891-79029891 on Build GRCh38
rs964919748 Chr.5: 79030010-79030010 on Build GRCh38
rs771080265 Chr.5: 79030046-79030046 on Build GRCh38
rs980574208 Chr.5: 79029738-79029738 on Build GRCh38
rs114971596 Chr.5: 79030163-79030163 on Build GRCh38
rs201183749 Chr.5: 79029872-79029872 on Build GRCh38
rs1031020884 Chr.5: 79030107-79030107 on Build GRCh38
rs923570828 Chr.5: 79030087-79030087 on Build GRCh38
rs762622638 Chr.5: 79029952-79029952 on Build GRCh38
rs7731290 Chr.5: 79029767-79029767 on Build GRCh38
rs1052082429 Chr.5: 79029769-79029769 on Build GRCh38
rs927610470 Chr.5: 79029758-79029759 on Build GRCh38
rs764148418 Chr.5: 79030051-79030055 on Build GRCh38
rs892363922 Chr.5: 79030123-79030123 on Build GRCh38
rs200459665 Chr.5: 79029894-79029894 on Build GRCh38
rs769116632 Chr.5: 79029914-79029914 on Build GRCh38
rs773219098 Chr.5: 79029938-79029938 on Build GRCh38
rs772215706 Chr.5: 79030031-79030031 on Build GRCh38
rs910588331 Chr.5: 79030001-79030001 on Build GRCh38
rs540048739 Chr.5: 79029819-79029819 on Build GRCh38
rs767278482 Chr.5: 79029980-79029980 on Build GRCh38
rs913565691 Chr.5: 79030172-79030172 on Build GRCh38
rs933662822 Chr.5: 79029742-79029742 on Build GRCh38
rs772125882 Chr.5: 79029937-79029937 on Build GRCh38
rs756908579 Chr.5: 79029978-79029978 on Build GRCh38
rs918806785 Chr.5: 79029826-79029826 on Build GRCh38
rs558617324 Chr.5: 79029849-79029849 on Build GRCh38
rs1010846809 Chr.5: 79030158-79030158 on Build GRCh38
rs779394442 Chr.5: 79030021-79030021 on Build GRCh38
rs1040637409 Chr.5: 79029730-79029730 on Build GRCh38
rs753917594 Chr.5: 79029769-79029769 on Build GRCh38
rs772315241 Chr.5: 79029728-79029728 on Build GRCh38
rs751263321 Chr.5: 79029864-79029864 on Build GRCh38
rs139749124 Chr.5: 79029941-79029941 on Build GRCh38
rs74619873 Chr.5: 79029964-79029964 on Build GRCh38
rs552947242 Chr.5: 79030173-79030173 on Build GRCh38
rs111601498 Chr.5: 79029929-79029929 on Build GRCh38
rs755711429 Chr.5: 79029990-79029990 on Build GRCh38
rs775545785 Chr.5: 79029943-79029943 on Build GRCh38
rs1010650749 Chr.5: 79029835-79029835 on Build GRCh38
rs745380831 Chr.5: 79029892-79029892 on Build GRCh38
rs998152299 Chr.5: 79030096-79030096 on Build GRCh38
rs756142928 Chr.5: 79030016-79030017 on Build GRCh38

View Full Product Details