Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79069422-79069939 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT2
Entrez Gene ID
23743
Gene Name
betaine--homocysteine S-methyltransferase 2
Gene Aliases
-
UniGene
Hs.114172
Gene Chromosome Location
Chr.5: 79069724-79090074 on Build GRCh38
Transcripts
NM_001178005.1 NM_017614.4 NM_013391.3 NR_104002.1 NR_104003.1 XM_006714597.2 XM_011543354.2 XM_011543355.2

Gene Symbol
DMGDH
Entrez Gene ID
29958
Gene Name
dimethylglycine dehydrogenase
Gene Aliases
DMGDHD, ME2GLYDH
UniGene
Hs.655653
Gene Chromosome Location
Chr.5: 78997564-79069674 on Build GRCh38
Transcripts
NM_001178005.1 NM_017614.4 NM_013391.3 NR_104002.1 NR_104003.1 XM_006714597.2 XM_011543354.2 XM_011543355.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6413069 c.91G>T p.G31C Chr.5: 79069530-79069530 on Build GRCh38
6416497 c.16G>T p.A6S Chr.5: 79069605-79069605 on Build GRCh38
6373534 c.2T>G p.M1R Chr.5: 79069784-79069784 on Build GRCh38
289913 c.9T>C p.P3P Chr.5: 79069791-79069791 on Build GRCh38
5725400 c.62G>A p.G21D Chr.5: 79069559-79069559 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs13189440 Chr.5: 79069793-79069793 on Build GRCh38
rs556440039 Chr.5: 79069543-79069543 on Build GRCh38
rs751895011 Chr.5: 79069559-79069559 on Build GRCh38
rs997539459 Chr.5: 79069700-79069700 on Build GRCh38
rs1051573999 Chr.5: 79069764-79069764 on Build GRCh38
rs913207363 Chr.5: 79069702-79069702 on Build GRCh38
rs376333321 Chr.5: 79069832-79069832 on Build GRCh38
rs918048623 Chr.5: 79069552-79069552 on Build GRCh38
rs1019896974 Chr.5: 79069554-79069554 on Build GRCh38
rs750536578 Chr.5: 79069802-79069802 on Build GRCh38
rs375741505 Chr.5: 79069817-79069817 on Build GRCh38
rs908627183 Chr.5: 79069905-79069905 on Build GRCh38
rs1043158306 Chr.5: 79069805-79069805 on Build GRCh38
rs1028663524 Chr.5: 79069525-79069525 on Build GRCh38
rs991330843 Chr.5: 79069576-79069576 on Build GRCh38
rs371478130 Chr.5: 79069853-79069853 on Build GRCh38
rs968357302 Chr.5: 79069520-79069520 on Build GRCh38
rs900352904 Chr.5: 79069604-79069604 on Build GRCh38
rs540788847 Chr.5: 79069846-79069846 on Build GRCh38
rs1034000532 Chr.5: 79069916-79069916 on Build GRCh38
rs13188340 Chr.5: 79069777-79069777 on Build GRCh38
rs929359642 Chr.5: 79069552-79069552 on Build GRCh38
rs775664830 Chr.5: 79069657-79069657 on Build GRCh38
rs1020012218 Chr.5: 79069737-79069737 on Build GRCh38
rs954452575 Chr.5: 79069536-79069536 on Build GRCh38
rs891602273 Chr.5: 79069797-79069797 on Build GRCh38
rs564075355 Chr.5: 79069862-79069862 on Build GRCh38
rs891569770 Chr.5: 79069618-79069618 on Build GRCh38
rs891324403 Chr.5: 79069719-79069719 on Build GRCh38
rs371850102 Chr.5: 79069483-79069483 on Build GRCh38
rs895911393 Chr.5: 79069458-79069458 on Build GRCh38
rs992248968 Chr.5: 79069548-79069548 on Build GRCh38
rs978459630 Chr.5: 79069752-79069752 on Build GRCh38
rs56774046 Chr.5: 79069518-79069518 on Build GRCh38
rs900321836 Chr.5: 79069444-79069444 on Build GRCh38
rs1038986625 Chr.5: 79069673-79069673 on Build GRCh38
rs904194651 Chr.5: 79069840-79069840 on Build GRCh38
rs761969540 Chr.5: 79069759-79069759 on Build GRCh38
rs941992160 Chr.5: 79069720-79069720 on Build GRCh38
rs764191756 Chr.5: 79069660-79069660 on Build GRCh38
rs967412458 Chr.5: 79069751-79069751 on Build GRCh38
rs899976135 Chr.5: 79069510-79069510 on Build GRCh38
rs555715126 Chr.5: 79069644-79069644 on Build GRCh38
rs770498546 Chr.5: 79069470-79069470 on Build GRCh38
rs937136493 Chr.5: 79069574-79069574 on Build GRCh38
rs1034310929 Chr.5: 79069686-79069686 on Build GRCh38
rs941978077 Chr.5: 79069583-79069583 on Build GRCh38
rs373250750 Chr.5: 79069825-79069825 on Build GRCh38
rs1021303526 Chr.5: 79069653-79069653 on Build GRCh38
rs1047092441 Chr.5: 79069475-79069475 on Build GRCh38
rs13188159 Chr.5: 79069671-79069671 on Build GRCh38
rs992353515 Chr.5: 79069696-79069696 on Build GRCh38
rs13188199 Chr.5: 79069730-79069730 on Build GRCh38
rs13188201 Chr.5: 79069740-79069740 on Build GRCh38
rs57739060 Chr.5: 79069632-79069632 on Build GRCh38
rs757204104 Chr.5: 79069733-79069767 on Build GRCh38
rs1039325694 Chr.5: 79069587-79069587 on Build GRCh38
rs917128060 Chr.5: 79069607-79069607 on Build GRCh38
rs1056244137 Chr.5: 79069456-79069456 on Build GRCh38
rs777289278 Chr.5: 79069850-79069850 on Build GRCh38
rs1051543845 Chr.5: 79069610-79069610 on Build GRCh38
rs950792876 Chr.5: 79069711-79069711 on Build GRCh38
rs370147287 Chr.5: 79069807-79069807 on Build GRCh38
rs1017199728 Chr.5: 79069506-79069506 on Build GRCh38
rs1010004996 Chr.5: 79069799-79069799 on Build GRCh38
rs763422986 Chr.5: 79069659-79069659 on Build GRCh38
rs13189433 Chr.5: 79069776-79069776 on Build GRCh38
rs997339456 Chr.5: 79069605-79069605 on Build GRCh38
rs941389029 Chr.5: 79069666-79069666 on Build GRCh38
rs764366356 Chr.5: 79069497-79069497 on Build GRCh38
rs576372257 Chr.5: 79069562-79069562 on Build GRCh38
rs765141747 Chr.5: 79069776-79069777 on Build GRCh38
rs369709170 Chr.5: 79069522-79069522 on Build GRCh38
rs59310526 Chr.5: 79069449-79069449 on Build GRCh38
rs925897230 Chr.5: 79069570-79069570 on Build GRCh38
rs572083610 Chr.5: 79069733-79069733 on Build GRCh38
rs113466769 Chr.5: 79069772-79069772 on Build GRCh38
rs13171089 Chr.5: 79069819-79069819 on Build GRCh38
rs983919203 Chr.5: 79069717-79069717 on Build GRCh38
rs950095698 Chr.5: 79069613-79069613 on Build GRCh38
rs900087609 Chr.5: 79069683-79069683 on Build GRCh38
rs1039027963 Chr.5: 79069721-79069721 on Build GRCh38
rs751894373 Chr.5: 79069754-79069754 on Build GRCh38
rs13188470 Chr.5: 79069875-79069875 on Build GRCh38
rs959642195 Chr.5: 79069917-79069917 on Build GRCh38
rs766601673 Chr.5: 79069629-79069629 on Build GRCh38
rs925750436 Chr.5: 79069447-79069447 on Build GRCh38
rs1029781679 Chr.5: 79069715-79069715 on Build GRCh38
rs926828033 Chr.5: 79069531-79069531 on Build GRCh38
rs868724472 Chr.5: 79069868-79069868 on Build GRCh38
rs59281454 Chr.5: 79069887-79069887 on Build GRCh38
rs776129607 Chr.5: 79069623-79069623 on Build GRCh38
rs949877429 Chr.5: 79069841-79069841 on Build GRCh38
rs767648371 Chr.5: 79069781-79069781 on Build GRCh38
rs959601532 Chr.5: 79069688-79069688 on Build GRCh38
rs979065177 Chr.5: 79069568-79069568 on Build GRCh38

View Full Product Details