Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79088224-79088742 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT2
Entrez Gene ID
23743
Gene Name
betaine--homocysteine S-methyltransferase 2
Gene Aliases
-
UniGene
Hs.114172
Gene Chromosome Location
Chr.5: 79069724-79090074 on Build GRCh38
Transcripts
NM_001178005.1 NM_017614.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5061216 c.1036C>A p.L346M Chr.5: 79088518-79088518 on Build GRCh38
7296140 c.1087T>G p.F363V Chr.5: 79088569-79088569 on Build GRCh38
3618053 c.1086C>T p.D362D Chr.5: 79088568-79088568 on Build GRCh38
3828461 c.1066C>G p.P356A Chr.5: 79088548-79088548 on Build GRCh38
309476 c.1089C>T p.F363F Chr.5: 79088571-79088571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750798062 Chr.5: 79088459-79088462 on Build GRCh38
rs746838767 Chr.5: 79088498-79088498 on Build GRCh38
rs202040828 Chr.5: 79088358-79088359 on Build GRCh38
rs888076899 Chr.5: 79088343-79088343 on Build GRCh38
rs753981682 Chr.5: 79088553-79088553 on Build GRCh38
rs780186405 Chr.5: 79088460-79088460 on Build GRCh38
rs756254218 Chr.5: 79088449-79088449 on Build GRCh38
rs946548855 Chr.5: 79088699-79088699 on Build GRCh38
rs1029596385 Chr.5: 79088676-79088676 on Build GRCh38
rs560000225 Chr.5: 79088427-79088427 on Build GRCh38
rs777812797 Chr.5: 79088554-79088554 on Build GRCh38
rs368310862 Chr.5: 79088537-79088537 on Build GRCh38
rs938112111 Chr.5: 79088264-79088264 on Build GRCh38
rs368539921 Chr.5: 79088473-79088473 on Build GRCh38
rs745936914 Chr.5: 79088447-79088447 on Build GRCh38
rs574045062 Chr.5: 79088298-79088298 on Build GRCh38
rs984124070 Chr.5: 79088309-79088309 on Build GRCh38
rs56053829 Chr.5: 79088500-79088500 on Build GRCh38
rs780970063 Chr.5: 79088568-79088571 on Build GRCh38
rs1056446074 Chr.5: 79088276-79088276 on Build GRCh38
rs772067632 Chr.5: 79088491-79088491 on Build GRCh38
rs573161305 Chr.5: 79088379-79088398 on Build GRCh38
rs372951107 Chr.5: 79088357-79088358 on Build GRCh38
rs746347820 Chr.5: 79088592-79088592 on Build GRCh38
rs148438687 Chr.5: 79088388-79088389 on Build GRCh38
rs551084921 Chr.5: 79088643-79088643 on Build GRCh38
rs72306524 Chr.5: 79088366-79088367 on Build GRCh38
rs148183648 Chr.5: 79088540-79088540 on Build GRCh38
rs746785482 Chr.5: 79088556-79088556 on Build GRCh38
rs769033413 Chr.5: 79088621-79088621 on Build GRCh38
rs373665798 Chr.5: 79088614-79088614 on Build GRCh38
rs138961781 Chr.5: 79088501-79088501 on Build GRCh38
rs536623621 Chr.5: 79088691-79088691 on Build GRCh38
rs1006377965 Chr.5: 79088399-79088399 on Build GRCh38
rs549037477 Chr.5: 79088579-79088579 on Build GRCh38
rs751330926 Chr.5: 79088476-79088479 on Build GRCh38
rs926697847 Chr.5: 79088260-79088260 on Build GRCh38
rs763331564 Chr.5: 79088519-79088519 on Build GRCh38
rs899558993 Chr.5: 79088423-79088423 on Build GRCh38
rs55861901 Chr.5: 79088715-79088715 on Build GRCh38
rs762892535 Chr.5: 79088457-79088457 on Build GRCh38
rs775091606 Chr.5: 79088518-79088518 on Build GRCh38
rs546218419 Chr.5: 79088403-79088403 on Build GRCh38
rs757526868 Chr.5: 79088398-79088399 on Build GRCh38
rs770913184 Chr.5: 79088583-79088583 on Build GRCh38
rs375611100 Chr.5: 79088505-79088505 on Build GRCh38
rs887200956 Chr.5: 79088605-79088605 on Build GRCh38
rs774832651 Chr.5: 79088657-79088657 on Build GRCh38
rs879167468 Chr.5: 79088359-79088360 on Build GRCh38
rs539862060 Chr.5: 79088303-79088303 on Build GRCh38
rs774836253 Chr.5: 79088609-79088609 on Build GRCh38
rs764769227 Chr.5: 79088508-79088508 on Build GRCh38
rs779948612 Chr.5: 79088552-79088552 on Build GRCh38
rs188348653 Chr.5: 79088293-79088293 on Build GRCh38
rs975789694 Chr.5: 79088664-79088664 on Build GRCh38
rs979173745 Chr.5: 79088717-79088717 on Build GRCh38
rs933851959 Chr.5: 79088458-79088462 on Build GRCh38
rs60166823 Chr.5: 79088608-79088608 on Build GRCh38
rs543984298 Chr.5: 79088300-79088300 on Build GRCh38
rs369874568 Chr.5: 79088589-79088589 on Build GRCh38
rs193302760 Chr.5: 79088330-79088330 on Build GRCh38
rs922494590 Chr.5: 79088402-79088402 on Build GRCh38
rs762225803 Chr.5: 79088533-79088533 on Build GRCh38
rs372903910 Chr.5: 79088317-79088317 on Build GRCh38
rs996599140 Chr.5: 79088432-79088432 on Build GRCh38
rs4020529 Chr.5: 79088395-79088396 on Build GRCh38
rs763691230 Chr.5: 79088698-79088698 on Build GRCh38
rs567806868 Chr.5: 79088687-79088687 on Build GRCh38
rs745456442 Chr.5: 79088593-79088593 on Build GRCh38
rs750578244 Chr.5: 79088539-79088539 on Build GRCh38
rs975396742 Chr.5: 79088386-79088386 on Build GRCh38
rs149000124 Chr.5: 79088527-79088527 on Build GRCh38
rs532321076 Chr.5: 79088522-79088522 on Build GRCh38
rs372276376 Chr.5: 79088497-79088497 on Build GRCh38
rs143035984 Chr.5: 79088537-79088537 on Build GRCh38
rs762576972 Chr.5: 79088661-79088661 on Build GRCh38

View Full Product Details