Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79115671-79116211 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT
Entrez Gene ID
635
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
BHMT1, HEL-S-61p
UniGene
Hs.80756
Gene Chromosome Location
Chr.5: 79111781-79132290 on Build GRCh38
Transcripts
NM_001713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123975 c.90C>G p.V30V Chr.5: 79115823-79115823 on Build GRCh38
5029803 c.80G>A p.G27E Chr.5: 79115813-79115813 on Build GRCh38
5486502 c.36C>T p.G12G Chr.5: 79115769-79115769 on Build GRCh38
1070387 c.98T>C p.L33P Chr.5: 79115831-79115831 on Build GRCh38
483072 c.106A>G p.R36G Chr.5: 79115839-79115839 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1027559434 Chr.5: 79116116-79116116 on Build GRCh38
rs542489636 Chr.5: 79115741-79115741 on Build GRCh38
rs372541519 Chr.5: 79115780-79115780 on Build GRCh38
rs773765369 Chr.5: 79115797-79115797 on Build GRCh38
rs763364284 Chr.5: 79115862-79115862 on Build GRCh38
rs751917997 Chr.5: 79116097-79116097 on Build GRCh38
rs777010353 Chr.5: 79115906-79115906 on Build GRCh38
rs778585214 Chr.5: 79115855-79115855 on Build GRCh38
rs1003573942 Chr.5: 79115715-79115715 on Build GRCh38
rs564379184 Chr.5: 79115923-79115923 on Build GRCh38
rs376693924 Chr.5: 79115920-79115920 on Build GRCh38
rs534858860 Chr.5: 79116095-79116095 on Build GRCh38
rs985513424 Chr.5: 79115783-79115783 on Build GRCh38
rs1019121930 Chr.5: 79115917-79115917 on Build GRCh38
rs939191410 Chr.5: 79116072-79116072 on Build GRCh38
rs757968469 Chr.5: 79115874-79115874 on Build GRCh38
rs376169567 Chr.5: 79115936-79115936 on Build GRCh38
rs766268143 Chr.5: 79115929-79115929 on Build GRCh38
rs200127835 Chr.5: 79115942-79115942 on Build GRCh38
rs1036833147 Chr.5: 79116077-79116077 on Build GRCh38
rs953600240 Chr.5: 79115861-79115861 on Build GRCh38
rs549684616 Chr.5: 79116063-79116063 on Build GRCh38
rs756007113 Chr.5: 79115739-79115739 on Build GRCh38
rs775693955 Chr.5: 79115828-79115828 on Build GRCh38
rs746271866 Chr.5: 79115907-79115907 on Build GRCh38
rs535880396 Chr.5: 79116053-79116053 on Build GRCh38
rs772543723 Chr.5: 79115788-79115788 on Build GRCh38
rs994377607 Chr.5: 79116101-79116101 on Build GRCh38
rs771258014 Chr.5: 79115808-79115808 on Build GRCh38
rs537144250 Chr.5: 79116173-79116173 on Build GRCh38
rs373553877 Chr.5: 79115847-79115847 on Build GRCh38
rs766149888 Chr.5: 79115848-79115848 on Build GRCh38
rs556663914 Chr.5: 79116174-79116174 on Build GRCh38
rs976709806 Chr.5: 79116177-79116177 on Build GRCh38
rs578139197 Chr.5: 79116150-79116150 on Build GRCh38
rs765599787 Chr.5: 79115821-79115821 on Build GRCh38
rs369885790 Chr.5: 79115835-79115835 on Build GRCh38
rs906167690 Chr.5: 79116026-79116026 on Build GRCh38
rs989147649 Chr.5: 79115953-79115953 on Build GRCh38
rs750231986 Chr.5: 79115726-79115726 on Build GRCh38
rs35826365 Chr.5: 79115967-79115968 on Build GRCh38
rs777266312 Chr.5: 79115880-79115880 on Build GRCh38
rs528235368 Chr.5: 79115815-79115815 on Build GRCh38
rs763276045 Chr.5: 79115919-79115919 on Build GRCh38
rs58691147 Chr.5: 79115940-79115940 on Build GRCh38
rs947968027 Chr.5: 79115994-79115994 on Build GRCh38
rs566393266 Chr.5: 79116085-79116085 on Build GRCh38
rs376343021 Chr.5: 79115809-79115809 on Build GRCh38
rs753607977 Chr.5: 79115854-79115854 on Build GRCh38
rs906341431 Chr.5: 79116021-79116021 on Build GRCh38
rs771359725 Chr.5: 79115900-79115900 on Build GRCh38
rs897924814 Chr.5: 79116099-79116099 on Build GRCh38
rs371308517 Chr.5: 79115939-79115939 on Build GRCh38
rs747489317 Chr.5: 79115887-79115887 on Build GRCh38
rs888548010 Chr.5: 79116117-79116117 on Build GRCh38
rs755346884 Chr.5: 79116132-79116132 on Build GRCh38
rs619100 Chr.5: 79115999-79115999 on Build GRCh38
rs747327020 Chr.5: 79115804-79115804 on Build GRCh38
rs370246578 Chr.5: 79115944-79115944 on Build GRCh38
rs767237180 Chr.5: 79115957-79115957 on Build GRCh38
rs752276097 Chr.5: 79115941-79115941 on Build GRCh38
rs770132110 Chr.5: 79115908-79115908 on Build GRCh38
rs760611373 Chr.5: 79115814-79115814 on Build GRCh38
rs535666974 Chr.5: 79115829-79115829 on Build GRCh38
rs772588128 Chr.5: 79115881-79115882 on Build GRCh38
rs775924721 Chr.5: 79115916-79115916 on Build GRCh38
rs528901081 Chr.5: 79115740-79115740 on Build GRCh38
rs1027095099 Chr.5: 79115768-79115768 on Build GRCh38
rs746363908 Chr.5: 79115719-79115719 on Build GRCh38
rs897392949 Chr.5: 79116096-79116096 on Build GRCh38
rs141007396 Chr.5: 79116109-79116109 on Build GRCh38
rs56709544 Chr.5: 79115779-79115779 on Build GRCh38

View Full Product Details