Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 79119167-79119669 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BHMT
Entrez Gene ID
635
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
BHMT1, HEL-S-61p
UniGene
Hs.80756
Gene Chromosome Location
Chr.5: 79111781-79132290 on Build GRCh38
Transcripts
NM_001713.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6171938 c.175C>G p.L59V Chr.5: 79119267-79119267 on Build GRCh38
3393674 c.258G>C p.R86S Chr.5: 79119350-79119350 on Build GRCh38
3429671 c.170G>A p.R57H Chr.5: 79119262-79119262 on Build GRCh38
5666702 c.189C>A p.F63L Chr.5: 79119281-79119281 on Build GRCh38
450090 c.215A>C p.Q72P Chr.5: 79119307-79119307 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747373233 Chr.5: 79119372-79119373 on Build GRCh38
rs770452767 Chr.5: 79119562-79119562 on Build GRCh38
rs925882963 Chr.5: 79119323-79119323 on Build GRCh38
rs773754654 Chr.5: 79119638-79119642 on Build GRCh38
rs1028899430 Chr.5: 79119350-79119350 on Build GRCh38
rs759472468 Chr.5: 79119248-79119248 on Build GRCh38
rs369270690 Chr.5: 79119419-79119419 on Build GRCh38
rs1055467885 Chr.5: 79119500-79119500 on Build GRCh38
rs764950849 Chr.5: 79119254-79119254 on Build GRCh38
rs967164768 Chr.5: 79119421-79119421 on Build GRCh38
rs772644856 Chr.5: 79119303-79119303 on Build GRCh38
rs377251028 Chr.5: 79119213-79119213 on Build GRCh38
rs771977077 Chr.5: 79119308-79119308 on Build GRCh38
rs754240613 Chr.5: 79119409-79119409 on Build GRCh38
rs758665842 Chr.5: 79119428-79119428 on Build GRCh38
rs368457430 Chr.5: 79119239-79119239 on Build GRCh38
rs201963562 Chr.5: 79119400-79119400 on Build GRCh38
rs575221062 Chr.5: 79119453-79119453 on Build GRCh38
rs1029830079 Chr.5: 79119493-79119493 on Build GRCh38
rs749474539 Chr.5: 79119225-79119225 on Build GRCh38
rs773813393 Chr.5: 79119391-79119391 on Build GRCh38
rs949583133 Chr.5: 79119535-79119535 on Build GRCh38
rs778379680 Chr.5: 79119354-79119354 on Build GRCh38
rs762797454 Chr.5: 79119338-79119338 on Build GRCh38
rs374909768 Chr.5: 79119231-79119231 on Build GRCh38
rs886534149 Chr.5: 79119276-79119276 on Build GRCh38
rs370122161 Chr.5: 79119214-79119214 on Build GRCh38
rs57651345 Chr.5: 79119509-79119509 on Build GRCh38
rs1005033366 Chr.5: 79119625-79119625 on Build GRCh38
rs761846550 Chr.5: 79119236-79119236 on Build GRCh38
rs145214742 Chr.5: 79119211-79119211 on Build GRCh38
rs769031815 Chr.5: 79119227-79119227 on Build GRCh38
rs1038605478 Chr.5: 79119435-79119435 on Build GRCh38
rs548467116 Chr.5: 79119299-79119299 on Build GRCh38
rs955735254 Chr.5: 79119557-79119557 on Build GRCh38
rs997125818 Chr.5: 79119481-79119481 on Build GRCh38
rs756712401 Chr.5: 79119281-79119281 on Build GRCh38
rs373954954 Chr.5: 79119212-79119212 on Build GRCh38
rs577010343 Chr.5: 79119403-79119403 on Build GRCh38
rs746456103 Chr.5: 79119393-79119393 on Build GRCh38
rs978945466 Chr.5: 79119461-79119461 on Build GRCh38
rs978384042 Chr.5: 79119622-79119622 on Build GRCh38
rs780760620 Chr.5: 79119273-79119273 on Build GRCh38
rs568737681 Chr.5: 79119244-79119244 on Build GRCh38
rs780300316 Chr.5: 79119320-79119320 on Build GRCh38
rs559039767 Chr.5: 79119423-79119423 on Build GRCh38
rs370702877 Chr.5: 79119274-79119274 on Build GRCh38
rs35041410 Chr.5: 79119314-79119314 on Build GRCh38
rs940638024 Chr.5: 79119194-79119194 on Build GRCh38
rs147391267 Chr.5: 79119262-79119262 on Build GRCh38
rs997032472 Chr.5: 79119246-79119246 on Build GRCh38
rs537874141 Chr.5: 79119271-79119271 on Build GRCh38
rs373259906 Chr.5: 79119543-79119543 on Build GRCh38
rs978623032 Chr.5: 79119317-79119317 on Build GRCh38
rs568794365 Chr.5: 79119438-79119438 on Build GRCh38
rs544215086 Chr.5: 79119506-79119506 on Build GRCh38
rs776047105 Chr.5: 79119586-79119586 on Build GRCh38
rs1020469039 Chr.5: 79119563-79119563 on Build GRCh38
rs763800908 Chr.5: 79119347-79119347 on Build GRCh38
rs996120287 Chr.5: 79119332-79119332 on Build GRCh38
rs554138869 Chr.5: 79119643-79119643 on Build GRCh38
rs780528487 Chr.5: 79119285-79119285 on Build GRCh38
rs372610894 Chr.5: 79119261-79119261 on Build GRCh38
rs181542647 Chr.5: 79119634-79119634 on Build GRCh38
rs189088164 Chr.5: 79119588-79119588 on Build GRCh38
rs748259122 Chr.5: 79119424-79119424 on Build GRCh38
rs767043338 Chr.5: 79119408-79119408 on Build GRCh38
rs749710704 Chr.5: 79119289-79119289 on Build GRCh38
rs925715086 Chr.5: 79119466-79119466 on Build GRCh38
rs193083153 Chr.5: 79119325-79119325 on Build GRCh38
rs768910093 Chr.5: 79119400-79119400 on Build GRCh38
rs766889112 Chr.5: 79119394-79119394 on Build GRCh38
rs752508939 Chr.5: 79119258-79119258 on Build GRCh38

View Full Product Details