Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80501778-80502311 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM151B
Entrez Gene ID
167555
Gene Name
family with sequence similarity 151 member B
Gene Aliases
UNQ9217
UniGene
Hs.338182
Gene Chromosome Location
Chr.5: 80487981-80542563 on Build GRCh38
Transcripts
NM_205548.2 XM_006714564.3 XM_011543234.2 XM_011543235.2 XM_017009166.1 XM_017009168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1634344 c.151+1G>A p.? Chr.5: 80501918-80501918 on Build GRCh38
5816167 c.49G>T p.E17* Chr.5: 80501815-80501815 on Build GRCh38
6171890 c.33G>T p.W11C Chr.5: 80501799-80501799 on Build GRCh38
4682615 c.89G>C p.G30A Chr.5: 80501855-80501855 on Build GRCh38
1070669 c.88G>A p.G30S Chr.5: 80501854-80501854 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs59410708 Chr.5: 80502173-80502173 on Build GRCh38
rs903862594 Chr.5: 80502264-80502264 on Build GRCh38
rs144997302 Chr.5: 80501983-80501983 on Build GRCh38
rs1051575364 Chr.5: 80502217-80502217 on Build GRCh38
rs573712190 Chr.5: 80502072-80502072 on Build GRCh38
rs771669308 Chr.5: 80501851-80501851 on Build GRCh38
rs149944081 Chr.5: 80501979-80501979 on Build GRCh38
rs373194560 Chr.5: 80501948-80501948 on Build GRCh38
rs747682698 Chr.5: 80501848-80501848 on Build GRCh38
rs201002289 Chr.5: 80501957-80501957 on Build GRCh38
rs891602435 Chr.5: 80502221-80502221 on Build GRCh38
rs536219059 Chr.5: 80502102-80502102 on Build GRCh38
rs779978732 Chr.5: 80501911-80501911 on Build GRCh38
rs908898459 Chr.5: 80502117-80502117 on Build GRCh38
rs967790749 Chr.5: 80502005-80502006 on Build GRCh38
rs747844083 Chr.5: 80501952-80501952 on Build GRCh38
rs752874602 Chr.5: 80501876-80501876 on Build GRCh38
rs763127533 Chr.5: 80501877-80501877 on Build GRCh38
rs557189315 Chr.5: 80502035-80502035 on Build GRCh38
rs565133930 Chr.5: 80502207-80502207 on Build GRCh38
rs919566593 Chr.5: 80502236-80502236 on Build GRCh38
rs781135092 Chr.5: 80501809-80501809 on Build GRCh38
rs375436982 Chr.5: 80501940-80501940 on Build GRCh38
rs560871185 Chr.5: 80501814-80501814 on Build GRCh38
rs999411040 Chr.5: 80502089-80502089 on Build GRCh38
rs185892198 Chr.5: 80502263-80502263 on Build GRCh38
rs779772925 Chr.5: 80501838-80501838 on Build GRCh38
rs749107242 Chr.5: 80501844-80501844 on Build GRCh38
rs544528188 Chr.5: 80502252-80502252 on Build GRCh38
rs746461511 Chr.5: 80501968-80501968 on Build GRCh38
rs911080990 Chr.5: 80502146-80502146 on Build GRCh38
rs1055919730 Chr.5: 80501960-80501960 on Build GRCh38
rs1010126580 Chr.5: 80502245-80502245 on Build GRCh38
rs756060319 Chr.5: 80501836-80501836 on Build GRCh38
rs149355931 Chr.5: 80501871-80501871 on Build GRCh38
rs545165799 Chr.5: 80502192-80502192 on Build GRCh38
rs986618741 Chr.5: 80502138-80502138 on Build GRCh38
rs372187543 Chr.5: 80501843-80501843 on Build GRCh38
rs374879012 Chr.5: 80501853-80501853 on Build GRCh38
rs983677680 Chr.5: 80502108-80502108 on Build GRCh38
rs551991757 Chr.5: 80501959-80501959 on Build GRCh38
rs900286050 Chr.5: 80502177-80502177 on Build GRCh38
rs771455070 Chr.5: 80502098-80502098 on Build GRCh38
rs768429430 Chr.5: 80501845-80501845 on Build GRCh38
rs11951382 Chr.5: 80502279-80502279 on Build GRCh38
rs750363890 Chr.5: 80501811-80501811 on Build GRCh38
rs373423516 Chr.5: 80501873-80501873 on Build GRCh38
rs776864023 Chr.5: 80501870-80501870 on Build GRCh38
rs575299992 Chr.5: 80502226-80502226 on Build GRCh38
rs543693575 Chr.5: 80501846-80501846 on Build GRCh38
rs35110098 Chr.5: 80502035-80502036 on Build GRCh38
rs753699675 Chr.5: 80501924-80501924 on Build GRCh38
rs751648264 Chr.5: 80501895-80501895 on Build GRCh38
rs759523231 Chr.5: 80501812-80501812 on Build GRCh38
rs146291904 Chr.5: 80501892-80501892 on Build GRCh38
rs941653982 Chr.5: 80502123-80502123 on Build GRCh38
rs558318475 Chr.5: 80501900-80501900 on Build GRCh38
rs369053764 Chr.5: 80501854-80501854 on Build GRCh38
rs146104489 Chr.5: 80502182-80502182 on Build GRCh38

View Full Product Details