Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80541529-80542049 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM151B
Entrez Gene ID
167555
Gene Name
family with sequence similarity 151 member B
Gene Aliases
UNQ9217
UniGene
Hs.338182
Gene Chromosome Location
Chr.5: 80487981-80542563 on Build GRCh38
Transcripts
NM_205548.2 XM_006714564.3 XM_011543234.2 XM_011543235.2 XM_011543236.2 XM_011543237.2 XM_017009166.1 XM_017009167.1 XM_017009168.1 XM_017009169.1 XM_017009170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3618309 c.789C>T p.N263N Chr.5: 80541790-80541790 on Build GRCh38
3618308 c.713C>T p.S238F Chr.5: 80541714-80541714 on Build GRCh38
3776891 c.828C>T p.L276L Chr.5: 80541829-80541829 on Build GRCh38
5995829 c.728T>A p.L243H Chr.5: 80541729-80541729 on Build GRCh38
3776890 c.816C>G p.I272M Chr.5: 80541817-80541817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138264758 Chr.5: 80541715-80541715 on Build GRCh38
rs749921216 Chr.5: 80541831-80541833 on Build GRCh38
rs186380131 Chr.5: 80541929-80541929 on Build GRCh38
rs907492480 Chr.5: 80541717-80541717 on Build GRCh38
rs940450147 Chr.5: 80541734-80541734 on Build GRCh38
rs754668019 Chr.5: 80541796-80541796 on Build GRCh38
rs892539564 Chr.5: 80542006-80542006 on Build GRCh38
rs760874824 Chr.5: 80541628-80541628 on Build GRCh38
rs771330562 Chr.5: 80541832-80541832 on Build GRCh38
rs780911515 Chr.5: 80541659-80541659 on Build GRCh38
rs772472704 Chr.5: 80541809-80541809 on Build GRCh38
rs555553887 Chr.5: 80541792-80541792 on Build GRCh38
rs185811036 Chr.5: 80541608-80541608 on Build GRCh38
rs752813118 Chr.5: 80541758-80541758 on Build GRCh38
rs1045731894 Chr.5: 80541827-80541827 on Build GRCh38
rs765376708 Chr.5: 80541850-80541850 on Build GRCh38
rs773726227 Chr.5: 80541810-80541810 on Build GRCh38
rs759821618 Chr.5: 80541844-80541844 on Build GRCh38
rs949576467 Chr.5: 80541807-80541807 on Build GRCh38
rs995274741 Chr.5: 80541897-80541897 on Build GRCh38
rs879250560 Chr.5: 80541908-80541908 on Build GRCh38
rs766557488 Chr.5: 80541634-80541634 on Build GRCh38
rs764889708 Chr.5: 80541752-80541752 on Build GRCh38
rs755009229 Chr.5: 80541646-80541646 on Build GRCh38
rs775444232 Chr.5: 80541858-80541858 on Build GRCh38
rs779806513 Chr.5: 80541688-80541688 on Build GRCh38
rs778116783 Chr.5: 80541716-80541716 on Build GRCh38
rs1023801748 Chr.5: 80541587-80541587 on Build GRCh38
rs747630538 Chr.5: 80541803-80541803 on Build GRCh38
rs748841886 Chr.5: 80541693-80541693 on Build GRCh38
rs1009643921 Chr.5: 80542017-80542017 on Build GRCh38
rs756809033 Chr.5: 80541744-80541745 on Build GRCh38
rs761044598 Chr.5: 80541719-80541719 on Build GRCh38
rs1028461476 Chr.5: 80541923-80541923 on Build GRCh38
rs779967842 Chr.5: 80541793-80541793 on Build GRCh38
rs771321376 Chr.5: 80541772-80541772 on Build GRCh38
rs190610363 Chr.5: 80541635-80541635 on Build GRCh38
rs773980596 Chr.5: 80541706-80541706 on Build GRCh38
rs569857436 Chr.5: 80541606-80541606 on Build GRCh38
rs776793995 Chr.5: 80541831-80541831 on Build GRCh38
rs373906305 Chr.5: 80541625-80541625 on Build GRCh38
rs367813400 Chr.5: 80541652-80541652 on Build GRCh38
rs778633681 Chr.5: 80541799-80541799 on Build GRCh38
rs142754508 Chr.5: 80541794-80541794 on Build GRCh38
rs753557834 Chr.5: 80541745-80541745 on Build GRCh38
rs200855590 Chr.5: 80541750-80541750 on Build GRCh38
rs542011842 Chr.5: 80541841-80541841 on Build GRCh38
rs543521901 Chr.5: 80541745-80541745 on Build GRCh38
rs1054915739 Chr.5: 80541676-80541676 on Build GRCh38
rs765358570 Chr.5: 80541757-80541757 on Build GRCh38
rs762285644 Chr.5: 80541712-80541712 on Build GRCh38
rs181339470 Chr.5: 80541669-80541669 on Build GRCh38
rs1004411834 Chr.5: 80541720-80541720 on Build GRCh38
rs905330027 Chr.5: 80541905-80541905 on Build GRCh38
rs368080662 Chr.5: 80541626-80541626 on Build GRCh38
rs945263029 Chr.5: 80541614-80541614 on Build GRCh38
rs372277066 Chr.5: 80541667-80541667 on Build GRCh38
rs766617543 Chr.5: 80541737-80541737 on Build GRCh38
rs373669675 Chr.5: 80541648-80541648 on Build GRCh38
rs747439432 Chr.5: 80541825-80541825 on Build GRCh38
rs757200992 Chr.5: 80541654-80541654 on Build GRCh38
rs750204491 Chr.5: 80541776-80541776 on Build GRCh38
rs767387859 Chr.5: 80541880-80541880 on Build GRCh38
rs747433865 Chr.5: 80541768-80541768 on Build GRCh38
rs953876874 Chr.5: 80542000-80542000 on Build GRCh38
rs768325355 Chr.5: 80541702-80541702 on Build GRCh38
rs545776704 Chr.5: 80541954-80541954 on Build GRCh38
rs763037726 Chr.5: 80541872-80541872 on Build GRCh38

View Full Product Details