Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80559696-80560193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34B
Entrez Gene ID
340120
Gene Name
ankyrin repeat domain 34B
Gene Aliases
DP58
UniGene
Hs.519404
Gene Chromosome Location
Chr.5: 80556755-80570485 on Build GRCh38
Transcripts
NM_001004441.2 XM_006714599.3 XM_011543366.2 XM_011543369.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6600681 c.131A>G p.E44G Chr.5: 80559889-80559889 on Build GRCh38
5580229 c.201G>A p.V67V Chr.5: 80559819-80559819 on Build GRCh38
1292165 c.280G>A p.E94K Chr.5: 80559740-80559740 on Build GRCh38
6575704 c.262C>G p.L88V Chr.5: 80559758-80559758 on Build GRCh38
5559228 c.86T>C p.L29S Chr.5: 80559934-80559934 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767862318 Chr.5: 80559955-80559955 on Build GRCh38
rs765922987 Chr.5: 80559898-80559899 on Build GRCh38
rs983302797 Chr.5: 80559941-80559941 on Build GRCh38
rs755267962 Chr.5: 80560049-80560049 on Build GRCh38
rs779140867 Chr.5: 80559975-80559975 on Build GRCh38
rs372701643 Chr.5: 80559793-80559793 on Build GRCh38
rs865774055 Chr.5: 80559988-80559988 on Build GRCh38
rs775051217 Chr.5: 80559933-80559933 on Build GRCh38
rs992341636 Chr.5: 80559852-80559852 on Build GRCh38
rs753578611 Chr.5: 80559722-80559722 on Build GRCh38
rs377193284 Chr.5: 80559807-80559807 on Build GRCh38
rs552814333 Chr.5: 80559979-80559979 on Build GRCh38
rs142137939 Chr.5: 80559917-80559917 on Build GRCh38
rs148638555 Chr.5: 80559883-80559883 on Build GRCh38
rs752084873 Chr.5: 80559744-80559744 on Build GRCh38
rs750831837 Chr.5: 80559956-80559956 on Build GRCh38
rs865951841 Chr.5: 80559949-80559949 on Build GRCh38
rs760509384 Chr.5: 80559718-80559718 on Build GRCh38
rs185775880 Chr.5: 80560050-80560050 on Build GRCh38
rs775271852 Chr.5: 80559794-80559794 on Build GRCh38
rs746160231 Chr.5: 80559902-80559902 on Build GRCh38
rs144658829 Chr.5: 80559855-80559855 on Build GRCh38
rs761232546 Chr.5: 80560032-80560032 on Build GRCh38
rs753280742 Chr.5: 80559979-80559979 on Build GRCh38
rs200291996 Chr.5: 80560004-80560004 on Build GRCh38
rs766247967 Chr.5: 80559720-80559720 on Build GRCh38
rs758452396 Chr.5: 80560058-80560058 on Build GRCh38
rs367911402 Chr.5: 80560046-80560046 on Build GRCh38
rs774562997 Chr.5: 80559916-80559916 on Build GRCh38
rs781522784 Chr.5: 80559785-80559785 on Build GRCh38
rs761273142 Chr.5: 80560009-80560009 on Build GRCh38
rs1021087764 Chr.5: 80560068-80560068 on Build GRCh38
rs767023197 Chr.5: 80560014-80560014 on Build GRCh38
rs752116185 Chr.5: 80559891-80559891 on Build GRCh38
rs765046677 Chr.5: 80559926-80559926 on Build GRCh38
rs559948107 Chr.5: 80559900-80559900 on Build GRCh38
rs759235997 Chr.5: 80559872-80559872 on Build GRCh38
rs771760179 Chr.5: 80559751-80559751 on Build GRCh38
rs904118108 Chr.5: 80560110-80560110 on Build GRCh38
rs1033316735 Chr.5: 80559817-80559817 on Build GRCh38
rs773871880 Chr.5: 80560005-80560005 on Build GRCh38
rs778209016 Chr.5: 80559757-80559757 on Build GRCh38
rs775347673 Chr.5: 80559996-80559996 on Build GRCh38
rs376018983 Chr.5: 80559797-80559797 on Build GRCh38
rs145042037 Chr.5: 80559822-80559822 on Build GRCh38
rs757774393 Chr.5: 80559954-80559954 on Build GRCh38
rs1053163348 Chr.5: 80560040-80560040 on Build GRCh38
rs756381871 Chr.5: 80559906-80559906 on Build GRCh38
rs778555790 Chr.5: 80559728-80559728 on Build GRCh38
rs752279417 Chr.5: 80559878-80559878 on Build GRCh38
rs772167886 Chr.5: 80559812-80559812 on Build GRCh38
rs762566361 Chr.5: 80559833-80559834 on Build GRCh38
rs370883986 Chr.5: 80559784-80559784 on Build GRCh38
rs750943122 Chr.5: 80560027-80560027 on Build GRCh38
rs749580989 Chr.5: 80559967-80559967 on Build GRCh38
rs996745246 Chr.5: 80560100-80560100 on Build GRCh38
rs199801377 Chr.5: 80559880-80559880 on Build GRCh38
rs551674336 Chr.5: 80559923-80559923 on Build GRCh38
rs201143692 Chr.5: 80559896-80559896 on Build GRCh38
rs769532469 Chr.5: 80559993-80559993 on Build GRCh38
rs750012958 Chr.5: 80559798-80559799 on Build GRCh38
rs757755790 Chr.5: 80559895-80559895 on Build GRCh38
rs968250233 Chr.5: 80559772-80559772 on Build GRCh38
rs367829617 Chr.5: 80559938-80559938 on Build GRCh38
rs762597114 Chr.5: 80559997-80559997 on Build GRCh38
rs528734024 Chr.5: 80559907-80559907 on Build GRCh38
rs770794401 Chr.5: 80560028-80560028 on Build GRCh38
rs755251156 Chr.5: 80559970-80559970 on Build GRCh38
rs187262 Chr.5: 80560057-80560057 on Build GRCh38
rs369135971 Chr.5: 80559904-80559904 on Build GRCh38
rs759208087 Chr.5: 80559924-80559924 on Build GRCh38
rs749496251 Chr.5: 80559909-80559909 on Build GRCh38
rs764975825 Chr.5: 80559788-80559789 on Build GRCh38
rs775652032 Chr.5: 80559798-80559798 on Build GRCh38
rs746166737 Chr.5: 80559790-80559790 on Build GRCh38
rs543830365 Chr.5: 80559719-80559719 on Build GRCh38
rs375779222 Chr.5: 80559743-80559743 on Build GRCh38
rs763658538 Chr.5: 80559944-80559944 on Build GRCh38
rs771330677 Chr.5: 80559782-80559782 on Build GRCh38
rs772087145 Chr.5: 80559727-80559727 on Build GRCh38
rs371220836 Chr.5: 80559939-80559939 on Build GRCh38
rs151166785 Chr.5: 80559978-80559978 on Build GRCh38
rs776469711 Chr.5: 80559984-80559984 on Build GRCh38
rs747481528 Chr.5: 80559763-80559763 on Build GRCh38
rs768907369 Chr.5: 80559913-80559913 on Build GRCh38
rs770926920 Chr.5: 80559981-80559981 on Build GRCh38
rs1010151112 Chr.5: 80560066-80560066 on Build GRCh38
rs201133010 Chr.5: 80559950-80559950 on Build GRCh38
rs762578409 Chr.5: 80559935-80559935 on Build GRCh38
rs745792603 Chr.5: 80559991-80559991 on Build GRCh38
rs777893126 Chr.5: 80560071-80560071 on Build GRCh38
rs760668942 Chr.5: 80559835-80559835 on Build GRCh38
rs754575567 Chr.5: 80559724-80559724 on Build GRCh38
rs373542062 Chr.5: 80559783-80559783 on Build GRCh38
rs756476173 Chr.5: 80559958-80559958 on Build GRCh38
rs200046558 Chr.5: 80559964-80559964 on Build GRCh38
rs151244325 Chr.5: 80559873-80559873 on Build GRCh38

View Full Product Details