Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80563652-80564153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD34B
Entrez Gene ID
340120
Gene Name
ankyrin repeat domain 34B
Gene Aliases
DP58
UniGene
Hs.519404
Gene Chromosome Location
Chr.5: 80556755-80570485 on Build GRCh38
Transcripts
NM_001004441.2 XM_006714599.3 XM_011543366.2 XM_011543369.2

SNP Detail

SNP ID Chromosome Location
rs570680522 Chr.5: 80563894-80563894 on Build GRCh38
rs756078246 Chr.5: 80564094-80564094 on Build GRCh38
rs902024264 Chr.5: 80563756-80563756 on Build GRCh38
rs527532157 Chr.5: 80563830-80563830 on Build GRCh38
rs972498340 Chr.5: 80563879-80563879 on Build GRCh38
rs139492434 Chr.5: 80563945-80563945 on Build GRCh38
rs555219319 Chr.5: 80564114-80564114 on Build GRCh38
rs763814710 Chr.5: 80563690-80563690 on Build GRCh38
rs73769141 Chr.5: 80563947-80563947 on Build GRCh38
rs769422353 Chr.5: 80563997-80564000 on Build GRCh38
rs112996507 Chr.5: 80563806-80563806 on Build GRCh38
rs890412252 Chr.5: 80563996-80563996 on Build GRCh38
rs141612331 Chr.5: 80563737-80563737 on Build GRCh38
rs573691934 Chr.5: 80563939-80563939 on Build GRCh38
rs893391143 Chr.5: 80563803-80563803 on Build GRCh38
rs943451244 Chr.5: 80563708-80563708 on Build GRCh38
rs952621815 Chr.5: 80563911-80563911 on Build GRCh38
rs915320748 Chr.5: 80563880-80563880 on Build GRCh38
rs946818296 Chr.5: 80563899-80563899 on Build GRCh38
rs756787812 Chr.5: 80563948-80563948 on Build GRCh38
rs1040839063 Chr.5: 80563717-80563717 on Build GRCh38
rs1053368263 Chr.5: 80563795-80563795 on Build GRCh38
rs919881570 Chr.5: 80563892-80563892 on Build GRCh38
rs998991671 Chr.5: 80563767-80563767 on Build GRCh38
rs1007341055 Chr.5: 80564088-80564088 on Build GRCh38
rs768218084 Chr.5: 80563891-80563894 on Build GRCh38
rs934855725 Chr.5: 80564085-80564085 on Build GRCh38
rs750265749 Chr.5: 80564058-80564058 on Build GRCh38
rs1035818306 Chr.5: 80563854-80563854 on Build GRCh38
rs368589671 Chr.5: 80563844-80563844 on Build GRCh38
rs918943439 Chr.5: 80563808-80563808 on Build GRCh38
rs954511635 Chr.5: 80563716-80563716 on Build GRCh38
rs34914263 Chr.5: 80563987-80563987 on Build GRCh38
rs1017422157 Chr.5: 80564111-80564111 on Build GRCh38
rs555221661 Chr.5: 80563874-80563874 on Build GRCh38
rs538202931 Chr.5: 80563837-80563837 on Build GRCh38

View Full Product Details