Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 91369700-91370197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC3
Entrez Gene ID
57561
Gene Name
arrestin domain containing 3
Gene Aliases
TLIMP
UniGene
Hs.24684
Gene Chromosome Location
Chr.5: 91368724-91383359 on Build GRCh38
Transcripts
NM_020801.2

SNP Detail

SNP ID Chromosome Location
rs994770197 Chr.5: 91369844-91369844 on Build GRCh38
rs559913447 Chr.5: 91369961-91369961 on Build GRCh38
rs192055939 Chr.5: 91369908-91369908 on Build GRCh38
rs542366336 Chr.5: 91370011-91370011 on Build GRCh38
rs187333080 Chr.5: 91369874-91369874 on Build GRCh38
rs574992490 Chr.5: 91370054-91370054 on Build GRCh38
rs577498205 Chr.5: 91369914-91369914 on Build GRCh38
rs961757070 Chr.5: 91370074-91370074 on Build GRCh38
rs912030158 Chr.5: 91369995-91369995 on Build GRCh38
rs987582438 Chr.5: 91369793-91369793 on Build GRCh38
rs763445240 Chr.5: 91369759-91369760 on Build GRCh38
rs1036043580 Chr.5: 91369837-91369839 on Build GRCh38
rs572006061 Chr.5: 91370154-91370154 on Build GRCh38
rs1027370297 Chr.5: 91370121-91370121 on Build GRCh38
rs1019051097 Chr.5: 91370068-91370068 on Build GRCh38
rs1002036049 Chr.5: 91369901-91369901 on Build GRCh38
rs1041836622 Chr.5: 91369963-91369963 on Build GRCh38
rs551982773 Chr.5: 91370153-91370153 on Build GRCh38
rs972112662 Chr.5: 91370108-91370108 on Build GRCh38
rs868395080 Chr.5: 91369931-91369931 on Build GRCh38
rs750854633 Chr.5: 91369957-91369957 on Build GRCh38
rs946014041 Chr.5: 91369962-91369962 on Build GRCh38
rs951762187 Chr.5: 91370123-91370123 on Build GRCh38
rs889066480 Chr.5: 91370025-91370025 on Build GRCh38
rs986248176 Chr.5: 91369987-91369987 on Build GRCh38
rs944397015 Chr.5: 91369830-91369830 on Build GRCh38
rs930299859 Chr.5: 91370059-91370059 on Build GRCh38
rs556221112 Chr.5: 91369734-91369734 on Build GRCh38
rs971822575 Chr.5: 91369884-91369884 on Build GRCh38
rs753541069 Chr.5: 91369872-91369872 on Build GRCh38
rs984285248 Chr.5: 91370131-91370131 on Build GRCh38
rs201912294 Chr.5: 91369756-91369757 on Build GRCh38
rs115222203 Chr.5: 91370021-91370021 on Build GRCh38
rs920323486 Chr.5: 91369893-91369893 on Build GRCh38
rs960100049 Chr.5: 91369905-91369905 on Build GRCh38
rs140200762 Chr.5: 91369967-91369967 on Build GRCh38
rs74345264 Chr.5: 91369960-91369960 on Build GRCh38
rs369735759 Chr.5: 91369941-91369941 on Build GRCh38
rs200438683 Chr.5: 91369757-91369758 on Build GRCh38
rs544589267 Chr.5: 91369862-91369862 on Build GRCh38
rs202077168 Chr.5: 91370053-91370053 on Build GRCh38
rs931496357 Chr.5: 91369903-91369903 on Build GRCh38
rs557655439 Chr.5: 91369909-91369909 on Build GRCh38
rs761728226 Chr.5: 91369757-91369759 on Build GRCh38
rs536371471 Chr.5: 91369838-91369838 on Build GRCh38
rs114643676 Chr.5: 91369873-91369873 on Build GRCh38
rs1000032233 Chr.5: 91370017-91370017 on Build GRCh38
rs114355367 Chr.5: 91370026-91370026 on Build GRCh38

View Full Product Details