Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 91374031-91374545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARRDC3
Entrez Gene ID
57561
Gene Name
arrestin domain containing 3
Gene Aliases
TLIMP
UniGene
Hs.24684
Gene Chromosome Location
Chr.5: 91368724-91383359 on Build GRCh38
Transcripts
NM_020801.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183677 c.1032A>C p.E344D Chr.5: 91374115-91374115 on Build GRCh38
6605245 c.1032A>T p.E344D Chr.5: 91374115-91374115 on Build GRCh38
1132086 c.928G>A p.G310S Chr.5: 91374219-91374219 on Build GRCh38
4586082 c.888T>C p.P296P Chr.5: 91374259-91374259 on Build GRCh38
3393712 c.961A>G p.T321A Chr.5: 91374186-91374186 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1024206955 Chr.5: 91374477-91374477 on Build GRCh38
rs777743615 Chr.5: 91374074-91374074 on Build GRCh38
rs775993938 Chr.5: 91374196-91374196 on Build GRCh38
rs386352279 Chr.5: 91374125-91374125 on Build GRCh38
rs541834208 Chr.5: 91374454-91374454 on Build GRCh38
rs997410236 Chr.5: 91374184-91374184 on Build GRCh38
rs755014961 Chr.5: 91374288-91374288 on Build GRCh38
rs760026211 Chr.5: 91374082-91374082 on Build GRCh38
rs939438512 Chr.5: 91374457-91374457 on Build GRCh38
rs879087673 Chr.5: 91374241-91374241 on Build GRCh38
rs774572344 Chr.5: 91374109-91374109 on Build GRCh38
rs1045992398 Chr.5: 91374446-91374446 on Build GRCh38
rs757255643 Chr.5: 91374272-91374272 on Build GRCh38
rs998491506 Chr.5: 91374396-91374396 on Build GRCh38
rs748320156 Chr.5: 91374123-91374123 on Build GRCh38
rs191101238 Chr.5: 91374291-91374291 on Build GRCh38
rs149605403 Chr.5: 91374274-91374274 on Build GRCh38
rs927938341 Chr.5: 91374266-91374266 on Build GRCh38
rs769300749 Chr.5: 91374318-91374318 on Build GRCh38
rs773585411 Chr.5: 91374145-91374145 on Build GRCh38
rs763959919 Chr.5: 91374231-91374231 on Build GRCh38
rs377395596 Chr.5: 91374096-91374096 on Build GRCh38
rs745804769 Chr.5: 91374276-91374276 on Build GRCh38
rs1003809514 Chr.5: 91374452-91374452 on Build GRCh38
rs113916429 Chr.5: 91374456-91374456 on Build GRCh38
rs757592968 Chr.5: 91374500-91374500 on Build GRCh38
rs767336839 Chr.5: 91374085-91374085 on Build GRCh38
rs899787323 Chr.5: 91374392-91374392 on Build GRCh38
rs377567633 Chr.5: 91374310-91374310 on Build GRCh38
rs772383068 Chr.5: 91374142-91374142 on Build GRCh38
rs751440152 Chr.5: 91374244-91374244 on Build GRCh38
rs760138850 Chr.5: 91374102-91374103 on Build GRCh38
rs116170695 Chr.5: 91374306-91374306 on Build GRCh38
rs553215264 Chr.5: 91374417-91374417 on Build GRCh38
rs974027859 Chr.5: 91374369-91374369 on Build GRCh38
rs146325981 Chr.5: 91374183-91374183 on Build GRCh38
rs201768181 Chr.5: 91374089-91374089 on Build GRCh38
rs1050745414 Chr.5: 91374425-91374425 on Build GRCh38
rs1010842787 Chr.5: 91374503-91374503 on Build GRCh38
rs747436050 Chr.5: 91374309-91374309 on Build GRCh38
rs886785721 Chr.5: 91374448-91374448 on Build GRCh38
rs576976301 Chr.5: 91374354-91374354 on Build GRCh38
rs955320381 Chr.5: 91374300-91374300 on Build GRCh38
rs748349590 Chr.5: 91374295-91374295 on Build GRCh38
rs772325924 Chr.5: 91374299-91374299 on Build GRCh38
rs573221895 Chr.5: 91374419-91374419 on Build GRCh38
rs982785307 Chr.5: 91374512-91374512 on Build GRCh38
rs539765237 Chr.5: 91374410-91374410 on Build GRCh38
rs548294485 Chr.5: 91374097-91374097 on Build GRCh38
rs995056917 Chr.5: 91374476-91374476 on Build GRCh38
rs948633203 Chr.5: 91374430-91374430 on Build GRCh38
rs758146770 Chr.5: 91374222-91374222 on Build GRCh38
rs758708335 Chr.5: 91374094-91374095 on Build GRCh38
rs946578453 Chr.5: 91374345-91374345 on Build GRCh38
rs759031409 Chr.5: 91374199-91374199 on Build GRCh38
rs138440730 Chr.5: 91374212-91374212 on Build GRCh38
rs752356907 Chr.5: 91374219-91374219 on Build GRCh38
rs768745371 Chr.5: 91374108-91374108 on Build GRCh38
rs1016639556 Chr.5: 91374455-91374455 on Build GRCh38
rs761144028 Chr.5: 91374154-91374154 on Build GRCh38
rs376935799 Chr.5: 91374319-91374319 on Build GRCh38
rs557142213 Chr.5: 91374316-91374316 on Build GRCh38
rs750691126 Chr.5: 91374072-91374074 on Build GRCh38
rs969697492 Chr.5: 91374490-91374490 on Build GRCh38
rs1011147044 Chr.5: 91374104-91374104 on Build GRCh38

View Full Product Details