Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95885020-95885516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL2
Entrez Gene ID
22936
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
-
UniGene
Hs.192221
Gene Chromosome Location
Chr.5: 95885098-95962288 on Build GRCh38
Transcripts
NM_012081.5 XM_006714575.2 XM_017009240.1 XM_017009241.1 XM_017009242.1 XM_017009243.1

SNP Detail

SNP ID Chromosome Location
rs970897774 Chr.5: 95885055-95885055 on Build GRCh38
rs575072466 Chr.5: 95885357-95885357 on Build GRCh38
rs1031846959 Chr.5: 95885343-95885343 on Build GRCh38
rs142416372 Chr.5: 95885134-95885134 on Build GRCh38
rs919386889 Chr.5: 95885045-95885045 on Build GRCh38
rs1000162146 Chr.5: 95885285-95885285 on Build GRCh38
rs543963052 Chr.5: 95885397-95885397 on Build GRCh38
rs530081747 Chr.5: 95885396-95885396 on Build GRCh38
rs1041774163 Chr.5: 95885187-95885187 on Build GRCh38
rs1054013177 Chr.5: 95885159-95885159 on Build GRCh38
rs904569291 Chr.5: 95885217-95885217 on Build GRCh38
rs974511522 Chr.5: 95885290-95885290 on Build GRCh38
rs984071831 Chr.5: 95885082-95885082 on Build GRCh38
rs779588109 Chr.5: 95885401-95885401 on Build GRCh38
rs561291854 Chr.5: 95885160-95885160 on Build GRCh38
rs746624629 Chr.5: 95885424-95885424 on Build GRCh38
rs370672982 Chr.5: 95885367-95885367 on Build GRCh38
rs980451468 Chr.5: 95885072-95885072 on Build GRCh38
rs1033338004 Chr.5: 95885417-95885417 on Build GRCh38
rs35457589 Chr.5: 95885454-95885455 on Build GRCh38
rs943619066 Chr.5: 95885137-95885137 on Build GRCh38
rs779311219 Chr.5: 95885257-95885257 on Build GRCh38
rs7918 Chr.5: 95885412-95885412 on Build GRCh38
rs541632615 Chr.5: 95885152-95885152 on Build GRCh38
rs147620971 Chr.5: 95885100-95885100 on Build GRCh38
rs539039936 Chr.5: 95885076-95885076 on Build GRCh38
rs754386438 Chr.5: 95885079-95885079 on Build GRCh38
rs561611053 Chr.5: 95885347-95885347 on Build GRCh38
rs1014396318 Chr.5: 95885352-95885352 on Build GRCh38
rs550891967 Chr.5: 95885292-95885292 on Build GRCh38
rs908467784 Chr.5: 95885098-95885098 on Build GRCh38
rs771474169 Chr.5: 95885356-95885356 on Build GRCh38
rs1049696450 Chr.5: 95885216-95885216 on Build GRCh38
rs952322975 Chr.5: 95885418-95885418 on Build GRCh38
rs946997121 Chr.5: 95885169-95885169 on Build GRCh38
rs932755871 Chr.5: 95885203-95885203 on Build GRCh38
rs749838541 Chr.5: 95885351-95885351 on Build GRCh38

View Full Product Details