Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95888769-95889221 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL2
Entrez Gene ID
22936
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
-
UniGene
Hs.192221
Gene Chromosome Location
Chr.5: 95885098-95962288 on Build GRCh38
Transcripts
NM_012081.5 XM_006714575.2 XM_017009239.1 XM_017009240.1 XM_017009241.1 XM_017009242.1 XM_017009243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3715281 c.1786G>T p.E596* Chr.5: 95889106-95889106 on Build GRCh38
5868980 c.1822C>T p.H608Y Chr.5: 95888972-95888972 on Build GRCh38
5807984 c.1855A>T p.N619Y Chr.5: 95888939-95888939 on Build GRCh38
6379499 c.1918T>C p.S640P Chr.5: 95888876-95888876 on Build GRCh38
5482015 c.1824T>C p.H608H Chr.5: 95888970-95888970 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371582892 Chr.5: 95889022-95889022 on Build GRCh38
rs769043715 Chr.5: 95888997-95888998 on Build GRCh38
rs779799397 Chr.5: 95888916-95888916 on Build GRCh38
rs760084933 Chr.5: 95888996-95888997 on Build GRCh38
rs748263765 Chr.5: 95888933-95888933 on Build GRCh38
rs199699795 Chr.5: 95888997-95888997 on Build GRCh38
rs544392198 Chr.5: 95889196-95889196 on Build GRCh38
rs750239854 Chr.5: 95888894-95888894 on Build GRCh38
rs112809853 Chr.5: 95889192-95889192 on Build GRCh38
rs746265461 Chr.5: 95889163-95889163 on Build GRCh38
rs566870740 Chr.5: 95889151-95889151 on Build GRCh38
rs369787361 Chr.5: 95888838-95888838 on Build GRCh38
rs759286686 Chr.5: 95888997-95889000 on Build GRCh38
rs777750798 Chr.5: 95889134-95889134 on Build GRCh38
rs763414952 Chr.5: 95889174-95889174 on Build GRCh38
rs755904077 Chr.5: 95889061-95889061 on Build GRCh38
rs200787935 Chr.5: 95889149-95889149 on Build GRCh38
rs976946931 Chr.5: 95888936-95888936 on Build GRCh38
rs752306101 Chr.5: 95888824-95888824 on Build GRCh38
rs775824181 Chr.5: 95889007-95889007 on Build GRCh38
rs1055198924 Chr.5: 95889038-95889038 on Build GRCh38
rs763840848 Chr.5: 95888842-95888842 on Build GRCh38
rs562499940 Chr.5: 95888813-95888813 on Build GRCh38
rs759878944 Chr.5: 95888995-95888995 on Build GRCh38
rs756951552 Chr.5: 95888863-95888863 on Build GRCh38
rs753668462 Chr.5: 95889112-95889112 on Build GRCh38
rs796780666 Chr.5: 95888997-95888998 on Build GRCh38
rs12515492 Chr.5: 95889079-95889079 on Build GRCh38
rs752544236 Chr.5: 95889014-95889015 on Build GRCh38
rs781174690 Chr.5: 95888889-95888889 on Build GRCh38
rs777043476 Chr.5: 95888985-95888985 on Build GRCh38
rs776384561 Chr.5: 95888997-95888999 on Build GRCh38
rs762550314 Chr.5: 95889011-95889011 on Build GRCh38
rs1016131695 Chr.5: 95888876-95888876 on Build GRCh38
rs775905401 Chr.5: 95889173-95889173 on Build GRCh38
rs200228969 Chr.5: 95888857-95888857 on Build GRCh38
rs965443805 Chr.5: 95888897-95888897 on Build GRCh38
rs571114020 Chr.5: 95889001-95889001 on Build GRCh38
rs749229622 Chr.5: 95889054-95889054 on Build GRCh38
rs188188780 Chr.5: 95888949-95888949 on Build GRCh38
rs769462915 Chr.5: 95889175-95889175 on Build GRCh38
rs1026997178 Chr.5: 95888793-95888793 on Build GRCh38
rs756180847 Chr.5: 95888915-95888915 on Build GRCh38
rs750258365 Chr.5: 95889042-95889042 on Build GRCh38
rs771107682 Chr.5: 95888951-95888951 on Build GRCh38
rs763566281 Chr.5: 95889014-95889014 on Build GRCh38
rs771197675 Chr.5: 95889146-95889146 on Build GRCh38
rs113652532 Chr.5: 95889015-95889016 on Build GRCh38
rs780415568 Chr.5: 95888839-95888840 on Build GRCh38
rs762873736 Chr.5: 95889078-95889079 on Build GRCh38
rs868195204 Chr.5: 95888917-95888917 on Build GRCh38
rs761625881 Chr.5: 95889026-95889026 on Build GRCh38
rs749734407 Chr.5: 95888853-95888854 on Build GRCh38
rs770008048 Chr.5: 95889167-95889167 on Build GRCh38
rs767267861 Chr.5: 95889041-95889041 on Build GRCh38
rs954296066 Chr.5: 95888997-95889004 on Build GRCh38
rs924981429 Chr.5: 95889017-95889017 on Build GRCh38
rs747084414 Chr.5: 95889135-95889135 on Build GRCh38
rs772003479 Chr.5: 95888937-95888937 on Build GRCh38
rs754920792 Chr.5: 95889124-95889124 on Build GRCh38

View Full Product Details