Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 95898105-95898624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELL2
Entrez Gene ID
22936
Gene Name
elongation factor for RNA polymerase II 2
Gene Aliases
-
UniGene
Hs.192221
Gene Chromosome Location
Chr.5: 95885098-95962288 on Build GRCh38
Transcripts
NM_012081.5 XM_006714575.2 XM_017009239.1 XM_017009240.1 XM_017009241.1 XM_017009242.1 XM_017009243.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1487057 c.1468C>A p.L490I Chr.5: 95898297-95898297 on Build GRCh38
4625622 c.1415A>G p.K472R Chr.5: 95898350-95898350 on Build GRCh38
1439273 c.1239C>T p.D413D Chr.5: 95898526-95898526 on Build GRCh38
1439272 c.1404delA p.K468fs*8 Chr.5: 95898361-95898361 on Build GRCh38
4680780 c.1315C>T p.P439S Chr.5: 95898450-95898450 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021386932 Chr.5: 95898275-95898275 on Build GRCh38
rs745975751 Chr.5: 95898356-95898356 on Build GRCh38
rs192546844 Chr.5: 95898129-95898129 on Build GRCh38
rs141910684 Chr.5: 95898435-95898435 on Build GRCh38
rs895715873 Chr.5: 95898584-95898584 on Build GRCh38
rs982031559 Chr.5: 95898519-95898519 on Build GRCh38
rs761236024 Chr.5: 95898511-95898511 on Build GRCh38
rs1035837642 Chr.5: 95898561-95898561 on Build GRCh38
rs766854852 Chr.5: 95898512-95898512 on Build GRCh38
rs755585057 Chr.5: 95898313-95898313 on Build GRCh38
rs779707618 Chr.5: 95898315-95898315 on Build GRCh38
rs751389108 Chr.5: 95898551-95898551 on Build GRCh38
rs772374807 Chr.5: 95898506-95898506 on Build GRCh38
rs768928599 Chr.5: 95898483-95898483 on Build GRCh38
rs764561455 Chr.5: 95898290-95898290 on Build GRCh38
rs1050934339 Chr.5: 95898402-95898402 on Build GRCh38
rs749644542 Chr.5: 95898194-95898195 on Build GRCh38
rs372134332 Chr.5: 95898271-95898271 on Build GRCh38
rs192085807 Chr.5: 95898245-95898245 on Build GRCh38
rs749416548 Chr.5: 95898568-95898568 on Build GRCh38
rs757595594 Chr.5: 95898417-95898417 on Build GRCh38
rs757275862 Chr.5: 95898552-95898552 on Build GRCh38
rs376803324 Chr.5: 95898206-95898206 on Build GRCh38
rs145361722 Chr.5: 95898182-95898182 on Build GRCh38
rs769116737 Chr.5: 95898488-95898490 on Build GRCh38
rs762017859 Chr.5: 95898396-95898396 on Build GRCh38
rs567513100 Chr.5: 95898578-95898578 on Build GRCh38
rs200683081 Chr.5: 95898524-95898524 on Build GRCh38
rs577613428 Chr.5: 95898188-95898188 on Build GRCh38
rs764595047 Chr.5: 95898199-95898199 on Build GRCh38
rs759631959 Chr.5: 95898268-95898268 on Build GRCh38
rs774792969 Chr.5: 95898393-95898393 on Build GRCh38
rs201506781 Chr.5: 95898446-95898446 on Build GRCh38
rs775814335 Chr.5: 95898276-95898276 on Build GRCh38
rs777749619 Chr.5: 95898550-95898550 on Build GRCh38
rs749944628 Chr.5: 95898303-95898303 on Build GRCh38
rs779615458 Chr.5: 95898207-95898207 on Build GRCh38
rs763335907 Chr.5: 95898363-95898363 on Build GRCh38
rs998504163 Chr.5: 95898526-95898526 on Build GRCh38
rs915378030 Chr.5: 95898486-95898486 on Build GRCh38
rs973418304 Chr.5: 95898388-95898388 on Build GRCh38
rs1011532633 Chr.5: 95898451-95898451 on Build GRCh38
rs150448906 Chr.5: 95898151-95898151 on Build GRCh38
rs199535968 Chr.5: 95898450-95898450 on Build GRCh38
rs756595083 Chr.5: 95898430-95898430 on Build GRCh38
rs780340562 Chr.5: 95898476-95898476 on Build GRCh38
rs753355776 Chr.5: 95898515-95898515 on Build GRCh38
rs149812788 Chr.5: 95898448-95898448 on Build GRCh38
rs534114256 Chr.5: 95898505-95898505 on Build GRCh38
rs148844512 Chr.5: 95898216-95898216 on Build GRCh38
rs747878291 Chr.5: 95898341-95898341 on Build GRCh38
rs184261883 Chr.5: 95898150-95898150 on Build GRCh38
rs369188132 Chr.5: 95898510-95898510 on Build GRCh38
rs763430711 Chr.5: 95898283-95898283 on Build GRCh38
rs929309661 Chr.5: 95898138-95898138 on Build GRCh38
rs547693201 Chr.5: 95898558-95898558 on Build GRCh38
rs773993208 Chr.5: 95898212-95898212 on Build GRCh38
rs772956391 Chr.5: 95898345-95898345 on Build GRCh38
rs200730667 Chr.5: 95898298-95898298 on Build GRCh38
rs748992028 Chr.5: 95898330-95898330 on Build GRCh38
rs146881354 Chr.5: 95898520-95898520 on Build GRCh38
rs150632934 Chr.5: 95898438-95898438 on Build GRCh38
rs532846531 Chr.5: 95898331-95898331 on Build GRCh38
rs752242129 Chr.5: 95898544-95898544 on Build GRCh38
rs764210324 Chr.5: 95898387-95898387 on Build GRCh38
rs35528842 Chr.5: 95898355-95898356 on Build GRCh38
rs769944982 Chr.5: 95898359-95898359 on Build GRCh38
rs770990543 Chr.5: 95898462-95898462 on Build GRCh38
rs746990147 Chr.5: 95898460-95898460 on Build GRCh38
rs772055336 Chr.5: 95898343-95898343 on Build GRCh38
rs960008717 Chr.5: 95898355-95898355 on Build GRCh38
rs201482564 Chr.5: 95898334-95898334 on Build GRCh38
rs902827181 Chr.5: 95898281-95898281 on Build GRCh38
rs764973449 Chr.5: 95898529-95898529 on Build GRCh38
rs187406997 Chr.5: 95898196-95898196 on Build GRCh38
rs771895334 Chr.5: 95898230-95898230 on Build GRCh38
rs768392475 Chr.5: 95898210-95898210 on Build GRCh38
rs536412366 Chr.5: 95898593-95898593 on Build GRCh38
rs1039603824 Chr.5: 95898478-95898478 on Build GRCh38
rs780825682 Chr.5: 95898200-95898200 on Build GRCh38
rs758045644 Chr.5: 95898547-95898547 on Build GRCh38
rs750884400 Chr.5: 95898427-95898427 on Build GRCh38
rs748362880 Chr.5: 95898594-95898594 on Build GRCh38
rs139908232 Chr.5: 95898447-95898447 on Build GRCh38
rs372488423 Chr.5: 95898204-95898204 on Build GRCh38
rs774400321 Chr.5: 95898501-95898501 on Build GRCh38
rs200144988 Chr.5: 95898208-95898208 on Build GRCh38

View Full Product Details