Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 98855101-98855598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1
Entrez Gene ID
1105
Gene Name
chromodomain helicase DNA binding protein 1
Gene Aliases
-
UniGene
Hs.643465
Gene Chromosome Location
Chr.5: 98855204-98929014 on Build GRCh38
Transcripts
NM_001270.2 XM_005271866.4 XM_005271867.4 XM_011543112.2 XM_017008991.1 XM_017008992.1

SNP Detail

SNP ID Chromosome Location
rs529310149 Chr.5: 98855363-98855363 on Build GRCh38
rs867454912 Chr.5: 98855285-98855285 on Build GRCh38
rs1033958584 Chr.5: 98855531-98855531 on Build GRCh38
rs1000248631 Chr.5: 98855261-98855261 on Build GRCh38
rs1032405722 Chr.5: 98855191-98855191 on Build GRCh38
rs772198625 Chr.5: 98855428-98855428 on Build GRCh38
rs993392647 Chr.5: 98855323-98855324 on Build GRCh38
rs893927991 Chr.5: 98855477-98855477 on Build GRCh38
rs1036237357 Chr.5: 98855351-98855351 on Build GRCh38
rs35736545 Chr.5: 98855137-98855137 on Build GRCh38
rs945915792 Chr.5: 98855400-98855400 on Build GRCh38
rs904528114 Chr.5: 98855248-98855248 on Build GRCh38
rs931023537 Chr.5: 98855151-98855151 on Build GRCh38
rs1008072396 Chr.5: 98855164-98855164 on Build GRCh38
rs942522927 Chr.5: 98855162-98855165 on Build GRCh38
rs1024854212 Chr.5: 98855527-98855527 on Build GRCh38
rs983789255 Chr.5: 98855364-98855364 on Build GRCh38
rs904475682 Chr.5: 98855417-98855417 on Build GRCh38
rs1003768502 Chr.5: 98855519-98855519 on Build GRCh38
rs571415381 Chr.5: 98855472-98855472 on Build GRCh38
rs77918687 Chr.5: 98855138-98855138 on Build GRCh38
rs942496679 Chr.5: 98855373-98855373 on Build GRCh38
rs1021084905 Chr.5: 98855501-98855501 on Build GRCh38
rs1054186377 Chr.5: 98855476-98855476 on Build GRCh38
rs958374883 Chr.5: 98855290-98855290 on Build GRCh38
rs933234416 Chr.5: 98855377-98855377 on Build GRCh38
rs749500789 Chr.5: 98855228-98855228 on Build GRCh38
rs953780512 Chr.5: 98855139-98855139 on Build GRCh38
rs970954604 Chr.5: 98855247-98855247 on Build GRCh38
rs997432115 Chr.5: 98855145-98855145 on Build GRCh38
rs763444141 Chr.5: 98855264-98855265 on Build GRCh38
rs552549342 Chr.5: 98855440-98855440 on Build GRCh38
rs779177439 Chr.5: 98855337-98855337 on Build GRCh38
rs1034426621 Chr.5: 98855262-98855262 on Build GRCh38
rs1010169755 Chr.5: 98855233-98855233 on Build GRCh38
rs796667420 Chr.5: 98855201-98855201 on Build GRCh38
rs889090641 Chr.5: 98855156-98855156 on Build GRCh38
rs918270049 Chr.5: 98855149-98855149 on Build GRCh38
rs982293498 Chr.5: 98855266-98855266 on Build GRCh38
rs1041666419 Chr.5: 98855410-98855410 on Build GRCh38
rs549889479 Chr.5: 98855325-98855325 on Build GRCh38
rs1009565499 Chr.5: 98855485-98855485 on Build GRCh38
rs745940620 Chr.5: 98855340-98855340 on Build GRCh38
rs991543997 Chr.5: 98855297-98855297 on Build GRCh38
rs908991345 Chr.5: 98855327-98855327 on Build GRCh38
rs147480848 Chr.5: 98855126-98855126 on Build GRCh38
rs918217598 Chr.5: 98855359-98855360 on Build GRCh38
rs769929336 Chr.5: 98855153-98855153 on Build GRCh38
rs928301324 Chr.5: 98855283-98855283 on Build GRCh38
rs1007584596 Chr.5: 98855403-98855403 on Build GRCh38
rs910950907 Chr.5: 98855371-98855371 on Build GRCh38
rs1021073590 Chr.5: 98855231-98855231 on Build GRCh38
rs1014358489 Chr.5: 98855511-98855511 on Build GRCh38
rs894522443 Chr.5: 98855461-98855461 on Build GRCh38
rs367881297 Chr.5: 98855159-98855159 on Build GRCh38
rs532467831 Chr.5: 98855537-98855537 on Build GRCh38
rs560679935 Chr.5: 98855152-98855152 on Build GRCh38
rs940981349 Chr.5: 98855138-98855139 on Build GRCh38
rs1051653985 Chr.5: 98855380-98855380 on Build GRCh38
rs569611445 Chr.5: 98855327-98855327 on Build GRCh38
rs544848789 Chr.5: 98855333-98855333 on Build GRCh38
rs1040909286 Chr.5: 98855166-98855169 on Build GRCh38
rs561453710 Chr.5: 98855393-98855393 on Build GRCh38
rs970839618 Chr.5: 98855515-98855515 on Build GRCh38

View Full Product Details