Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 100535670-100536181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM174A
Entrez Gene ID
345757
Gene Name
family with sequence similarity 174 member A
Gene Aliases
HGS_RE408, TMEM157, UNQ1912
UniGene
Hs.356108
Gene Chromosome Location
Chr.5: 100535305-100586741 on Build GRCh38
Transcripts
NM_198507.2 XM_006714600.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3857063 c.143C>A p.P48H Chr.5: 100535673-100535673 on Build GRCh38
3857065 c.263T>C p.V88A Chr.5: 100535793-100535793 on Build GRCh38
3857064 c.201C>T p.G67G Chr.5: 100535731-100535731 on Build GRCh38
738844 c.306A>T p.E102D Chr.5: 100535836-100535836 on Build GRCh38
7313446 c.403G>T p.V135L Chr.5: 100535933-100535933 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs893610292 Chr.5: 100536134-100536134 on Build GRCh38
rs759880540 Chr.5: 100535947-100535947 on Build GRCh38
rs991987171 Chr.5: 100535942-100535942 on Build GRCh38
rs375162382 Chr.5: 100535731-100535731 on Build GRCh38
rs763670180 Chr.5: 100535826-100535826 on Build GRCh38
rs773628854 Chr.5: 100535775-100535775 on Build GRCh38
rs761872265 Chr.5: 100536121-100536121 on Build GRCh38
rs779426117 Chr.5: 100535736-100535736 on Build GRCh38
rs749237204 Chr.5: 100535894-100535894 on Build GRCh38
rs960008278 Chr.5: 100535914-100535914 on Build GRCh38
rs755921624 Chr.5: 100535981-100535981 on Build GRCh38
rs756741908 Chr.5: 100535964-100535964 on Build GRCh38
rs767202041 Chr.5: 100535973-100535973 on Build GRCh38
rs574798483 Chr.5: 100536082-100536082 on Build GRCh38
rs113665794 Chr.5: 100535752-100535752 on Build GRCh38
rs996465616 Chr.5: 100535694-100535694 on Build GRCh38
rs761188844 Chr.5: 100535698-100535698 on Build GRCh38
rs760994288 Chr.5: 100535794-100535794 on Build GRCh38
rs372797030 Chr.5: 100535900-100535900 on Build GRCh38
rs11548396 Chr.5: 100535789-100535789 on Build GRCh38
rs745626051 Chr.5: 100535758-100535758 on Build GRCh38
rs753238312 Chr.5: 100535820-100535820 on Build GRCh38
rs775952388 Chr.5: 100536015-100536015 on Build GRCh38
rs556251875 Chr.5: 100536052-100536052 on Build GRCh38
rs764547900 Chr.5: 100535958-100535958 on Build GRCh38
rs757890212 Chr.5: 100535751-100535751 on Build GRCh38
rs369657992 Chr.5: 100535766-100535766 on Build GRCh38
rs149747209 Chr.5: 100536110-100536110 on Build GRCh38
rs115404031 Chr.5: 100536008-100536008 on Build GRCh38
rs772189882 Chr.5: 100535767-100535767 on Build GRCh38
rs375418043 Chr.5: 100535885-100535885 on Build GRCh38
rs751095320 Chr.5: 100535961-100535961 on Build GRCh38
rs754275569 Chr.5: 100535810-100535810 on Build GRCh38
rs765617668 Chr.5: 100535948-100535948 on Build GRCh38
rs113914887 Chr.5: 100535763-100535763 on Build GRCh38
rs201541686 Chr.5: 100535895-100535895 on Build GRCh38
rs771326074 Chr.5: 100535936-100535936 on Build GRCh38
rs185049199 Chr.5: 100535992-100535992 on Build GRCh38
rs372498908 Chr.5: 100536000-100536000 on Build GRCh38
rs754199628 Chr.5: 100535719-100535719 on Build GRCh38
rs531483245 Chr.5: 100535818-100535818 on Build GRCh38
rs770904098 Chr.5: 100535874-100535874 on Build GRCh38
rs776908556 Chr.5: 100535959-100535959 on Build GRCh38
rs746111753 Chr.5: 100536002-100536002 on Build GRCh38
rs770197899 Chr.5: 100536017-100536017 on Build GRCh38
rs750071967 Chr.5: 100535977-100535977 on Build GRCh38
rs200948075 Chr.5: 100535980-100535980 on Build GRCh38
rs932985306 Chr.5: 100536033-100536033 on Build GRCh38
rs1009430698 Chr.5: 100536149-100536149 on Build GRCh38
rs372245282 Chr.5: 100535692-100535692 on Build GRCh38
rs766838722 Chr.5: 100535709-100535709 on Build GRCh38
rs776960906 Chr.5: 100535937-100535937 on Build GRCh38
rs369516601 Chr.5: 100535993-100535993 on Build GRCh38
rs570853705 Chr.5: 100536031-100536031 on Build GRCh38
rs1051443550 Chr.5: 100535728-100535728 on Build GRCh38
rs751002251 Chr.5: 100535846-100535846 on Build GRCh38
rs777505637 Chr.5: 100535750-100535750 on Build GRCh38
rs766666353 Chr.5: 100535797-100535797 on Build GRCh38
rs747035012 Chr.5: 100535930-100535930 on Build GRCh38
rs554030894 Chr.5: 100536114-100536114 on Build GRCh38
rs745716053 Chr.5: 100535870-100535870 on Build GRCh38
rs760119092 Chr.5: 100535695-100535695 on Build GRCh38
rs771129761 Chr.5: 100535996-100535996 on Build GRCh38
rs546024871 Chr.5: 100535755-100535755 on Build GRCh38
rs763518688 Chr.5: 100535951-100535951 on Build GRCh38
rs770510840 Chr.5: 100535754-100535754 on Build GRCh38
rs538347940 Chr.5: 100536035-100536035 on Build GRCh38
rs765834127 Chr.5: 100535772-100535791 on Build GRCh38
rs566008413 Chr.5: 100535989-100535989 on Build GRCh38
rs1011149361 Chr.5: 100535838-100535838 on Build GRCh38
rs756936364 Chr.5: 100535855-100535855 on Build GRCh38
rs372056361 Chr.5: 100535824-100535824 on Build GRCh38
rs755911105 Chr.5: 100535871-100535871 on Build GRCh38
rs796598051 Chr.5: 100535849-100535849 on Build GRCh38
rs774173259 Chr.5: 100536009-100536009 on Build GRCh38
rs769645263 Chr.5: 100535760-100535760 on Build GRCh38
rs780777327 Chr.5: 100535859-100535859 on Build GRCh38
rs139597477 Chr.5: 100535814-100535814 on Build GRCh38
rs773432657 Chr.5: 100535897-100535897 on Build GRCh38
rs969694870 Chr.5: 100535872-100535872 on Build GRCh38
rs779806256 Chr.5: 100535708-100535708 on Build GRCh38
rs746647890 Chr.5: 100535753-100535753 on Build GRCh38

View Full Product Details