Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 111224057-111224593 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMK4
Entrez Gene ID
814
Gene Name
calcium/calmodulin dependent protein kinase IV
Gene Aliases
CaMK IV, CaMK-GR, caMK
UniGene
Hs.438801
Gene Chromosome Location
Chr.5: 111223653-111498503 on Build GRCh38
Transcripts
NM_001323374.1 NM_001323375.1 NM_001323376.1 NM_001323377.1 NM_001744.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5356597 c.106G>A p.G36S Chr.5: 111224589-111224589 on Build GRCh38
6027106 c.54C>T p.T18T Chr.5: 111224537-111224537 on Build GRCh38
4668955 c.44C>A p.S15Y Chr.5: 111224527-111224527 on Build GRCh38
5528088 c.88C>T p.P30S Chr.5: 111224571-111224571 on Build GRCh38
7144506 c.103G>T p.D35Y Chr.5: 111224586-111224586 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756016616 Chr.5: 111224451-111224451 on Build GRCh38
rs1047143854 Chr.5: 111224415-111224415 on Build GRCh38
rs1016907284 Chr.5: 111224373-111224373 on Build GRCh38
rs1030523198 Chr.5: 111224115-111224115 on Build GRCh38
rs749183758 Chr.5: 111224277-111224277 on Build GRCh38
rs960484551 Chr.5: 111224386-111224386 on Build GRCh38
rs943219574 Chr.5: 111224288-111224288 on Build GRCh38
rs781692773 Chr.5: 111224480-111224480 on Build GRCh38
rs768721428 Chr.5: 111224377-111224377 on Build GRCh38
rs767617440 Chr.5: 111224434-111224434 on Build GRCh38
rs1018080529 Chr.5: 111224128-111224128 on Build GRCh38
rs892751439 Chr.5: 111224082-111224082 on Build GRCh38
rs748901600 Chr.5: 111224499-111224499 on Build GRCh38
rs958333650 Chr.5: 111224454-111224454 on Build GRCh38
rs768256755 Chr.5: 111224502-111224502 on Build GRCh38
rs778631289 Chr.5: 111224440-111224448 on Build GRCh38
rs543574477 Chr.5: 111224179-111224179 on Build GRCh38
rs74294411 Chr.5: 111224123-111224123 on Build GRCh38
rs530474251 Chr.5: 111224403-111224403 on Build GRCh38
rs115090272 Chr.5: 111224385-111224385 on Build GRCh38
rs958172344 Chr.5: 111224374-111224374 on Build GRCh38
rs1052537444 Chr.5: 111224410-111224410 on Build GRCh38
rs746404317 Chr.5: 111224490-111224490 on Build GRCh38
rs772826235 Chr.5: 111224440-111224442 on Build GRCh38
rs111716534 Chr.5: 111224167-111224167 on Build GRCh38
rs936659347 Chr.5: 111224202-111224202 on Build GRCh38
rs576097029 Chr.5: 111224177-111224177 on Build GRCh38
rs1026273887 Chr.5: 111224334-111224334 on Build GRCh38
rs913568832 Chr.5: 111224240-111224240 on Build GRCh38
rs569750472 Chr.5: 111224413-111224413 on Build GRCh38
rs926385595 Chr.5: 111224204-111224204 on Build GRCh38
rs199721159 Chr.5: 111224554-111224554 on Build GRCh38
rs758643186 Chr.5: 111224522-111224522 on Build GRCh38
rs73230459 Chr.5: 111224087-111224087 on Build GRCh38
rs530657052 Chr.5: 111224408-111224409 on Build GRCh38
rs1026262549 Chr.5: 111224166-111224166 on Build GRCh38
rs775728276 Chr.5: 111224498-111224498 on Build GRCh38
rs145633003 Chr.5: 111224530-111224530 on Build GRCh38
rs773893938 Chr.5: 111224504-111224504 on Build GRCh38
rs759969062 Chr.5: 111224439-111224440 on Build GRCh38
rs943648847 Chr.5: 111224556-111224556 on Build GRCh38
rs1044003289 Chr.5: 111224244-111224244 on Build GRCh38
rs903646674 Chr.5: 111224120-111224120 on Build GRCh38
rs1053111992 Chr.5: 111224079-111224079 on Build GRCh38
rs1035863406 Chr.5: 111224523-111224523 on Build GRCh38
rs750225738 Chr.5: 111224447-111224447 on Build GRCh38
rs777843446 Chr.5: 111224470-111224470 on Build GRCh38
rs111263260 Chr.5: 111224438-111224438 on Build GRCh38
rs760714420 Chr.5: 111224518-111224518 on Build GRCh38
rs898223578 Chr.5: 111224411-111224411 on Build GRCh38
rs886777897 Chr.5: 111224423-111224423 on Build GRCh38
rs1051850901 Chr.5: 111224246-111224246 on Build GRCh38
rs987467299 Chr.5: 111224534-111224534 on Build GRCh38
rs868009254 Chr.5: 111224390-111224390 on Build GRCh38
rs532077628 Chr.5: 111224276-111224276 on Build GRCh38
rs960162314 Chr.5: 111224540-111224540 on Build GRCh38
rs760500088 Chr.5: 111224442-111224443 on Build GRCh38
rs1039425879 Chr.5: 111224296-111224296 on Build GRCh38
rs570938398 Chr.5: 111224537-111224537 on Build GRCh38
rs367616588 Chr.5: 111224514-111224514 on Build GRCh38
rs564950082 Chr.5: 111224193-111224193 on Build GRCh38
rs747839230 Chr.5: 111224440-111224445 on Build GRCh38
rs995569661 Chr.5: 111224317-111224317 on Build GRCh38
rs544500715 Chr.5: 111224173-111224173 on Build GRCh38
rs980708866 Chr.5: 111224382-111224383 on Build GRCh38
rs980566270 Chr.5: 111224187-111224187 on Build GRCh38
rs749668332 Chr.5: 111224560-111224560 on Build GRCh38
rs753581560 Chr.5: 111224521-111224521 on Build GRCh38
rs1006555779 Chr.5: 111224285-111224285 on Build GRCh38
rs761462897 Chr.5: 111224512-111224512 on Build GRCh38
rs935139697 Chr.5: 111224324-111224324 on Build GRCh38
rs945316219 Chr.5: 111224243-111224243 on Build GRCh38
rs879312645 Chr.5: 111224327-111224327 on Build GRCh38
rs1052474279 Chr.5: 111224212-111224212 on Build GRCh38
rs370357101 Chr.5: 111224545-111224545 on Build GRCh38
rs564921572 Chr.5: 111224220-111224220 on Build GRCh38
rs910792342 Chr.5: 111224552-111224552 on Build GRCh38
rs925344244 Chr.5: 111224360-111224360 on Build GRCh38
rs369076960 Chr.5: 111224395-111224395 on Build GRCh38
rs1006489154 Chr.5: 111224364-111224364 on Build GRCh38
rs913916885 Chr.5: 111224382-111224382 on Build GRCh38
rs942609543 Chr.5: 111224308-111224308 on Build GRCh38
rs943292524 Chr.5: 111224425-111224425 on Build GRCh38
rs951352861 Chr.5: 111224322-111224322 on Build GRCh38
rs542509226 Chr.5: 111224139-111224139 on Build GRCh38
rs910844672 Chr.5: 111224400-111224400 on Build GRCh38
rs914121100 Chr.5: 111224216-111224216 on Build GRCh38
rs571137042 Chr.5: 111224106-111224106 on Build GRCh38
rs752917380 Chr.5: 111224464-111224464 on Build GRCh38
rs757739319 Chr.5: 111224478-111224478 on Build GRCh38
rs969424568 Chr.5: 111224178-111224178 on Build GRCh38
rs951004536 Chr.5: 111224168-111224168 on Build GRCh38
rs751606904 Chr.5: 111224538-111224538 on Build GRCh38
rs757802886 Chr.5: 111224435-111224436 on Build GRCh38
rs1038880761 Chr.5: 111224431-111224431 on Build GRCh38
rs766121127 Chr.5: 111224463-111224463 on Build GRCh38
rs889290826 Chr.5: 111224255-111224255 on Build GRCh38
rs548464045 Chr.5: 111224409-111224409 on Build GRCh38
rs572314398 Chr.5: 111224134-111224134 on Build GRCh38
rs62376844 Chr.5: 111224501-111224501 on Build GRCh38
rs979322418 Chr.5: 111224314-111224314 on Build GRCh38
rs1025976285 Chr.5: 111224141-111224141 on Build GRCh38
rs767534006 Chr.5: 111224513-111224513 on Build GRCh38
rs768328095 Chr.5: 111224568-111224568 on Build GRCh38
rs1037864791 Chr.5: 111224404-111224404 on Build GRCh38
rs933605583 Chr.5: 111224239-111224239 on Build GRCh38
rs766141765 Chr.5: 111224519-111224519 on Build GRCh38
rs758564731 Chr.5: 111224467-111224467 on Build GRCh38
rs906144758 Chr.5: 111224506-111224506 on Build GRCh38
rs905137891 Chr.5: 111224442-111224442 on Build GRCh38
rs374698334 Chr.5: 111224278-111224278 on Build GRCh38
rs746314655 Chr.5: 111224546-111224546 on Build GRCh38
rs925396019 Chr.5: 111224189-111224189 on Build GRCh38
rs1024771725 Chr.5: 111224328-111224328 on Build GRCh38
rs770093750 Chr.5: 111224497-111224497 on Build GRCh38
rs939072918 Chr.5: 111224210-111224210 on Build GRCh38
rs925115899 Chr.5: 111224320-111224320 on Build GRCh38
rs997501519 Chr.5: 111224143-111224143 on Build GRCh38
rs896540832 Chr.5: 111224551-111224551 on Build GRCh38
rs747124636 Chr.5: 111224474-111224474 on Build GRCh38
rs541623904 Chr.5: 111224319-111224319 on Build GRCh38
rs111991449 Chr.5: 111224341-111224341 on Build GRCh38
rs980632387 Chr.5: 111224356-111224356 on Build GRCh38
rs898279793 Chr.5: 111224312-111224312 on Build GRCh38
rs1006608087 Chr.5: 111224121-111224121 on Build GRCh38
rs970603976 Chr.5: 111224186-111224186 on Build GRCh38
rs901969449 Chr.5: 111224326-111224326 on Build GRCh38
rs780459960 Chr.5: 111224559-111224559 on Build GRCh38
rs991858555 Chr.5: 111224217-111224217 on Build GRCh38
rs372340803 Chr.5: 111224267-111224267 on Build GRCh38
rs114063690 Chr.5: 111224119-111224119 on Build GRCh38
rs1013294846 Chr.5: 111224428-111224428 on Build GRCh38
rs532099051 Chr.5: 111224561-111224561 on Build GRCh38

View Full Product Details