Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 116293049-116293595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
UniGene
Hs.483136
Gene Chromosome Location
Chr.5: 116084592-116446462 on Build GRCh38
Transcripts
NM_001308080.1 NM_016144.3 XM_017009553.1

SNP Detail

SNP ID Chromosome Location
rs949811595 Chr.5: 116293225-116293225 on Build GRCh38
rs1005681511 Chr.5: 116293417-116293417 on Build GRCh38
rs949862492 Chr.5: 116293496-116293496 on Build GRCh38
rs1006169617 Chr.5: 116293560-116293560 on Build GRCh38
rs1045477843 Chr.5: 116293497-116293497 on Build GRCh38
rs188816426 Chr.5: 116293291-116293291 on Build GRCh38
rs537586038 Chr.5: 116293368-116293368 on Build GRCh38
rs972409972 Chr.5: 116293193-116293193 on Build GRCh38
rs770894963 Chr.5: 116293250-116293250 on Build GRCh38
rs1023019771 Chr.5: 116293405-116293405 on Build GRCh38
rs902873790 Chr.5: 116293263-116293263 on Build GRCh38
rs553690555 Chr.5: 116293509-116293509 on Build GRCh38
rs1015840409 Chr.5: 116293417-116293420 on Build GRCh38
rs951994494 Chr.5: 116293471-116293471 on Build GRCh38
rs911905979 Chr.5: 116293168-116293168 on Build GRCh38
rs987516006 Chr.5: 116293156-116293156 on Build GRCh38
rs712571 Chr.5: 116293461-116293461 on Build GRCh38
rs1029167938 Chr.5: 116293326-116293326 on Build GRCh38
rs184383090 Chr.5: 116293275-116293275 on Build GRCh38
rs779808852 Chr.5: 116293251-116293251 on Build GRCh38
rs922922246 Chr.5: 116293425-116293425 on Build GRCh38
rs573896350 Chr.5: 116293526-116293526 on Build GRCh38
rs745713396 Chr.5: 116293159-116293159 on Build GRCh38
rs6873096 Chr.5: 116293490-116293490 on Build GRCh38
rs547029575 Chr.5: 116293222-116293222 on Build GRCh38
rs369766155 Chr.5: 116293358-116293358 on Build GRCh38
rs947246810 Chr.5: 116293465-116293465 on Build GRCh38
rs569893804 Chr.5: 116293367-116293367 on Build GRCh38
rs986298295 Chr.5: 116293436-116293436 on Build GRCh38
rs923060925 Chr.5: 116293206-116293206 on Build GRCh38
rs191507349 Chr.5: 116293221-116293221 on Build GRCh38
rs754745062 Chr.5: 116293294-116293294 on Build GRCh38
rs953442762 Chr.5: 116293139-116293139 on Build GRCh38
rs895924545 Chr.5: 116293356-116293356 on Build GRCh38
rs372026937 Chr.5: 116293214-116293214 on Build GRCh38
rs560720817 Chr.5: 116293090-116293090 on Build GRCh38
rs952886109 Chr.5: 116293144-116293145 on Build GRCh38
rs140181352 Chr.5: 116293314-116293314 on Build GRCh38
rs144414023 Chr.5: 116293125-116293125 on Build GRCh38
rs562667158 Chr.5: 116293086-116293086 on Build GRCh38
rs954355498 Chr.5: 116293430-116293431 on Build GRCh38
rs1029523102 Chr.5: 116293096-116293096 on Build GRCh38
rs1013435699 Chr.5: 116293384-116293384 on Build GRCh38
rs934354208 Chr.5: 116293546-116293546 on Build GRCh38
rs746666855 Chr.5: 116293252-116293252 on Build GRCh38
rs527422430 Chr.5: 116293204-116293204 on Build GRCh38
rs1012883397 Chr.5: 116293543-116293543 on Build GRCh38
rs972077855 Chr.5: 116293417-116293417 on Build GRCh38
rs1011528833 Chr.5: 116293324-116293324 on Build GRCh38
rs903099119 Chr.5: 116293268-116293268 on Build GRCh38
rs1055623895 Chr.5: 116293220-116293220 on Build GRCh38
rs1057139914 Chr.5: 116293337-116293337 on Build GRCh38
rs1057046888 Chr.5: 116293494-116293494 on Build GRCh38
rs953002734 Chr.5: 116293416-116293417 on Build GRCh38
rs116583503 Chr.5: 116293188-116293188 on Build GRCh38
rs533558973 Chr.5: 116293491-116293491 on Build GRCh38
rs984791326 Chr.5: 116293158-116293158 on Build GRCh38
rs971733752 Chr.5: 116293423-116293423 on Build GRCh38
rs377330363 Chr.5: 116293467-116293468 on Build GRCh38
rs1054123369 Chr.5: 116293553-116293553 on Build GRCh38
rs768317889 Chr.5: 116293472-116293472 on Build GRCh38
rs542494930 Chr.5: 116293075-116293075 on Build GRCh38
rs892756717 Chr.5: 116293558-116293558 on Build GRCh38
rs756227245 Chr.5: 116293373-116293373 on Build GRCh38
rs1019017587 Chr.5: 116293372-116293372 on Build GRCh38
rs998457404 Chr.5: 116293272-116293272 on Build GRCh38
rs998890065 Chr.5: 116293316-116293316 on Build GRCh38

View Full Product Details