Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 118839733-118840263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTWD2
Entrez Gene ID
285605
Gene Name
DTW domain containing 2
Gene Aliases
-
UniGene
Hs.655891
Gene Chromosome Location
Chr.5: 118836074-118989938 on Build GRCh38
Transcripts
NM_001308081.1 NM_173666.3 XM_011543338.2 XM_011543340.2

SNP Detail

SNP ID Chromosome Location
rs187635645 Chr.5: 118840134-118840134 on Build GRCh38
rs776550017 Chr.5: 118839942-118839942 on Build GRCh38
rs369957116 Chr.5: 118840126-118840126 on Build GRCh38
rs879200327 Chr.5: 118839854-118839854 on Build GRCh38
rs750515710 Chr.5: 118839925-118839925 on Build GRCh38
rs915565546 Chr.5: 118840000-118840000 on Build GRCh38
rs1035578512 Chr.5: 118840206-118840206 on Build GRCh38
rs1036918748 Chr.5: 118839859-118839859 on Build GRCh38
rs999738252 Chr.5: 118840200-118840200 on Build GRCh38
rs902588432 Chr.5: 118840199-118840199 on Build GRCh38
rs1022436724 Chr.5: 118839932-118839932 on Build GRCh38
rs948462540 Chr.5: 118840001-118840001 on Build GRCh38
rs573882752 Chr.5: 118839897-118839897 on Build GRCh38
rs765095179 Chr.5: 118839819-118839819 on Build GRCh38
rs529858753 Chr.5: 118840117-118840117 on Build GRCh38
rs901113082 Chr.5: 118840109-118840109 on Build GRCh38
rs192223899 Chr.5: 118840210-118840210 on Build GRCh38
rs879894997 Chr.5: 118839969-118839969 on Build GRCh38
rs17132677 Chr.5: 118839979-118839979 on Build GRCh38
rs983834748 Chr.5: 118840057-118840057 on Build GRCh38
rs184051753 Chr.5: 118840222-118840222 on Build GRCh38
rs939936641 Chr.5: 118839822-118839822 on Build GRCh38
rs978757578 Chr.5: 118839957-118839957 on Build GRCh38
rs888251031 Chr.5: 118839808-118839808 on Build GRCh38
rs889761267 Chr.5: 118840149-118840149 on Build GRCh38
rs184456112 Chr.5: 118840111-118840111 on Build GRCh38
rs17132673 Chr.5: 118839945-118839945 on Build GRCh38
rs1010917886 Chr.5: 118839928-118839928 on Build GRCh38
rs73236915 Chr.5: 118840229-118840229 on Build GRCh38
rs1028262850 Chr.5: 118839921-118839921 on Build GRCh38
rs533595067 Chr.5: 118839835-118839835 on Build GRCh38
rs961014917 Chr.5: 118840209-118840209 on Build GRCh38
rs1022654269 Chr.5: 118840230-118840230 on Build GRCh38
rs939906498 Chr.5: 118840078-118840078 on Build GRCh38
rs369399613 Chr.5: 118839781-118839781 on Build GRCh38
rs553715013 Chr.5: 118839862-118839862 on Build GRCh38
rs1022301353 Chr.5: 118840166-118840166 on Build GRCh38
rs909716876 Chr.5: 118840077-118840078 on Build GRCh38
rs1010947529 Chr.5: 118840162-118840162 on Build GRCh38
rs901080564 Chr.5: 118839893-118839893 on Build GRCh38
rs1037272219 Chr.5: 118840089-118840089 on Build GRCh38
rs1048285045 Chr.5: 118839783-118839783 on Build GRCh38
rs992515055 Chr.5: 118839981-118839981 on Build GRCh38

View Full Product Details