Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119110014-119110505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
UniGene
Hs.181042
Gene Chromosome Location
Chr.5: 119071002-119250170 on Build GRCh38
Transcripts
NM_001290321.2 NM_001290322.2 NM_005509.5 XM_005271909.4 XM_005271910.4 XM_005271912.2 XM_011543212.2 XM_011543213.2 XM_011543214.2 XM_011543215.2 XM_017009141.1 XM_017009142.1 XM_017009143.1 XM_017009144.1 XM_017009145.1 XM_017009146.1 XM_017009147.1 XM_017009148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5105855 c.390C>G p.S130R Chr.5: 119110176-119110176 on Build GRCh38
7192671 c.369T>A p.S123R Chr.5: 119110155-119110155 on Build GRCh38
7141616 c.488G>T p.W163L Chr.5: 119110274-119110274 on Build GRCh38
7400793 c.436G>T p.D146Y Chr.5: 119110222-119110222 on Build GRCh38
6469844 c.407C>G p.S136C Chr.5: 119110193-119110193 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779083658 Chr.5: 119110313-119110313 on Build GRCh38
rs750676579 Chr.5: 119110404-119110404 on Build GRCh38
rs752555916 Chr.5: 119110121-119110121 on Build GRCh38
rs146514647 Chr.5: 119110153-119110153 on Build GRCh38
rs184385094 Chr.5: 119110435-119110435 on Build GRCh38
rs780317156 Chr.5: 119110290-119110290 on Build GRCh38
rs561110440 Chr.5: 119110319-119110321 on Build GRCh38
rs34420938 Chr.5: 119110054-119110054 on Build GRCh38
rs549446657 Chr.5: 119110291-119110291 on Build GRCh38
rs776067413 Chr.5: 119110198-119110198 on Build GRCh38
rs760252585 Chr.5: 119110193-119110193 on Build GRCh38
rs190524932 Chr.5: 119110442-119110442 on Build GRCh38
rs752499902 Chr.5: 119110353-119110353 on Build GRCh38
rs751308539 Chr.5: 119110259-119110259 on Build GRCh38
rs565844922 Chr.5: 119110382-119110382 on Build GRCh38
rs1034528475 Chr.5: 119110425-119110425 on Build GRCh38
rs562904417 Chr.5: 119110074-119110074 on Build GRCh38
rs772342945 Chr.5: 119110178-119110178 on Build GRCh38
rs769352967 Chr.5: 119110164-119110164 on Build GRCh38
rs776235199 Chr.5: 119110332-119110332 on Build GRCh38
rs534866770 Chr.5: 119110397-119110397 on Build GRCh38
rs759486687 Chr.5: 119110111-119110111 on Build GRCh38
rs763734872 Chr.5: 119110400-119110400 on Build GRCh38
rs755081908 Chr.5: 119110300-119110300 on Build GRCh38
rs371482414 Chr.5: 119110328-119110328 on Build GRCh38
rs748568077 Chr.5: 119110170-119110170 on Build GRCh38
rs767474937 Chr.5: 119110235-119110235 on Build GRCh38
rs921420234 Chr.5: 119110360-119110360 on Build GRCh38
rs547590187 Chr.5: 119110157-119110157 on Build GRCh38
rs757965604 Chr.5: 119110141-119110141 on Build GRCh38
rs748692839 Chr.5: 119110316-119110316 on Build GRCh38
rs947808661 Chr.5: 119110384-119110384 on Build GRCh38
rs778015272 Chr.5: 119110327-119110327 on Build GRCh38
rs900007456 Chr.5: 119110052-119110052 on Build GRCh38
rs576425967 Chr.5: 119110460-119110460 on Build GRCh38
rs756349296 Chr.5: 119110266-119110266 on Build GRCh38
rs184092263 Chr.5: 119110246-119110246 on Build GRCh38
rs1009847546 Chr.5: 119110392-119110392 on Build GRCh38
rs180999628 Chr.5: 119110422-119110422 on Build GRCh38
rs765840834 Chr.5: 119110106-119110106 on Build GRCh38
rs936869317 Chr.5: 119110378-119110378 on Build GRCh38
rs188137634 Chr.5: 119110352-119110352 on Build GRCh38
rs565432755 Chr.5: 119110128-119110128 on Build GRCh38
rs1019989191 Chr.5: 119110105-119110105 on Build GRCh38
rs750634781 Chr.5: 119110142-119110142 on Build GRCh38
rs772567479 Chr.5: 119110323-119110323 on Build GRCh38
rs1009880877 Chr.5: 119110104-119110104 on Build GRCh38
rs759517316 Chr.5: 119110196-119110196 on Build GRCh38
rs752609116 Chr.5: 119110237-119110237 on Build GRCh38
rs758158476 Chr.5: 119110133-119110133 on Build GRCh38
rs773812396 Chr.5: 119110076-119110076 on Build GRCh38
rs191604911 Chr.5: 119110152-119110152 on Build GRCh38
rs951310810 Chr.5: 119110476-119110476 on Build GRCh38
rs765149895 Chr.5: 119110120-119110120 on Build GRCh38
rs938891968 Chr.5: 119110330-119110330 on Build GRCh38
rs397883913 Chr.5: 119110061-119110061 on Build GRCh38
rs892759180 Chr.5: 119110386-119110386 on Build GRCh38
rs376312939 Chr.5: 119110297-119110297 on Build GRCh38
rs545085295 Chr.5: 119110463-119110463 on Build GRCh38
rs779527923 Chr.5: 119110169-119110169 on Build GRCh38
rs1023264646 Chr.5: 119110062-119110062 on Build GRCh38
rs529753640 Chr.5: 119110243-119110243 on Build GRCh38
rs1056133487 Chr.5: 119110345-119110345 on Build GRCh38
rs1032568023 Chr.5: 119110086-119110086 on Build GRCh38
rs531585725 Chr.5: 119110087-119110087 on Build GRCh38
rs919872500 Chr.5: 119110256-119110256 on Build GRCh38
rs759046996 Chr.5: 119110217-119110217 on Build GRCh38
rs879205085 Chr.5: 119110304-119110304 on Build GRCh38
rs897653188 Chr.5: 119110401-119110401 on Build GRCh38
rs763827730 Chr.5: 119110258-119110258 on Build GRCh38
rs376336568 Chr.5: 119110137-119110137 on Build GRCh38
rs374255427 Chr.5: 119110108-119110108 on Build GRCh38
rs201249787 Chr.5: 119110113-119110113 on Build GRCh38
rs981604844 Chr.5: 119110148-119110148 on Build GRCh38
rs574671004 Chr.5: 119110423-119110423 on Build GRCh38
rs199987228 Chr.5: 119110102-119110102 on Build GRCh38
rs200428684 Chr.5: 119110358-119110358 on Build GRCh38
rs567489539 Chr.5: 119110195-119110195 on Build GRCh38
rs773583019 Chr.5: 119110188-119110188 on Build GRCh38

View Full Product Details