Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 119118704-119119253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DMXL1
Entrez Gene ID
1657
Gene Name
Dmx like 1
Gene Aliases
-
UniGene
Hs.181042
Gene Chromosome Location
Chr.5: 119071002-119250170 on Build GRCh38
Transcripts
NM_001290321.2 NM_001290322.2 NM_005509.5 XM_005271909.4 XM_005271910.4 XM_005271912.2 XM_011543212.2 XM_011543213.2 XM_011543214.2 XM_011543215.2 XM_017009141.1 XM_017009142.1 XM_017009143.1 XM_017009144.1 XM_017009145.1 XM_017009146.1 XM_017009147.1 XM_017009148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3134415 c.833T>C p.L278P Chr.5: 119118904-119118904 on Build GRCh38
3134417 c.910G>T p.E304* Chr.5: 119118981-119118981 on Build GRCh38
7417546 c.854A>T p.H285L Chr.5: 119118925-119118925 on Build GRCh38
6168463 c.868A>G p.I290V Chr.5: 119118939-119118939 on Build GRCh38
5829999 c.756T>C p.C252C Chr.5: 119118827-119118827 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375271214 Chr.5: 119119025-119119025 on Build GRCh38
rs933756895 Chr.5: 119119084-119119085 on Build GRCh38
rs763705818 Chr.5: 119118824-119118824 on Build GRCh38
rs749067891 Chr.5: 119118978-119118978 on Build GRCh38
rs551057796 Chr.5: 119119112-119119112 on Build GRCh38
rs946623469 Chr.5: 119119110-119119110 on Build GRCh38
rs930237289 Chr.5: 119119207-119119207 on Build GRCh38
rs201520761 Chr.5: 119118801-119118801 on Build GRCh38
rs72784079 Chr.5: 119118751-119118751 on Build GRCh38
rs779854170 Chr.5: 119118857-119118857 on Build GRCh38
rs922333886 Chr.5: 119119084-119119084 on Build GRCh38
rs776127486 Chr.5: 119118895-119118895 on Build GRCh38
rs974293420 Chr.5: 119119163-119119163 on Build GRCh38
rs776982814 Chr.5: 119118907-119118907 on Build GRCh38
rs144725692 Chr.5: 119119094-119119094 on Build GRCh38
rs765552138 Chr.5: 119118911-119118911 on Build GRCh38
rs922361817 Chr.5: 119118747-119118747 on Build GRCh38
rs140972662 Chr.5: 119118920-119118920 on Build GRCh38
rs767000856 Chr.5: 119118948-119118948 on Build GRCh38
rs544699912 Chr.5: 119118760-119118760 on Build GRCh38
rs772679787 Chr.5: 119118748-119118748 on Build GRCh38
rs770938719 Chr.5: 119119001-119119001 on Build GRCh38
rs985097366 Chr.5: 119119022-119119022 on Build GRCh38
rs1047389401 Chr.5: 119118931-119118931 on Build GRCh38
rs141939679 Chr.5: 119118820-119118820 on Build GRCh38
rs773205071 Chr.5: 119118795-119118795 on Build GRCh38
rs886155994 Chr.5: 119118985-119118985 on Build GRCh38
rs766937787 Chr.5: 119119055-119119055 on Build GRCh38
rs748374686 Chr.5: 119118873-119118873 on Build GRCh38
rs774412610 Chr.5: 119118776-119118776 on Build GRCh38
rs761325204 Chr.5: 119119028-119119028 on Build GRCh38
rs777942863 Chr.5: 119118992-119118992 on Build GRCh38
rs781056671 Chr.5: 119118770-119118770 on Build GRCh38
rs761180932 Chr.5: 119118802-119118802 on Build GRCh38
rs778558865 Chr.5: 119118750-119118750 on Build GRCh38
rs779836749 Chr.5: 119118976-119118976 on Build GRCh38
rs367822456 Chr.5: 119118898-119118898 on Build GRCh38
rs748319783 Chr.5: 119118779-119118779 on Build GRCh38
rs758685188 Chr.5: 119118984-119118984 on Build GRCh38
rs902430480 Chr.5: 119119133-119119133 on Build GRCh38
rs755880455 Chr.5: 119118850-119118850 on Build GRCh38
rs775934966 Chr.5: 119118916-119118916 on Build GRCh38
rs750355441 Chr.5: 119118957-119118957 on Build GRCh38
rs376204152 Chr.5: 119118973-119118973 on Build GRCh38
rs759790134 Chr.5: 119118804-119118804 on Build GRCh38
rs927442349 Chr.5: 119119147-119119147 on Build GRCh38
rs762926803 Chr.5: 119119200-119119200 on Build GRCh38
rs751059947 Chr.5: 119118829-119118829 on Build GRCh38
rs370839757 Chr.5: 119119083-119119083 on Build GRCh38
rs746370261 Chr.5: 119119012-119119012 on Build GRCh38
rs59649653 Chr.5: 119118901-119118901 on Build GRCh38
rs939766246 Chr.5: 119118739-119118739 on Build GRCh38
rs766294288 Chr.5: 119119206-119119206 on Build GRCh38
rs776356918 Chr.5: 119118994-119118994 on Build GRCh38
rs958937956 Chr.5: 119119149-119119149 on Build GRCh38
rs1032959573 Chr.5: 119119059-119119059 on Build GRCh38
rs758087682 Chr.5: 119118817-119118817 on Build GRCh38
rs889672368 Chr.5: 119118862-119118862 on Build GRCh38
rs79099492 Chr.5: 119119076-119119076 on Build GRCh38
rs376075158 Chr.5: 119118831-119118831 on Build GRCh38
rs570951271 Chr.5: 119119127-119119127 on Build GRCh38
rs770281161 Chr.5: 119119021-119119021 on Build GRCh38
rs566166382 Chr.5: 119119096-119119096 on Build GRCh38
rs780110326 Chr.5: 119118867-119118868 on Build GRCh38
rs542053975 Chr.5: 119118747-119118749 on Build GRCh38
rs780958245 Chr.5: 119118833-119118833 on Build GRCh38
rs371066411 Chr.5: 119118748-119118750 on Build GRCh38
rs547531689 Chr.5: 119118757-119118757 on Build GRCh38
rs764989768 Chr.5: 119118809-119118809 on Build GRCh38
rs191440572 Chr.5: 119119030-119119030 on Build GRCh38
rs756696696 Chr.5: 119118934-119118934 on Build GRCh38
rs996482770 Chr.5: 119119045-119119045 on Build GRCh38
rs762711155 Chr.5: 119118918-119118918 on Build GRCh38
rs535815460 Chr.5: 119118761-119118770 on Build GRCh38
rs957384842 Chr.5: 119118754-119118754 on Build GRCh38
rs981574777 Chr.5: 119119142-119119142 on Build GRCh38
rs560633408 Chr.5: 119118921-119118921 on Build GRCh38
rs372998467 Chr.5: 119118908-119118908 on Build GRCh38
rs766779715 Chr.5: 119118803-119118803 on Build GRCh38
rs981990565 Chr.5: 119118851-119118851 on Build GRCh38
rs373143457 Chr.5: 119119002-119119002 on Build GRCh38
rs200933738 Chr.5: 119118904-119118904 on Build GRCh38
rs752421311 Chr.5: 119118812-119118812 on Build GRCh38
rs747393980 Chr.5: 119118876-119118876 on Build GRCh38
rs745721380 Chr.5: 119118839-119118839 on Build GRCh38
rs772678680 Chr.5: 119119185-119119185 on Build GRCh38
rs772166376 Chr.5: 119118787-119118787 on Build GRCh38
rs761483906 Chr.5: 119119039-119119039 on Build GRCh38
rs771511327 Chr.5: 119118905-119118905 on Build GRCh38
rs957353650 Chr.5: 119119091-119119091 on Build GRCh38
rs200673180 Chr.5: 119118875-119118875 on Build GRCh38
rs549335030 Chr.5: 119119032-119119032 on Build GRCh38
rs747460415 Chr.5: 119119081-119119081 on Build GRCh38
rs768961476 Chr.5: 119118772-119118772 on Build GRCh38
rs781291695 Chr.5: 119119004-119119004 on Build GRCh38

View Full Product Details