Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127043650-127044166 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
UniGene
Hs.49573
Gene Chromosome Location
Chr.5: 127040752-127073539 on Build GRCh38
Transcripts
NM_001164479.1

SNP Detail

SNP ID Chromosome Location
rs6872893 Chr.5: 127043974-127043974 on Build GRCh38
rs150126703 Chr.5: 127043707-127043707 on Build GRCh38
rs745531920 Chr.5: 127043830-127043830 on Build GRCh38
rs34758773 Chr.5: 127043891-127043891 on Build GRCh38
rs984060030 Chr.5: 127043787-127043787 on Build GRCh38
rs763492245 Chr.5: 127044055-127044055 on Build GRCh38
rs556200115 Chr.5: 127043967-127043967 on Build GRCh38
rs148826828 Chr.5: 127043761-127043761 on Build GRCh38
rs905839662 Chr.5: 127044022-127044022 on Build GRCh38
rs193104436 Chr.5: 127043897-127043897 on Build GRCh38
rs955361676 Chr.5: 127043739-127043739 on Build GRCh38
rs996876309 Chr.5: 127043959-127043959 on Build GRCh38
rs933479232 Chr.5: 127043941-127043941 on Build GRCh38
rs559145676 Chr.5: 127043763-127043763 on Build GRCh38
rs570101220 Chr.5: 127043949-127043949 on Build GRCh38
rs543018900 Chr.5: 127043824-127043824 on Build GRCh38
rs539094199 Chr.5: 127043965-127043965 on Build GRCh38
rs770702669 Chr.5: 127043677-127043677 on Build GRCh38
rs375423355 Chr.5: 127043999-127043999 on Build GRCh38
rs895903419 Chr.5: 127044084-127044084 on Build GRCh38
rs922345853 Chr.5: 127043940-127043940 on Build GRCh38
rs910886219 Chr.5: 127043806-127043806 on Build GRCh38
rs138417830 Chr.5: 127043760-127043760 on Build GRCh38
rs145206725 Chr.5: 127043814-127043814 on Build GRCh38
rs1057195876 Chr.5: 127044074-127044074 on Build GRCh38
rs994294978 Chr.5: 127044089-127044089 on Build GRCh38
rs1033739322 Chr.5: 127043799-127043799 on Build GRCh38
rs947637853 Chr.5: 127043987-127043987 on Build GRCh38
rs967874551 Chr.5: 127044106-127044106 on Build GRCh38
rs1029594469 Chr.5: 127043732-127043732 on Build GRCh38
rs979089138 Chr.5: 127043777-127043777 on Build GRCh38
rs988016818 Chr.5: 127043747-127043747 on Build GRCh38
rs899817325 Chr.5: 127043951-127043951 on Build GRCh38
rs896085492 Chr.5: 127043873-127043873 on Build GRCh38
rs572911262 Chr.5: 127043946-127043946 on Build GRCh38
rs950244751 Chr.5: 127043882-127043883 on Build GRCh38
rs551701931 Chr.5: 127043945-127043945 on Build GRCh38
rs926237800 Chr.5: 127043785-127043785 on Build GRCh38
rs1025466507 Chr.5: 127044125-127044125 on Build GRCh38
rs1043324901 Chr.5: 127043994-127043994 on Build GRCh38

View Full Product Details