Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 127046053-127046586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf63
Entrez Gene ID
401207
Gene Name
chromosome 5 open reading frame 63
Gene Aliases
YDR286C
UniGene
Hs.49573
Gene Chromosome Location
Chr.5: 127040752-127073539 on Build GRCh38
Transcripts
NM_001164479.1

SNP Detail

SNP ID Chromosome Location
rs1032416769 Chr.5: 127046375-127046375 on Build GRCh38
rs528148079 Chr.5: 127046547-127046547 on Build GRCh38
rs535149310 Chr.5: 127046391-127046391 on Build GRCh38
rs145318238 Chr.5: 127046276-127046276 on Build GRCh38
rs764378881 Chr.5: 127046194-127046194 on Build GRCh38
rs1037526293 Chr.5: 127046511-127046511 on Build GRCh38
rs940509840 Chr.5: 127046509-127046509 on Build GRCh38
rs147640333 Chr.5: 127046378-127046378 on Build GRCh38
rs766570822 Chr.5: 127046106-127046106 on Build GRCh38
rs34779298 Chr.5: 127046109-127046110 on Build GRCh38
rs779073316 Chr.5: 127046521-127046521 on Build GRCh38
rs896162545 Chr.5: 127046191-127046191 on Build GRCh38
rs59584486 Chr.5: 127046081-127046081 on Build GRCh38
rs917582862 Chr.5: 127046540-127046540 on Build GRCh38
rs955595279 Chr.5: 127046337-127046337 on Build GRCh38
rs937923297 Chr.5: 127046519-127046519 on Build GRCh38
rs887239580 Chr.5: 127046262-127046262 on Build GRCh38
rs571348381 Chr.5: 127046550-127046550 on Build GRCh38
rs937643836 Chr.5: 127046179-127046179 on Build GRCh38
rs898942369 Chr.5: 127046086-127046086 on Build GRCh38
rs760866277 Chr.5: 127046082-127046082 on Build GRCh38
rs916530254 Chr.5: 127046431-127046431 on Build GRCh38
rs572965519 Chr.5: 127046131-127046131 on Build GRCh38
rs982407746 Chr.5: 127046485-127046485 on Build GRCh38
rs931761021 Chr.5: 127046523-127046523 on Build GRCh38
rs979245907 Chr.5: 127046470-127046470 on Build GRCh38
rs970762995 Chr.5: 127046475-127046475 on Build GRCh38
rs59500308 Chr.5: 127046549-127046549 on Build GRCh38
rs575841260 Chr.5: 127046198-127046198 on Build GRCh38
rs1050183727 Chr.5: 127046537-127046537 on Build GRCh38
rs58371737 Chr.5: 127046187-127046187 on Build GRCh38
rs77125592 Chr.5: 127046178-127046178 on Build GRCh38
rs558216424 Chr.5: 127046396-127046396 on Build GRCh38
rs1047546207 Chr.5: 127046220-127046220 on Build GRCh38
rs544563716 Chr.5: 127046228-127046228 on Build GRCh38
rs907814821 Chr.5: 127046502-127046506 on Build GRCh38
rs945222270 Chr.5: 127046108-127046108 on Build GRCh38
rs1017169673 Chr.5: 127046278-127046278 on Build GRCh38
rs925145987 Chr.5: 127046510-127046510 on Build GRCh38
rs997531609 Chr.5: 127046314-127046314 on Build GRCh38
rs530439515 Chr.5: 127046351-127046351 on Build GRCh38
rs750271371 Chr.5: 127046245-127046245 on Build GRCh38
rs1028323771 Chr.5: 127046329-127046329 on Build GRCh38
rs559401523 Chr.5: 127046506-127046506 on Build GRCh38
rs966490477 Chr.5: 127046427-127046427 on Build GRCh38
rs552969808 Chr.5: 127046123-127046123 on Build GRCh38
rs962980964 Chr.5: 127046308-127046308 on Build GRCh38
rs1008854714 Chr.5: 127046190-127046190 on Build GRCh38

View Full Product Details