Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 130185712-130186219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY3
Entrez Gene ID
337876
Gene Name
chondroitin sulfate synthase 3
Gene Aliases
CHSY2, CSS3
UniGene
Hs.213137
Gene Chromosome Location
Chr.5: 129903415-130186637 on Build GRCh38
Transcripts
NM_175856.4 XM_011543363.2 XM_011543364.2 XM_017009434.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776991 c.2635C>T p.R879* Chr.5: 130185777-130185777 on Build GRCh38
1165852 c.2574C>T p.F858F Chr.5: 130185716-130185716 on Build GRCh38
4453188 c.2632A>G p.N878D Chr.5: 130185774-130185774 on Build GRCh38
5918609 c.2638A>G p.T880A Chr.5: 130185780-130185780 on Build GRCh38
6482837 c.2615A>C p.H872P Chr.5: 130185757-130185757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008257755 Chr.5: 130185785-130185785 on Build GRCh38
rs115392300 Chr.5: 130185845-130185845 on Build GRCh38
rs781636623 Chr.5: 130185927-130185927 on Build GRCh38
rs73788439 Chr.5: 130186144-130186144 on Build GRCh38
rs890931839 Chr.5: 130185964-130185964 on Build GRCh38
rs377277338 Chr.5: 130186115-130186115 on Build GRCh38
rs767995342 Chr.5: 130185766-130185766 on Build GRCh38
rs916659883 Chr.5: 130186106-130186106 on Build GRCh38
rs940322621 Chr.5: 130186020-130186020 on Build GRCh38
rs763106343 Chr.5: 130185822-130185822 on Build GRCh38
rs999817040 Chr.5: 130185887-130185887 on Build GRCh38
rs958242273 Chr.5: 130185864-130185864 on Build GRCh38
rs777029388 Chr.5: 130185778-130185778 on Build GRCh38
rs774062228 Chr.5: 130185774-130185774 on Build GRCh38
rs991372656 Chr.5: 130185870-130185870 on Build GRCh38
rs561079307 Chr.5: 130185755-130185755 on Build GRCh38
rs748868435 Chr.5: 130185759-130185759 on Build GRCh38
rs1019669451 Chr.5: 130186143-130186143 on Build GRCh38
rs77918956 Chr.5: 130186083-130186083 on Build GRCh38
rs1026611841 Chr.5: 130186015-130186015 on Build GRCh38
rs554684476 Chr.5: 130186071-130186071 on Build GRCh38
rs981775245 Chr.5: 130185978-130185978 on Build GRCh38
rs780105499 Chr.5: 130185758-130185758 on Build GRCh38
rs889847315 Chr.5: 130185780-130185780 on Build GRCh38
rs187472554 Chr.5: 130185875-130185875 on Build GRCh38
rs780908345 Chr.5: 130186036-130186039 on Build GRCh38
rs996378544 Chr.5: 130185761-130185761 on Build GRCh38
rs1043795444 Chr.5: 130186142-130186142 on Build GRCh38
rs201876337 Chr.5: 130185777-130185777 on Build GRCh38
rs183232436 Chr.5: 130185953-130185953 on Build GRCh38
rs963822632 Chr.5: 130185995-130185995 on Build GRCh38
rs568828313 Chr.5: 130185989-130185989 on Build GRCh38
rs10070570 Chr.5: 130186084-130186084 on Build GRCh38
rs542306046 Chr.5: 130186002-130186002 on Build GRCh38
rs764013289 Chr.5: 130185841-130185841 on Build GRCh38
rs745818019 Chr.5: 130185749-130185749 on Build GRCh38
rs554019827 Chr.5: 130186112-130186112 on Build GRCh38
rs1052379341 Chr.5: 130185905-130185905 on Build GRCh38
rs757694920 Chr.5: 130185827-130185827 on Build GRCh38
rs142213623 Chr.5: 130186134-130186134 on Build GRCh38
rs539716660 Chr.5: 130186139-130186139 on Build GRCh38
rs746669694 Chr.5: 130185837-130185838 on Build GRCh38
rs1018024145 Chr.5: 130185993-130185993 on Build GRCh38
rs369922143 Chr.5: 130186170-130186170 on Build GRCh38
rs761335482 Chr.5: 130185775-130185775 on Build GRCh38
rs187478194 Chr.5: 130186045-130186045 on Build GRCh38
rs879931680 Chr.5: 130185763-130185763 on Build GRCh38
rs948048154 Chr.5: 130185814-130185815 on Build GRCh38
rs192672097 Chr.5: 130185911-130185911 on Build GRCh38
rs530208922 Chr.5: 130185794-130185794 on Build GRCh38
rs992371729 Chr.5: 130186099-130186099 on Build GRCh38
rs76526369 Chr.5: 130186013-130186013 on Build GRCh38
rs371632478 Chr.5: 130185804-130185804 on Build GRCh38
rs78253116 Chr.5: 130185747-130185747 on Build GRCh38
rs192031558 Chr.5: 130186047-130186047 on Build GRCh38

View Full Product Details