Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 130186538-130187047 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHSY3
Entrez Gene ID
337876
Gene Name
chondroitin sulfate synthase 3
Gene Aliases
CHSY2, CSS3
UniGene
Hs.213137
Gene Chromosome Location
Chr.5: 129903415-130186637 on Build GRCh38
Transcripts
NM_175856.4 XM_011543363.2 XM_011543364.2 XM_017009434.1

SNP Detail

SNP ID Chromosome Location
rs144177824 Chr.5: 130186832-130186832 on Build GRCh38
rs911781701 Chr.5: 130186720-130186720 on Build GRCh38
rs912531273 Chr.5: 130186906-130186906 on Build GRCh38
rs576279177 Chr.5: 130186996-130186996 on Build GRCh38
rs1004282236 Chr.5: 130186754-130186754 on Build GRCh38
rs924050505 Chr.5: 130186800-130186800 on Build GRCh38
rs912467378 Chr.5: 130186631-130186631 on Build GRCh38
rs1053830346 Chr.5: 130186813-130186813 on Build GRCh38
rs539289378 Chr.5: 130186904-130186904 on Build GRCh38
rs1041552255 Chr.5: 130186787-130186787 on Build GRCh38
rs931125697 Chr.5: 130186942-130186942 on Build GRCh38
rs935318263 Chr.5: 130186612-130186612 on Build GRCh38
rs12520897 Chr.5: 130186579-130186579 on Build GRCh38
rs1032834761 Chr.5: 130186851-130186851 on Build GRCh38
rs1003637890 Chr.5: 130186628-130186628 on Build GRCh38
rs979465036 Chr.5: 130186576-130186576 on Build GRCh38
rs920248183 Chr.5: 130186684-130186684 on Build GRCh38
rs184510449 Chr.5: 130186679-130186679 on Build GRCh38
rs994994502 Chr.5: 130186974-130186974 on Build GRCh38
rs1045303457 Chr.5: 130186821-130186821 on Build GRCh38
rs961752951 Chr.5: 130186671-130186671 on Build GRCh38
rs944538506 Chr.5: 130186753-130186753 on Build GRCh38
rs975308903 Chr.5: 130186932-130186932 on Build GRCh38
rs764340026 Chr.5: 130186975-130186975 on Build GRCh38
rs531310948 Chr.5: 130186653-130186653 on Build GRCh38
rs1006791125 Chr.5: 130186740-130186769 on Build GRCh38
rs970878871 Chr.5: 130186599-130186599 on Build GRCh38
rs186361555 Chr.5: 130186947-130186947 on Build GRCh38
rs956852218 Chr.5: 130186880-130186880 on Build GRCh38
rs1003693268 Chr.5: 130186843-130186843 on Build GRCh38
rs944037874 Chr.5: 130186917-130186917 on Build GRCh38
rs1015035495 Chr.5: 130186907-130186907 on Build GRCh38
rs893802842 Chr.5: 130186815-130186815 on Build GRCh38
rs953362049 Chr.5: 130186695-130186695 on Build GRCh38
rs552822652 Chr.5: 130186598-130186598 on Build GRCh38
rs548225625 Chr.5: 130186677-130186677 on Build GRCh38
rs12515816 Chr.5: 130186804-130186804 on Build GRCh38
rs182077044 Chr.5: 130186830-130186830 on Build GRCh38
rs1039650420 Chr.5: 130186673-130186673 on Build GRCh38
rs895477600 Chr.5: 130186793-130186793 on Build GRCh38
rs751696375 Chr.5: 130186990-130186990 on Build GRCh38
rs188245717 Chr.5: 130186698-130186698 on Build GRCh38
rs1035322126 Chr.5: 130186776-130186776 on Build GRCh38
rs988634654 Chr.5: 130186892-130186892 on Build GRCh38
rs146262925 Chr.5: 130186639-130186639 on Build GRCh38
rs139434453 Chr.5: 130186818-130186818 on Build GRCh38
rs899644128 Chr.5: 130186688-130186688 on Build GRCh38
rs748109062 Chr.5: 130186935-130186937 on Build GRCh38

View Full Product Details