Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 131641931-131642479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FNIP1
Entrez Gene ID
96459
Gene Name
folliculin interacting protein 1
Gene Aliases
-
UniGene
Hs.591273
Gene Chromosome Location
Chr.5: 131641714-131797063 on Build GRCh38
Transcripts
NM_001008738.2 NM_133372.2

SNP Detail

SNP ID Chromosome Location
rs987686896 Chr.5: 131642301-131642301 on Build GRCh38
rs752507759 Chr.5: 131642411-131642411 on Build GRCh38
rs940206466 Chr.5: 131642356-131642356 on Build GRCh38
rs893646263 Chr.5: 131642103-131642103 on Build GRCh38
rs141219047 Chr.5: 131642092-131642092 on Build GRCh38
rs181510352 Chr.5: 131641962-131641962 on Build GRCh38
rs917326823 Chr.5: 131642380-131642380 on Build GRCh38
rs1035455580 Chr.5: 131642172-131642172 on Build GRCh38
rs752688466 Chr.5: 131642434-131642436 on Build GRCh38
rs1012537324 Chr.5: 131642234-131642234 on Build GRCh38
rs138049748 Chr.5: 131642256-131642256 on Build GRCh38
rs1039130386 Chr.5: 131642369-131642369 on Build GRCh38
rs74603102 Chr.5: 131642310-131642310 on Build GRCh38
rs924511872 Chr.5: 131642295-131642295 on Build GRCh38
rs943030577 Chr.5: 131641992-131642014 on Build GRCh38
rs568185234 Chr.5: 131642125-131642126 on Build GRCh38
rs1022551217 Chr.5: 131642239-131642239 on Build GRCh38
rs899424566 Chr.5: 131641989-131641989 on Build GRCh38
rs1014276921 Chr.5: 131642016-131642016 on Build GRCh38
rs529907747 Chr.5: 131642241-131642241 on Build GRCh38
rs879114236 Chr.5: 131642000-131642000 on Build GRCh38
rs145312650 Chr.5: 131642181-131642181 on Build GRCh38
rs955914328 Chr.5: 131642300-131642300 on Build GRCh38
rs1048004503 Chr.5: 131642447-131642447 on Build GRCh38
rs867481532 Chr.5: 131642377-131642377 on Build GRCh38
rs760805519 Chr.5: 131642233-131642233 on Build GRCh38
rs765151299 Chr.5: 131642398-131642398 on Build GRCh38
rs186749810 Chr.5: 131642147-131642147 on Build GRCh38
rs75501564 Chr.5: 131642327-131642327 on Build GRCh38
rs995120068 Chr.5: 131642082-131642082 on Build GRCh38
rs10052961 Chr.5: 131642439-131642439 on Build GRCh38
rs763497005 Chr.5: 131642448-131642448 on Build GRCh38
rs886632481 Chr.5: 131642134-131642134 on Build GRCh38
rs1032579896 Chr.5: 131642098-131642099 on Build GRCh38
rs895766450 Chr.5: 131642015-131642015 on Build GRCh38
rs1044443916 Chr.5: 131642079-131642079 on Build GRCh38
rs773402975 Chr.5: 131642211-131642211 on Build GRCh38
rs910007122 Chr.5: 131642344-131642344 on Build GRCh38

View Full Product Details