Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132073714-132074212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2
Entrez Gene ID
1437
Gene Name
colony stimulating factor 2
Gene Aliases
GMCSF
UniGene
Hs.1349
Gene Chromosome Location
Chr.5: 132073792-132076170 on Build GRCh38
Transcripts
NM_000758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060633 c.184G>T p.E62* Chr.5: 132074105-132074105 on Build GRCh38
5918615 c.148G>A p.A50T Chr.5: 132073971-132073971 on Build GRCh38
3409721 c.170T>C p.V57A Chr.5: 132074091-132074091 on Build GRCh38
3428895 c.99G>C p.V33V Chr.5: 132073922-132073922 on Build GRCh38
4403217 c.135G>A p.L45L Chr.5: 132073958-132073958 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757115989 Chr.5: 132073981-132073981 on Build GRCh38
rs755097799 Chr.5: 132073883-132073883 on Build GRCh38
rs766529017 Chr.5: 132073880-132073880 on Build GRCh38
rs749993132 Chr.5: 132074112-132074112 on Build GRCh38
rs750583291 Chr.5: 132073871-132073871 on Build GRCh38
rs182488161 Chr.5: 132074163-132074163 on Build GRCh38
rs200740065 Chr.5: 132073859-132073859 on Build GRCh38
rs747355813 Chr.5: 132074165-132074165 on Build GRCh38
rs372448237 Chr.5: 132073809-132073809 on Build GRCh38
rs761215509 Chr.5: 132073944-132073944 on Build GRCh38
rs776821581 Chr.5: 132074022-132074022 on Build GRCh38
rs761270332 Chr.5: 132074016-132074016 on Build GRCh38
rs752883786 Chr.5: 132074040-132074040 on Build GRCh38
rs779569419 Chr.5: 132074127-132074127 on Build GRCh38
rs201520008 Chr.5: 132073856-132073856 on Build GRCh38
rs2069621 Chr.5: 132073889-132073889 on Build GRCh38
rs762065062 Chr.5: 132073909-132073909 on Build GRCh38
rs763175811 Chr.5: 132074041-132074041 on Build GRCh38
rs755284838 Chr.5: 132073950-132073950 on Build GRCh38
rs764528813 Chr.5: 132073836-132073836 on Build GRCh38
rs907581684 Chr.5: 132074003-132074003 on Build GRCh38
rs778349474 Chr.5: 132074148-132074148 on Build GRCh38
rs939074730 Chr.5: 132074029-132074029 on Build GRCh38
rs745361354 Chr.5: 132073998-132073998 on Build GRCh38
rs763741354 Chr.5: 132073985-132073985 on Build GRCh38
rs530587752 Chr.5: 132073943-132073943 on Build GRCh38
rs772933488 Chr.5: 132073942-132073942 on Build GRCh38
rs746958091 Chr.5: 132073980-132073980 on Build GRCh38
rs759502463 Chr.5: 132074030-132074030 on Build GRCh38
rs550408367 Chr.5: 132074021-132074021 on Build GRCh38
rs771605705 Chr.5: 132074168-132074168 on Build GRCh38
rs777654384 Chr.5: 132073888-132073888 on Build GRCh38
rs748908623 Chr.5: 132074131-132074131 on Build GRCh38
rs936812640 Chr.5: 132074115-132074115 on Build GRCh38
rs368183845 Chr.5: 132073788-132073788 on Build GRCh38
rs369533011 Chr.5: 132073938-132073938 on Build GRCh38
rs201688947 Chr.5: 132073884-132073884 on Build GRCh38
rs1043956044 Chr.5: 132074169-132074169 on Build GRCh38
rs370362075 Chr.5: 132074010-132074010 on Build GRCh38
rs190462832 Chr.5: 132074032-132074032 on Build GRCh38
rs757238860 Chr.5: 132074090-132074090 on Build GRCh38
rs200145269 Chr.5: 132073945-132073945 on Build GRCh38
rs745535281 Chr.5: 132073902-132073902 on Build GRCh38
rs751348663 Chr.5: 132074087-132074087 on Build GRCh38
rs1052673266 Chr.5: 132073777-132073777 on Build GRCh38
rs761384799 Chr.5: 132073876-132073876 on Build GRCh38
rs779545301 Chr.5: 132074006-132074006 on Build GRCh38
rs752575776 Chr.5: 132073955-132073955 on Build GRCh38
rs373579996 Chr.5: 132074013-132074013 on Build GRCh38
rs755613206 Chr.5: 132074114-132074114 on Build GRCh38
rs145293289 Chr.5: 132073903-132073903 on Build GRCh38
rs774509300 Chr.5: 132073842-132073842 on Build GRCh38
rs899549095 Chr.5: 132073818-132073818 on Build GRCh38
rs759137100 Chr.5: 132073827-132073827 on Build GRCh38
rs770447592 Chr.5: 132073892-132073892 on Build GRCh38
rs751428188 Chr.5: 132074048-132074048 on Build GRCh38
rs781090023 Chr.5: 132074093-132074093 on Build GRCh38
rs200208265 Chr.5: 132073881-132073881 on Build GRCh38
rs925385229 Chr.5: 132074074-132074074 on Build GRCh38
rs758409716 Chr.5: 132073885-132073885 on Build GRCh38
rs955875687 Chr.5: 132073901-132073901 on Build GRCh38
rs185543647 Chr.5: 132073795-132073795 on Build GRCh38
rs10072253 Chr.5: 132074075-132074075 on Build GRCh38
rs768357906 Chr.5: 132074144-132074144 on Build GRCh38
rs2069622 Chr.5: 132073904-132073904 on Build GRCh38
rs753902523 Chr.5: 132073947-132073947 on Build GRCh38
rs375972077 Chr.5: 132073999-132073999 on Build GRCh38
rs776058350 Chr.5: 132073894-132073894 on Build GRCh38
rs780742719 Chr.5: 132073993-132073993 on Build GRCh38
rs758157313 Chr.5: 132073974-132073974 on Build GRCh38
rs765354692 Chr.5: 132073953-132073953 on Build GRCh38
rs768111482 Chr.5: 132073920-132073920 on Build GRCh38
rs749239852 Chr.5: 132073783-132073783 on Build GRCh38
rs368853369 Chr.5: 132074053-132074053 on Build GRCh38
rs756382017 Chr.5: 132074014-132074014 on Build GRCh38
rs992138223 Chr.5: 132073927-132073927 on Build GRCh38
rs143375057 Chr.5: 132073979-132073979 on Build GRCh38

View Full Product Details