Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132073991-132074491 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSF2
Entrez Gene ID
1437
Gene Name
colony stimulating factor 2
Gene Aliases
GMCSF
UniGene
Hs.1349
Gene Chromosome Location
Chr.5: 132073792-132076170 on Build GRCh38
Transcripts
NM_000758.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1060633 c.184G>T p.E62* Chr.5: 132074105-132074105 on Build GRCh38
3409721 c.170T>C p.V57A Chr.5: 132074091-132074091 on Build GRCh38
6415190 c.196C>T p.L66F Chr.5: 132074117-132074117 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs951808186 Chr.5: 132074312-132074312 on Build GRCh38
rs749993132 Chr.5: 132074112-132074112 on Build GRCh38
rs944297282 Chr.5: 132074215-132074215 on Build GRCh38
rs115044507 Chr.5: 132074434-132074434 on Build GRCh38
rs182488161 Chr.5: 132074163-132074163 on Build GRCh38
rs747355813 Chr.5: 132074165-132074165 on Build GRCh38
rs1031512130 Chr.5: 132074258-132074258 on Build GRCh38
rs907534662 Chr.5: 132074403-132074403 on Build GRCh38
rs776821581 Chr.5: 132074022-132074022 on Build GRCh38
rs761270332 Chr.5: 132074016-132074016 on Build GRCh38
rs752883786 Chr.5: 132074040-132074040 on Build GRCh38
rs779569419 Chr.5: 132074127-132074127 on Build GRCh38
rs763175811 Chr.5: 132074041-132074041 on Build GRCh38
rs991894557 Chr.5: 132074424-132074424 on Build GRCh38
rs939107232 Chr.5: 132074423-132074423 on Build GRCh38
rs1004000822 Chr.5: 132074303-132074303 on Build GRCh38
rs778349474 Chr.5: 132074148-132074148 on Build GRCh38
rs939074730 Chr.5: 132074029-132074029 on Build GRCh38
rs958109899 Chr.5: 132074441-132074441 on Build GRCh38
rs2074561 Chr.5: 132074254-132074254 on Build GRCh38
rs1044373816 Chr.5: 132074436-132074436 on Build GRCh38
rs1008265525 Chr.5: 132074276-132074276 on Build GRCh38
rs2069623 Chr.5: 132074420-132074420 on Build GRCh38
rs1022903814 Chr.5: 132074391-132074391 on Build GRCh38
rs759502463 Chr.5: 132074030-132074030 on Build GRCh38
rs550408367 Chr.5: 132074021-132074021 on Build GRCh38
rs932343718 Chr.5: 132074246-132074246 on Build GRCh38
rs771605705 Chr.5: 132074168-132074168 on Build GRCh38
rs936812640 Chr.5: 132074115-132074115 on Build GRCh38
rs748908623 Chr.5: 132074131-132074131 on Build GRCh38
rs765578927 Chr.5: 132074285-132074285 on Build GRCh38
rs1043956044 Chr.5: 132074169-132074169 on Build GRCh38
rs904071993 Chr.5: 132074439-132074439 on Build GRCh38
rs190462832 Chr.5: 132074032-132074032 on Build GRCh38
rs757238860 Chr.5: 132074090-132074090 on Build GRCh38
rs1040687329 Chr.5: 132074224-132074224 on Build GRCh38
rs751348663 Chr.5: 132074087-132074087 on Build GRCh38
rs35576728 Chr.5: 132074419-132074420 on Build GRCh38
rs551318109 Chr.5: 132074386-132074386 on Build GRCh38
rs373579996 Chr.5: 132074013-132074013 on Build GRCh38
rs755613206 Chr.5: 132074114-132074114 on Build GRCh38
rs25880 Chr.5: 132074278-132074278 on Build GRCh38
rs975081505 Chr.5: 132074287-132074287 on Build GRCh38
rs185902755 Chr.5: 132074241-132074241 on Build GRCh38
rs751428188 Chr.5: 132074048-132074048 on Build GRCh38
rs781090023 Chr.5: 132074093-132074093 on Build GRCh38
rs925385229 Chr.5: 132074074-132074074 on Build GRCh38
rs535022168 Chr.5: 132074327-132074327 on Build GRCh38
rs10072253 Chr.5: 132074075-132074075 on Build GRCh38
rs1027531128 Chr.5: 132074299-132074299 on Build GRCh38
rs768357906 Chr.5: 132074144-132074144 on Build GRCh38
rs888230705 Chr.5: 132074293-132074293 on Build GRCh38
rs999494673 Chr.5: 132074252-132074252 on Build GRCh38
rs895704112 Chr.5: 132074326-132074326 on Build GRCh38
rs566014633 Chr.5: 132074284-132074284 on Build GRCh38
rs2069624 Chr.5: 132074425-132074425 on Build GRCh38
rs1014604599 Chr.5: 132074352-132074352 on Build GRCh38
rs368853369 Chr.5: 132074053-132074053 on Build GRCh38
rs756382017 Chr.5: 132074014-132074014 on Build GRCh38
rs1048120891 Chr.5: 132074263-132074263 on Build GRCh38
rs891245956 Chr.5: 132074274-132074274 on Build GRCh38
rs557188416 Chr.5: 132074459-132074459 on Build GRCh38

View Full Product Details