Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132751461-132751983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNI2
Entrez Gene ID
645121
Gene Name
cyclin I family member 2
Gene Aliases
-
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132747402-132754403 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132750817-132778216 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

SNP Detail

SNP ID Chromosome Location
rs111234856 Chr.5: 132751724-132751724 on Build GRCh38
rs1032716070 Chr.5: 132751907-132751907 on Build GRCh38
rs544670339 Chr.5: 132751601-132751601 on Build GRCh38
rs908430097 Chr.5: 132751827-132751827 on Build GRCh38
rs746123751 Chr.5: 132751581-132751581 on Build GRCh38
rs542443175 Chr.5: 132751602-132751602 on Build GRCh38
rs1056857214 Chr.5: 132751882-132751882 on Build GRCh38
rs932642134 Chr.5: 132751723-132751723 on Build GRCh38
rs886927521 Chr.5: 132751504-132751504 on Build GRCh38
rs924753467 Chr.5: 132751777-132751777 on Build GRCh38
rs988639308 Chr.5: 132751626-132751626 on Build GRCh38
rs1012210145 Chr.5: 132751605-132751605 on Build GRCh38
rs770162115 Chr.5: 132751940-132751940 on Build GRCh38
rs938525338 Chr.5: 132751873-132751873 on Build GRCh38
rs747439972 Chr.5: 132751911-132751911 on Build GRCh38
rs749307408 Chr.5: 132751955-132751955 on Build GRCh38
rs536584015 Chr.5: 132751533-132751533 on Build GRCh38
rs768926275 Chr.5: 132751961-132751961 on Build GRCh38
rs753255741 Chr.5: 132751896-132751896 on Build GRCh38
rs1020545846 Chr.5: 132751647-132751647 on Build GRCh38
rs115141047 Chr.5: 132751902-132751902 on Build GRCh38
rs942396008 Chr.5: 132751729-132751729 on Build GRCh38
rs778692925 Chr.5: 132751904-132751904 on Build GRCh38
rs572843358 Chr.5: 132751577-132751577 on Build GRCh38
rs575416996 Chr.5: 132751623-132751623 on Build GRCh38
rs1035083050 Chr.5: 132751588-132751588 on Build GRCh38
rs957075457 Chr.5: 132751696-132751696 on Build GRCh38
rs879047407 Chr.5: 132751753-132751753 on Build GRCh38
rs559784246 Chr.5: 132751815-132751815 on Build GRCh38
rs182007156 Chr.5: 132751685-132751685 on Build GRCh38
rs1046933155 Chr.5: 132751497-132751500 on Build GRCh38
rs780182209 Chr.5: 132751732-132751732 on Build GRCh38
rs560757045 Chr.5: 132751930-132751930 on Build GRCh38
rs897327249 Chr.5: 132751806-132751806 on Build GRCh38
rs986943973 Chr.5: 132751704-132751704 on Build GRCh38
rs1045077329 Chr.5: 132751929-132751929 on Build GRCh38
rs757697071 Chr.5: 132751928-132751928 on Build GRCh38
rs968367419 Chr.5: 132751673-132751673 on Build GRCh38
rs766876414 Chr.5: 132751554-132751554 on Build GRCh38
rs892850723 Chr.5: 132751908-132751908 on Build GRCh38
rs941452943 Chr.5: 132751758-132751758 on Build GRCh38
rs753990750 Chr.5: 132751554-132751555 on Build GRCh38
rs1014928044 Chr.5: 132751486-132751486 on Build GRCh38
rs41298944 Chr.5: 132751560-132751560 on Build GRCh38
rs1023944234 Chr.5: 132751531-132751531 on Build GRCh38
rs1046385753 Chr.5: 132751851-132751851 on Build GRCh38
rs529485047 Chr.5: 132751773-132751773 on Build GRCh38
rs549695318 Chr.5: 132751801-132751801 on Build GRCh38
rs921898598 Chr.5: 132751719-132751719 on Build GRCh38
rs556759810 Chr.5: 132751500-132751500 on Build GRCh38
rs746412378 Chr.5: 132751933-132751933 on Build GRCh38
rs963800811 Chr.5: 132751768-132751768 on Build GRCh38
rs756086309 Chr.5: 132751675-132751675 on Build GRCh38
rs563890635 Chr.5: 132751738-132751738 on Build GRCh38
rs1001225027 Chr.5: 132751583-132751583 on Build GRCh38
rs957141297 Chr.5: 132751603-132751603 on Build GRCh38
rs775682601 Chr.5: 132751946-132751946 on Build GRCh38
rs573466381 Chr.5: 132751517-132751517 on Build GRCh38

View Full Product Details