Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132751823-132752342 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNI2
Entrez Gene ID
645121
Gene Name
cyclin I family member 2
Gene Aliases
-
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132747402-132754403 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

Gene Symbol
SEPT8
Entrez Gene ID
23176
Gene Name
septin 8
Gene Aliases
SEP2
UniGene
Hs.522057
Gene Chromosome Location
Chr.5: 132750817-132778216 on Build GRCh38
Transcripts
NM_001039780.3 NM_001287252.1 NM_001287253.1 XM_017009725.1 NM_001098811.1 XM_005271927.2 XM_005271928.2 XM_005271931.3 XM_011543288.1 XM_017009254.1 XM_017009255.1 XM_017009256.1 XM_017009257.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3826731 c.890A>C p.H297P Chr.5: 132752081-132752081 on Build GRCh38
4653888 c.991C>T p.L331L Chr.5: 132752182-132752182 on Build GRCh38
5517965 c.963C>T p.P321P Chr.5: 132752154-132752154 on Build GRCh38
1200165 c.847G>A p.V283I Chr.5: 132752038-132752038 on Build GRCh38
7423851 c.871C>A p.Q291K Chr.5: 132752062-132752062 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764769439 Chr.5: 132752008-132752008 on Build GRCh38
rs760371108 Chr.5: 132752075-132752075 on Build GRCh38
rs1032716070 Chr.5: 132751907-132751907 on Build GRCh38
rs760705615 Chr.5: 132751996-132751996 on Build GRCh38
rs763634938 Chr.5: 132752184-132752184 on Build GRCh38
rs376378013 Chr.5: 132752041-132752041 on Build GRCh38
rs973245890 Chr.5: 132752100-132752100 on Build GRCh38
rs748399608 Chr.5: 132752048-132752048 on Build GRCh38
rs1056857214 Chr.5: 132751882-132751882 on Build GRCh38
rs1045930570 Chr.5: 132752164-132752164 on Build GRCh38
rs758079357 Chr.5: 132752025-132752025 on Build GRCh38
rs759108150 Chr.5: 132752007-132752007 on Build GRCh38
rs781716088 Chr.5: 132752028-132752028 on Build GRCh38
rs770162115 Chr.5: 132751940-132751940 on Build GRCh38
rs918702721 Chr.5: 132752141-132752141 on Build GRCh38
rs938525338 Chr.5: 132751873-132751873 on Build GRCh38
rs915735995 Chr.5: 132752282-132752282 on Build GRCh38
rs747439972 Chr.5: 132751911-132751911 on Build GRCh38
rs749307408 Chr.5: 132751955-132751955 on Build GRCh38
rs768926275 Chr.5: 132751961-132751961 on Build GRCh38
rs919625405 Chr.5: 132752101-132752101 on Build GRCh38
rs143338609 Chr.5: 132752154-132752154 on Build GRCh38
rs199927703 Chr.5: 132751999-132751999 on Build GRCh38
rs753255741 Chr.5: 132751896-132751896 on Build GRCh38
rs115141047 Chr.5: 132751902-132751902 on Build GRCh38
rs778692925 Chr.5: 132751904-132751904 on Build GRCh38
rs776433041 Chr.5: 132752111-132752111 on Build GRCh38
rs759485146 Chr.5: 132752124-132752124 on Build GRCh38
rs553274041 Chr.5: 132752136-132752136 on Build GRCh38
rs115692489 Chr.5: 132751993-132751993 on Build GRCh38
rs34204192 Chr.5: 132752270-132752270 on Build GRCh38
rs1056933613 Chr.5: 132752263-132752263 on Build GRCh38
rs759907297 Chr.5: 132752036-132752037 on Build GRCh38
rs369479594 Chr.5: 132752224-132752224 on Build GRCh38
rs560757045 Chr.5: 132751930-132751930 on Build GRCh38
rs536055418 Chr.5: 132752070-132752070 on Build GRCh38
rs774473174 Chr.5: 132752052-132752052 on Build GRCh38
rs757697071 Chr.5: 132751928-132751928 on Build GRCh38
rs368331115 Chr.5: 132752137-132752137 on Build GRCh38
rs768698720 Chr.5: 132751977-132751977 on Build GRCh38
rs1045077329 Chr.5: 132751929-132751929 on Build GRCh38
rs756694987 Chr.5: 132752037-132752037 on Build GRCh38
rs908460573 Chr.5: 132752254-132752254 on Build GRCh38
rs982438073 Chr.5: 132752252-132752252 on Build GRCh38
rs201961929 Chr.5: 132752040-132752040 on Build GRCh38
rs892850723 Chr.5: 132751908-132751908 on Build GRCh38
rs750725957 Chr.5: 132752030-132752030 on Build GRCh38
rs756433145 Chr.5: 132752220-132752220 on Build GRCh38
rs750858553 Chr.5: 132752201-132752201 on Build GRCh38
rs780414471 Chr.5: 132752038-132752038 on Build GRCh38
rs371030907 Chr.5: 132752076-132752076 on Build GRCh38
rs375318962 Chr.5: 132752127-132752127 on Build GRCh38
rs200114738 Chr.5: 132751970-132751970 on Build GRCh38
rs1046385753 Chr.5: 132751851-132751851 on Build GRCh38
rs767320935 Chr.5: 132751990-132751990 on Build GRCh38
rs538880792 Chr.5: 132752158-132752158 on Build GRCh38
rs78904826 Chr.5: 132751992-132751992 on Build GRCh38
rs73787056 Chr.5: 132752256-132752256 on Build GRCh38
rs952500171 Chr.5: 132752134-132752134 on Build GRCh38
rs746412378 Chr.5: 132751933-132751933 on Build GRCh38
rs760380394 Chr.5: 132752297-132752297 on Build GRCh38
rs761957678 Chr.5: 132751973-132751973 on Build GRCh38
rs530189479 Chr.5: 132752296-132752296 on Build GRCh38
rs781436328 Chr.5: 132752282-132752283 on Build GRCh38
rs779417217 Chr.5: 132752132-132752133 on Build GRCh38
rs937577790 Chr.5: 132752285-132752285 on Build GRCh38
rs550644991 Chr.5: 132752019-132752019 on Build GRCh38
rs775682601 Chr.5: 132751946-132751946 on Build GRCh38
rs1045111576 Chr.5: 132752319-132752322 on Build GRCh38
rs754315732 Chr.5: 132752229-132752229 on Build GRCh38
rs115152193 Chr.5: 132752005-132752005 on Build GRCh38

View Full Product Details