Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 133198697-133199197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FSTL4
Entrez Gene ID
23105
Gene Name
follistatin like 4
Gene Aliases
-
UniGene
Hs.483390
Gene Chromosome Location
Chr.5: 133194827-133842066 on Build GRCh38
Transcripts
NM_015082.1 XM_011543283.1 XM_011543284.1 XM_011543286.2 XM_011543287.2 XM_017009251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6667567 c.2465C>T p.T822M Chr.5: 133199159-133199159 on Build GRCh38
7422237 c.2449A>T p.N817Y Chr.5: 133199175-133199175 on Build GRCh38
7422236 c.2500A>G p.T834A Chr.5: 133199124-133199124 on Build GRCh38
5101761 c.2499_2500insG p.T834fs*8 Chr.5: 133199124-133199125 on Build GRCh38
6667568 c.2470C>T p.R824W Chr.5: 133199154-133199154 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1038716646 Chr.5: 133198815-133198815 on Build GRCh38
rs1038829351 Chr.5: 133199101-133199101 on Build GRCh38
rs745485239 Chr.5: 133199143-133199143 on Build GRCh38
rs1043007396 Chr.5: 133198805-133198805 on Build GRCh38
rs1017422539 Chr.5: 133199152-133199152 on Build GRCh38
rs544273813 Chr.5: 133199161-133199161 on Build GRCh38
rs1010038465 Chr.5: 133198839-133198860 on Build GRCh38
rs144854772 Chr.5: 133198875-133198875 on Build GRCh38
rs528695765 Chr.5: 133198769-133198769 on Build GRCh38
rs769509086 Chr.5: 133199153-133199153 on Build GRCh38
rs879344879 Chr.5: 133199011-133199011 on Build GRCh38
rs994741365 Chr.5: 133198828-133198828 on Build GRCh38
rs991575901 Chr.5: 133198898-133198898 on Build GRCh38
rs1035275108 Chr.5: 133198888-133198888 on Build GRCh38
rs746807141 Chr.5: 133199078-133199078 on Build GRCh38
rs781004091 Chr.5: 133199138-133199138 on Build GRCh38
rs777106231 Chr.5: 133199172-133199172 on Build GRCh38
rs960114455 Chr.5: 133198897-133198897 on Build GRCh38
rs750751038 Chr.5: 133199109-133199109 on Build GRCh38
rs916176126 Chr.5: 133199019-133199019 on Build GRCh38
rs73273844 Chr.5: 133198750-133198750 on Build GRCh38
rs150466282 Chr.5: 133199154-133199154 on Build GRCh38
rs755270499 Chr.5: 133199129-133199129 on Build GRCh38
rs775114004 Chr.5: 133199085-133199085 on Build GRCh38
rs769777780 Chr.5: 133198822-133198822 on Build GRCh38
rs776249576 Chr.5: 133199080-133199080 on Build GRCh38
rs761135653 Chr.5: 133199123-133199123 on Build GRCh38
rs762177525 Chr.5: 133199089-133199089 on Build GRCh38
rs968719455 Chr.5: 133198909-133198909 on Build GRCh38
rs568484339 Chr.5: 133199071-133199071 on Build GRCh38
rs915917565 Chr.5: 133198747-133198747 on Build GRCh38
rs182483019 Chr.5: 133198917-133198917 on Build GRCh38
rs376932662 Chr.5: 133198929-133198929 on Build GRCh38
rs772650848 Chr.5: 133199156-133199156 on Build GRCh38
rs762405379 Chr.5: 133199124-133199125 on Build GRCh38
rs769154224 Chr.5: 133199082-133199082 on Build GRCh38
rs116786563 Chr.5: 133198799-133198799 on Build GRCh38
rs938985690 Chr.5: 133198738-133198738 on Build GRCh38
rs375184077 Chr.5: 133199159-133199159 on Build GRCh38
rs777498985 Chr.5: 133199077-133199077 on Build GRCh38
rs199609726 Chr.5: 133199126-133199126 on Build GRCh38
rs780428529 Chr.5: 133199055-133199055 on Build GRCh38
rs139750390 Chr.5: 133198986-133198986 on Build GRCh38
rs545038336 Chr.5: 133199079-133199079 on Build GRCh38
rs140495211 Chr.5: 133199125-133199125 on Build GRCh38
rs934915081 Chr.5: 133198959-133198959 on Build GRCh38
rs62374514 Chr.5: 133198922-133198922 on Build GRCh38
rs1031542262 Chr.5: 133198857-133198857 on Build GRCh38
rs1003875282 Chr.5: 133199175-133199175 on Build GRCh38
rs934870171 Chr.5: 133198803-133198803 on Build GRCh38
rs147903709 Chr.5: 133199116-133199116 on Build GRCh38
rs770705305 Chr.5: 133199136-133199136 on Build GRCh38
rs537516590 Chr.5: 133198889-133198889 on Build GRCh38
rs373390539 Chr.5: 133199092-133199092 on Build GRCh38
rs747004645 Chr.5: 133199131-133199131 on Build GRCh38
rs772099992 Chr.5: 133199075-133199075 on Build GRCh38
rs890177227 Chr.5: 133198809-133198809 on Build GRCh38
rs558686290 Chr.5: 133199130-133199130 on Build GRCh38
rs766822429 Chr.5: 133199166-133199166 on Build GRCh38
rs894698075 Chr.5: 133198793-133198793 on Build GRCh38
rs943102017 Chr.5: 133198814-133198814 on Build GRCh38
rs908744474 Chr.5: 133198934-133198934 on Build GRCh38
rs987617559 Chr.5: 133198996-133198996 on Build GRCh38
rs1026257163 Chr.5: 133198865-133198865 on Build GRCh38
rs979018250 Chr.5: 133198756-133198756 on Build GRCh38
rs759297651 Chr.5: 133199081-133199081 on Build GRCh38
rs779117842 Chr.5: 133199069-133199069 on Build GRCh38
rs983604809 Chr.5: 133198931-133198931 on Build GRCh38
rs773843699 Chr.5: 133199158-133199158 on Build GRCh38
rs904530045 Chr.5: 133199105-133199105 on Build GRCh38
rs756364860 Chr.5: 133199047-133199047 on Build GRCh38
rs1003351356 Chr.5: 133198877-133198877 on Build GRCh38
rs567297786 Chr.5: 133198919-133198919 on Build GRCh38
rs776653715 Chr.5: 133198727-133198727 on Build GRCh38
rs948958717 Chr.5: 133199025-133199025 on Build GRCh38
rs1052375844 Chr.5: 133198808-133198808 on Build GRCh38
rs753912649 Chr.5: 133199058-133199058 on Build GRCh38
rs572846957 Chr.5: 133198987-133198987 on Build GRCh38
rs528915419 Chr.5: 133198908-133198908 on Build GRCh38

View Full Product Details