Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134751512-134752017 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAMLG
Entrez Gene ID
819
Gene Name
calcium modulating ligand
Gene Aliases
CAML, GET2
UniGene
Hs.529846
Gene Chromosome Location
Chr.5: 134737927-134752160 on Build GRCh38
Transcripts
NM_001745.3

SNP Detail

SNP ID Chromosome Location
rs889331834 Chr.5: 134751873-134751873 on Build GRCh38
rs943620378 Chr.5: 134751902-134751902 on Build GRCh38
rs981128393 Chr.5: 134751749-134751749 on Build GRCh38
rs879192451 Chr.5: 134751601-134751601 on Build GRCh38
rs779512577 Chr.5: 134751713-134751713 on Build GRCh38
rs933317773 Chr.5: 134751799-134751799 on Build GRCh38
rs985872550 Chr.5: 134751829-134751829 on Build GRCh38
rs1025285910 Chr.5: 134751722-134751722 on Build GRCh38
rs993710165 Chr.5: 134751938-134751938 on Build GRCh38
rs568939841 Chr.5: 134751911-134751911 on Build GRCh38
rs147532001 Chr.5: 134751643-134751643 on Build GRCh38
rs539061684 Chr.5: 134751915-134751915 on Build GRCh38
rs567545376 Chr.5: 134751736-134751736 on Build GRCh38
rs976247805 Chr.5: 134751882-134751882 on Build GRCh38
rs958909377 Chr.5: 134751752-134751752 on Build GRCh38
rs1025381593 Chr.5: 134751957-134751957 on Build GRCh38
rs914394030 Chr.5: 134751758-134751758 on Build GRCh38
rs1024210620 Chr.5: 134751767-134751767 on Build GRCh38
rs1032644530 Chr.5: 134751785-134751785 on Build GRCh38
rs898204411 Chr.5: 134751709-134751709 on Build GRCh38
rs995682615 Chr.5: 134751711-134751711 on Build GRCh38
rs560619649 Chr.5: 134751683-134751683 on Build GRCh38
rs924720762 Chr.5: 134751790-134751790 on Build GRCh38
rs192678182 Chr.5: 134751918-134751918 on Build GRCh38
rs1035605022 Chr.5: 134751750-134751750 on Build GRCh38
rs1002532019 Chr.5: 134751974-134751974 on Build GRCh38
rs775422003 Chr.5: 134751909-134751910 on Build GRCh38
rs781195223 Chr.5: 134751777-134751777 on Build GRCh38
rs1049018509 Chr.5: 134751682-134751684 on Build GRCh38
rs768563841 Chr.5: 134751776-134751776 on Build GRCh38
rs944956424 Chr.5: 134751580-134751580 on Build GRCh38
rs566758882 Chr.5: 134751961-134751961 on Build GRCh38
rs1016851875 Chr.5: 134751671-134751671 on Build GRCh38
rs116166805 Chr.5: 134751981-134751981 on Build GRCh38
rs957359393 Chr.5: 134751827-134751827 on Build GRCh38
rs1051944685 Chr.5: 134751645-134751645 on Build GRCh38
rs549148844 Chr.5: 134751714-134751714 on Build GRCh38
rs528017616 Chr.5: 134751703-134751703 on Build GRCh38
rs113232868 Chr.5: 134751742-134751742 on Build GRCh38
rs886897156 Chr.5: 134751962-134751962 on Build GRCh38
rs1041682168 Chr.5: 134751583-134751583 on Build GRCh38
rs910690428 Chr.5: 134751549-134751549 on Build GRCh38
rs998478645 Chr.5: 134751783-134751783 on Build GRCh38
rs572250810 Chr.5: 134751707-134751707 on Build GRCh38
rs1014030533 Chr.5: 134751754-134751754 on Build GRCh38
rs1051746084 Chr.5: 134751834-134751834 on Build GRCh38
rs545388523 Chr.5: 134751657-134751657 on Build GRCh38
rs540944152 Chr.5: 134751847-134751847 on Build GRCh38
rs1007718044 Chr.5: 134751658-134751658 on Build GRCh38
rs903204039 Chr.5: 134751615-134751615 on Build GRCh38
rs919494876 Chr.5: 134751656-134751656 on Build GRCh38
rs550367395 Chr.5: 134751757-134751757 on Build GRCh38

View Full Product Details