Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135578320-135578816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL14
Entrez Gene ID
9547
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14
UniGene
Hs.483444
Gene Chromosome Location
Chr.5: 135570679-135579279 on Build GRCh38
Transcripts
NM_004887.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4675703 c.151G>A p.V51M Chr.5: 135578489-135578489 on Build GRCh38
4896318 c.128G>A p.G43E Chr.5: 135578512-135578512 on Build GRCh38
1619481 c.122G>T p.R41L Chr.5: 135578518-135578518 on Build GRCh38
4533513 c.198G>A p.K66K Chr.5: 135578442-135578442 on Build GRCh38
1249493 c.139C>T p.R47C Chr.5: 135578501-135578501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1008921067 Chr.5: 135578798-135578798 on Build GRCh38
rs560517036 Chr.5: 135578417-135578417 on Build GRCh38
rs945641563 Chr.5: 135578538-135578538 on Build GRCh38
rs376972531 Chr.5: 135578609-135578609 on Build GRCh38
rs771114824 Chr.5: 135578695-135578695 on Build GRCh38
rs766123170 Chr.5: 135578460-135578460 on Build GRCh38
rs187056781 Chr.5: 135578541-135578541 on Build GRCh38
rs890523920 Chr.5: 135578785-135578785 on Build GRCh38
rs992705085 Chr.5: 135578493-135578493 on Build GRCh38
rs753501987 Chr.5: 135578470-135578470 on Build GRCh38
rs952535462 Chr.5: 135578620-135578620 on Build GRCh38
rs773169539 Chr.5: 135578554-135578554 on Build GRCh38
rs777139682 Chr.5: 135578402-135578402 on Build GRCh38
rs866912043 Chr.5: 135578386-135578386 on Build GRCh38
rs1043327402 Chr.5: 135578739-135578739 on Build GRCh38
rs771365067 Chr.5: 135578399-135578399 on Build GRCh38
rs754785132 Chr.5: 135578574-135578574 on Build GRCh38
rs376803763 Chr.5: 135578681-135578681 on Build GRCh38
rs11553972 Chr.5: 135578728-135578728 on Build GRCh38
rs868752918 Chr.5: 135578749-135578749 on Build GRCh38
rs770174401 Chr.5: 135578517-135578517 on Build GRCh38
rs375752265 Chr.5: 135578698-135578698 on Build GRCh38
rs916141231 Chr.5: 135578349-135578349 on Build GRCh38
rs759898082 Chr.5: 135578404-135578404 on Build GRCh38
rs372053502 Chr.5: 135578553-135578553 on Build GRCh38
rs745590166 Chr.5: 135578442-135578442 on Build GRCh38
rs980887903 Chr.5: 135578401-135578401 on Build GRCh38
rs983027417 Chr.5: 135578623-135578623 on Build GRCh38
rs778702230 Chr.5: 135578692-135578692 on Build GRCh38
rs748420774 Chr.5: 135578715-135578715 on Build GRCh38
rs1008873324 Chr.5: 135578518-135578518 on Build GRCh38
rs11950696 Chr.5: 135578543-135578543 on Build GRCh38
rs775282762 Chr.5: 135578587-135578587 on Build GRCh38
rs538380085 Chr.5: 135578779-135578779 on Build GRCh38
rs992234606 Chr.5: 135578796-135578796 on Build GRCh38
rs896970399 Chr.5: 135578615-135578615 on Build GRCh38
rs904682667 Chr.5: 135578377-135578377 on Build GRCh38
rs780617344 Chr.5: 135578683-135578683 on Build GRCh38
rs749556019 Chr.5: 135578684-135578684 on Build GRCh38
rs376509196 Chr.5: 135578419-135578419 on Build GRCh38
rs373816158 Chr.5: 135578578-135578578 on Build GRCh38
rs531217502 Chr.5: 135578659-135578659 on Build GRCh38
rs755897425 Chr.5: 135578447-135578447 on Build GRCh38
rs914174075 Chr.5: 135578527-135578527 on Build GRCh38
rs372475912 Chr.5: 135578494-135578494 on Build GRCh38
rs772253716 Chr.5: 135578780-135578780 on Build GRCh38
rs201072568 Chr.5: 135578547-135578547 on Build GRCh38
rs764823853 Chr.5: 135578577-135578577 on Build GRCh38
rs753640621 Chr.5: 135578573-135578573 on Build GRCh38
rs751757658 Chr.5: 135578423-135578423 on Build GRCh38
rs567105771 Chr.5: 135578624-135578624 on Build GRCh38
rs1053711974 Chr.5: 135578784-135578784 on Build GRCh38
rs757876485 Chr.5: 135578584-135578584 on Build GRCh38
rs563142483 Chr.5: 135578689-135578689 on Build GRCh38
rs746373310 Chr.5: 135578591-135578591 on Build GRCh38
rs1008075171 Chr.5: 135578593-135578593 on Build GRCh38
rs948930491 Chr.5: 135578371-135578371 on Build GRCh38
rs371398248 Chr.5: 135578570-135578570 on Build GRCh38
rs774215455 Chr.5: 135578420-135578420 on Build GRCh38
rs367776635 Chr.5: 135578585-135578585 on Build GRCh38
rs952014914 Chr.5: 135578385-135578385 on Build GRCh38
rs760575317 Chr.5: 135578565-135578565 on Build GRCh38
rs368188288 Chr.5: 135578551-135578551 on Build GRCh38
rs781637591 Chr.5: 135578492-135578492 on Build GRCh38
rs747602303 Chr.5: 135578487-135578487 on Build GRCh38
rs760732075 Chr.5: 135578678-135578682 on Build GRCh38
rs773327721 Chr.5: 135578561-135578561 on Build GRCh38
rs757927757 Chr.5: 135578491-135578491 on Build GRCh38
rs761987854 Chr.5: 135578425-135578425 on Build GRCh38
rs2072347 Chr.5: 135578660-135578660 on Build GRCh38
rs980731970 Chr.5: 135578765-135578765 on Build GRCh38
rs749522745 Chr.5: 135578544-135578544 on Build GRCh38
rs926200666 Chr.5: 135578777-135578777 on Build GRCh38
rs34598055 Chr.5: 135578624-135578625 on Build GRCh38
rs11553971 Chr.5: 135578509-135578509 on Build GRCh38
rs1043359088 Chr.5: 135578372-135578372 on Build GRCh38
rs1015819896 Chr.5: 135578607-135578607 on Build GRCh38
rs770090101 Chr.5: 135578408-135578408 on Build GRCh38
rs371946511 Chr.5: 135578701-135578701 on Build GRCh38
rs182682697 Chr.5: 135578478-135578478 on Build GRCh38

View Full Product Details