Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135578627-135579120 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCL14
Entrez Gene ID
9547
Gene Name
C-X-C motif chemokine ligand 14
Gene Aliases
BMAC, BRAK, KEC, KS1, MIP-2g, MIP2G, NJAC, SCYB14
UniGene
Hs.483444
Gene Chromosome Location
Chr.5: 135570679-135579279 on Build GRCh38
Transcripts
NM_004887.4 XM_017009090.1 XM_017009092.1

Gene Symbol
SLC25A48
Entrez Gene ID
153328
Gene Name
solute carrier family 25 member 48
Gene Aliases
-
UniGene
Hs.412418
Gene Chromosome Location
Chr.5: 135579002-135888637 on Build GRCh38
Transcripts
NM_004887.4 XM_017009090.1 XM_017009092.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6397343 c.17G>A p.R6H Chr.5: 135578798-135578798 on Build GRCh38
5817765 c.15A>G p.P5P Chr.5: 135578800-135578800 on Build GRCh38
4930377 c.91C>T p.R31C Chr.5: 135578724-135578724 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1008921067 Chr.5: 135578798-135578798 on Build GRCh38
rs930613864 Chr.5: 135578953-135578953 on Build GRCh38
rs1039053759 Chr.5: 135578932-135578932 on Build GRCh38
rs745758248 Chr.5: 135578847-135578847 on Build GRCh38
rs771114824 Chr.5: 135578695-135578695 on Build GRCh38
rs944914214 Chr.5: 135578926-135578926 on Build GRCh38
rs890523920 Chr.5: 135578785-135578785 on Build GRCh38
rs760654374 Chr.5: 135578815-135578815 on Build GRCh38
rs895651001 Chr.5: 135579024-135579024 on Build GRCh38
rs1043327402 Chr.5: 135578739-135578739 on Build GRCh38
rs1049356113 Chr.5: 135578871-135578871 on Build GRCh38
rs967281920 Chr.5: 135579003-135579003 on Build GRCh38
rs376803763 Chr.5: 135578681-135578681 on Build GRCh38
rs934928269 Chr.5: 135578921-135578921 on Build GRCh38
rs11553972 Chr.5: 135578728-135578728 on Build GRCh38
rs868752918 Chr.5: 135578749-135578749 on Build GRCh38
rs375752265 Chr.5: 135578698-135578698 on Build GRCh38
rs556656061 Chr.5: 135578884-135578884 on Build GRCh38
rs369294181 Chr.5: 135579095-135579095 on Build GRCh38
rs369843926 Chr.5: 135578833-135578833 on Build GRCh38
rs555250714 Chr.5: 135578938-135578938 on Build GRCh38
rs866006140 Chr.5: 135578859-135578859 on Build GRCh38
rs925551799 Chr.5: 135579015-135579015 on Build GRCh38
rs960724442 Chr.5: 135578961-135578961 on Build GRCh38
rs576469567 Chr.5: 135578979-135578979 on Build GRCh38
rs773218386 Chr.5: 135578812-135578812 on Build GRCh38
rs978432604 Chr.5: 135578904-135578904 on Build GRCh38
rs932327393 Chr.5: 135578860-135578860 on Build GRCh38
rs1033701457 Chr.5: 135578953-135578955 on Build GRCh38
rs1022760692 Chr.5: 135578891-135578891 on Build GRCh38
rs759288541 Chr.5: 135578855-135578855 on Build GRCh38
rs945555366 Chr.5: 135578837-135578837 on Build GRCh38
rs577403512 Chr.5: 135579056-135579056 on Build GRCh38
rs778702230 Chr.5: 135578692-135578692 on Build GRCh38
rs979692569 Chr.5: 135578842-135578842 on Build GRCh38
rs769977099 Chr.5: 135579093-135579093 on Build GRCh38
rs748420774 Chr.5: 135578715-135578715 on Build GRCh38
rs536497276 Chr.5: 135578925-135578925 on Build GRCh38
rs989102079 Chr.5: 135578922-135578922 on Build GRCh38
rs1004522637 Chr.5: 135578984-135578984 on Build GRCh38
rs538380085 Chr.5: 135578779-135578779 on Build GRCh38
rs992234606 Chr.5: 135578796-135578796 on Build GRCh38
rs780617344 Chr.5: 135578683-135578683 on Build GRCh38
rs1004025232 Chr.5: 135578853-135578853 on Build GRCh38
rs1039124835 Chr.5: 135578806-135578806 on Build GRCh38
rs749556019 Chr.5: 135578684-135578684 on Build GRCh38
rs926199799 Chr.5: 135578914-135578914 on Build GRCh38
rs531217502 Chr.5: 135578659-135578659 on Build GRCh38
rs776655717 Chr.5: 135578839-135578839 on Build GRCh38
rs978430247 Chr.5: 135579064-135579064 on Build GRCh38
rs905673440 Chr.5: 135579083-135579083 on Build GRCh38
rs992570763 Chr.5: 135578890-135578890 on Build GRCh38
rs772253716 Chr.5: 135578780-135578780 on Build GRCh38
rs979535193 Chr.5: 135579006-135579006 on Build GRCh38
rs1014100310 Chr.5: 135579028-135579028 on Build GRCh38
rs969673964 Chr.5: 135578893-135578893 on Build GRCh38
rs1015544508 Chr.5: 135579013-135579013 on Build GRCh38
rs1053711974 Chr.5: 135578784-135578784 on Build GRCh38
rs563142483 Chr.5: 135578689-135578689 on Build GRCh38
rs111500143 Chr.5: 135578889-135578889 on Build GRCh38
rs931149866 Chr.5: 135579048-135579048 on Build GRCh38
rs543857157 Chr.5: 135578982-135578982 on Build GRCh38
rs1036771214 Chr.5: 135578920-135578920 on Build GRCh38
rs528340046 Chr.5: 135578950-135578961 on Build GRCh38
rs760732075 Chr.5: 135578678-135578682 on Build GRCh38
rs2072347 Chr.5: 135578660-135578660 on Build GRCh38
rs980731970 Chr.5: 135578765-135578765 on Build GRCh38
rs1033074882 Chr.5: 135579075-135579075 on Build GRCh38
rs926200666 Chr.5: 135578777-135578777 on Build GRCh38
rs1022688411 Chr.5: 135579029-135579029 on Build GRCh38
rs376806377 Chr.5: 135578863-135578863 on Build GRCh38
rs559017145 Chr.5: 135579039-135579039 on Build GRCh38
rs371946511 Chr.5: 135578701-135578701 on Build GRCh38
rs994797666 Chr.5: 135578822-135578822 on Build GRCh38

View Full Product Details