Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 137945766-137946280 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM13B
Entrez Gene ID
51306
Gene Name
family with sequence similarity 13 member B
Gene Aliases
ARHGAP49, C5orf5, FAM13B1, KHCHP, N61
UniGene
Hs.567453
Gene Chromosome Location
Chr.5: 137937909-138051961 on Build GRCh38
Transcripts
NM_001101800.2 NM_001101801.2 NM_016603.3 XM_005272007.1 XM_006714647.1 XM_006714648.1 XM_006714649.1 XM_011543448.1 XM_011543449.1 XM_011543450.1 XM_017009549.1 XM_017009550.1 XM_017009551.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6469948 c.2179-2A>T p.? Chr.5: 137945999-137945999 on Build GRCh38
269133 c.2229G>A p.R743R Chr.5: 137945947-137945947 on Build GRCh38
6658577 c.2151T>C p.L717L Chr.5: 137946255-137946255 on Build GRCh38
6469949 c.1807-2A>T p.? Chr.5: 137945999-137945999 on Build GRCh38
1251788 c.2127A>T p.K709N Chr.5: 137946279-137946279 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs933516861 Chr.5: 137945846-137945846 on Build GRCh38
rs756714258 Chr.5: 137946007-137946008 on Build GRCh38
rs766873343 Chr.5: 137946222-137946222 on Build GRCh38
rs182482927 Chr.5: 137945998-137945998 on Build GRCh38
rs373582194 Chr.5: 137946075-137946075 on Build GRCh38
rs41298974 Chr.5: 137945931-137945931 on Build GRCh38
rs780479298 Chr.5: 137945877-137945877 on Build GRCh38
rs575264812 Chr.5: 137945920-137945920 on Build GRCh38
rs770841043 Chr.5: 137945877-137945892 on Build GRCh38
rs1042638749 Chr.5: 137946061-137946061 on Build GRCh38
rs770656130 Chr.5: 137945933-137945933 on Build GRCh38
rs767972919 Chr.5: 137945947-137945947 on Build GRCh38
rs375497404 Chr.5: 137946020-137946020 on Build GRCh38
rs372676831 Chr.5: 137946031-137946031 on Build GRCh38
rs759942646 Chr.5: 137946238-137946238 on Build GRCh38
rs770746185 Chr.5: 137946029-137946029 on Build GRCh38
rs889893086 Chr.5: 137946078-137946078 on Build GRCh38
rs752144290 Chr.5: 137945851-137945851 on Build GRCh38
rs759066899 Chr.5: 137946120-137946120 on Build GRCh38
rs954777106 Chr.5: 137946227-137946227 on Build GRCh38
rs1008255713 Chr.5: 137946091-137946091 on Build GRCh38
rs775761202 Chr.5: 137946182-137946185 on Build GRCh38
rs776272103 Chr.5: 137945937-137945937 on Build GRCh38
rs1125979 Chr.5: 137946019-137946019 on Build GRCh38
rs773114267 Chr.5: 137945876-137945876 on Build GRCh38
rs908159996 Chr.5: 137945914-137945914 on Build GRCh38
rs1021923013 Chr.5: 137946101-137946101 on Build GRCh38
rs139843402 Chr.5: 137945956-137945956 on Build GRCh38
rs898156993 Chr.5: 137945997-137945997 on Build GRCh38
rs762426637 Chr.5: 137946210-137946210 on Build GRCh38
rs199713198 Chr.5: 137945977-137945977 on Build GRCh38
rs769963509 Chr.5: 137946008-137946008 on Build GRCh38
rs41298976 Chr.5: 137945986-137945986 on Build GRCh38
rs375876942 Chr.5: 137946049-137946049 on Build GRCh38
rs750789373 Chr.5: 137945865-137945865 on Build GRCh38
rs777587041 Chr.5: 137946021-137946021 on Build GRCh38
rs765435782 Chr.5: 137946253-137946253 on Build GRCh38
rs142201748 Chr.5: 137946069-137946069 on Build GRCh38
rs78781927 Chr.5: 137945943-137945943 on Build GRCh38
rs1018749157 Chr.5: 137946103-137946103 on Build GRCh38
rs761221584 Chr.5: 137946221-137946221 on Build GRCh38
rs773858172 Chr.5: 137946220-137946220 on Build GRCh38
rs942276277 Chr.5: 137945976-137945976 on Build GRCh38
rs759153274 Chr.5: 137945941-137945941 on Build GRCh38
rs187331921 Chr.5: 137945932-137945932 on Build GRCh38
rs200475728 Chr.5: 137946177-137946177 on Build GRCh38
rs755112177 Chr.5: 137945910-137945910 on Build GRCh38
rs768151938 Chr.5: 137946216-137946216 on Build GRCh38
rs1011490936 Chr.5: 137946092-137946092 on Build GRCh38
rs996552947 Chr.5: 137946128-137946128 on Build GRCh38
rs923317005 Chr.5: 137945809-137945809 on Build GRCh38
rs541759749 Chr.5: 137945845-137945845 on Build GRCh38
rs754302218 Chr.5: 137946229-137946229 on Build GRCh38
rs769368742 Chr.5: 137946045-137946045 on Build GRCh38
rs759646694 Chr.5: 137945997-137945998 on Build GRCh38
rs1042440145 Chr.5: 137945794-137945794 on Build GRCh38
rs776731227 Chr.5: 137945941-137945942 on Build GRCh38
rs901661938 Chr.5: 137946115-137946115 on Build GRCh38
rs746884881 Chr.5: 137946022-137946022 on Build GRCh38
rs139041315 Chr.5: 137946052-137946052 on Build GRCh38
rs771944306 Chr.5: 137945927-137945927 on Build GRCh38
rs918076686 Chr.5: 137946039-137946039 on Build GRCh38
rs745644540 Chr.5: 137946041-137946041 on Build GRCh38
rs556788319 Chr.5: 137945857-137945857 on Build GRCh38
rs369564411 Chr.5: 137945919-137945919 on Build GRCh38
rs749462301 Chr.5: 137945897-137945897 on Build GRCh38
rs993380354 Chr.5: 137946157-137946157 on Build GRCh38
rs146367502 Chr.5: 137946074-137946074 on Build GRCh38
rs766780763 Chr.5: 137945967-137945967 on Build GRCh38
rs756540601 Chr.5: 137945870-137945870 on Build GRCh38
rs757660624 Chr.5: 137945856-137945856 on Build GRCh38
rs746782941 Chr.5: 137945874-137945874 on Build GRCh38
rs767120132 Chr.5: 137946147-137946147 on Build GRCh38
rs761133442 Chr.5: 137945960-137945960 on Build GRCh38
rs1026614180 Chr.5: 137946186-137946186 on Build GRCh38

View Full Product Details