Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 138152404-138152918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD8
Entrez Gene ID
10902
Gene Name
bromodomain containing 8
Gene Aliases
SMAP, SMAP2, p120
UniGene
Hs.519337
Gene Chromosome Location
Chr.5: 138139766-138178669 on Build GRCh38
Transcripts
NM_139199.1 XM_005271855.2 XM_005271856.2 XM_005271857.2 XM_005271859.3 XM_011543109.1 XM_011543110.1 XM_017008969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5794040 c.2621C>T p.P874L Chr.5: 138152717-138152717 on Build GRCh38
3608880 c.2617C>T p.H873Y Chr.5: 138152721-138152721 on Build GRCh38
5867348 c.2758_2759delGA p.E920fs*2 Chr.5: 138152579-138152580 on Build GRCh38
4483944 c.2750C>T p.P917L Chr.5: 138152588-138152588 on Build GRCh38
1184908 c.2748C>A p.S916R Chr.5: 138152590-138152590 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761633285 Chr.5: 138152469-138152469 on Build GRCh38
rs754120454 Chr.5: 138152785-138152785 on Build GRCh38
rs774316641 Chr.5: 138152570-138152570 on Build GRCh38
rs780209773 Chr.5: 138152681-138152681 on Build GRCh38
rs551000114 Chr.5: 138152867-138152867 on Build GRCh38
rs757679501 Chr.5: 138152533-138152533 on Build GRCh38
rs6883021 Chr.5: 138152651-138152651 on Build GRCh38
rs766067381 Chr.5: 138152601-138152601 on Build GRCh38
rs776299411 Chr.5: 138152618-138152618 on Build GRCh38
rs376591896 Chr.5: 138152806-138152806 on Build GRCh38
rs557433008 Chr.5: 138152455-138152455 on Build GRCh38
rs748186283 Chr.5: 138152699-138152699 on Build GRCh38
rs755191358 Chr.5: 138152793-138152793 on Build GRCh38
rs375407999 Chr.5: 138152490-138152490 on Build GRCh38
rs774155127 Chr.5: 138152468-138152468 on Build GRCh38
rs930630095 Chr.5: 138152677-138152677 on Build GRCh38
rs767127696 Chr.5: 138152527-138152527 on Build GRCh38
rs368730028 Chr.5: 138152641-138152641 on Build GRCh38
rs377420819 Chr.5: 138152752-138152752 on Build GRCh38
rs764808196 Chr.5: 138152625-138152625 on Build GRCh38
rs138308684 Chr.5: 138152694-138152694 on Build GRCh38
rs941544177 Chr.5: 138152526-138152526 on Build GRCh38
rs374530177 Chr.5: 138152671-138152671 on Build GRCh38
rs111670295 Chr.5: 138152444-138152444 on Build GRCh38
rs755103270 Chr.5: 138152684-138152684 on Build GRCh38
rs201588407 Chr.5: 138152471-138152471 on Build GRCh38
rs369736947 Chr.5: 138152784-138152784 on Build GRCh38
rs767931244 Chr.5: 138152649-138152649 on Build GRCh38
rs199684486 Chr.5: 138152701-138152701 on Build GRCh38
rs373610851 Chr.5: 138152804-138152804 on Build GRCh38
rs999408030 Chr.5: 138152643-138152643 on Build GRCh38
rs75165147 Chr.5: 138152866-138152866 on Build GRCh38
rs539478006 Chr.5: 138152453-138152453 on Build GRCh38
rs552596205 Chr.5: 138152749-138152749 on Build GRCh38
rs200412361 Chr.5: 138152560-138152560 on Build GRCh38
rs1046573240 Chr.5: 138152872-138152872 on Build GRCh38
rs972171028 Chr.5: 138152666-138152666 on Build GRCh38
rs751136954 Chr.5: 138152481-138152481 on Build GRCh38
rs373344329 Chr.5: 138152838-138152838 on Build GRCh38
rs568495274 Chr.5: 138152829-138152829 on Build GRCh38
rs145741800 Chr.5: 138152683-138152683 on Build GRCh38
rs766051001 Chr.5: 138152496-138152496 on Build GRCh38
rs767862583 Chr.5: 138152766-138152766 on Build GRCh38
rs758983450 Chr.5: 138152522-138152522 on Build GRCh38
rs779456548 Chr.5: 138152706-138152707 on Build GRCh38
rs761072491 Chr.5: 138152770-138152770 on Build GRCh38
rs754201802 Chr.5: 138152467-138152468 on Build GRCh38
rs150159308 Chr.5: 138152558-138152558 on Build GRCh38
rs201442540 Chr.5: 138152645-138152645 on Build GRCh38
rs759077090 Chr.5: 138152623-138152623 on Build GRCh38
rs749230187 Chr.5: 138152457-138152457 on Build GRCh38
rs146791453 Chr.5: 138152646-138152646 on Build GRCh38
rs753328688 Chr.5: 138152504-138152504 on Build GRCh38
rs369431228 Chr.5: 138152542-138152542 on Build GRCh38
rs951462903 Chr.5: 138152777-138152777 on Build GRCh38
rs781386566 Chr.5: 138152536-138152536 on Build GRCh38
rs916323261 Chr.5: 138152857-138152857 on Build GRCh38
rs999897147 Chr.5: 138152587-138152587 on Build GRCh38
rs770735098 Chr.5: 138152714-138152714 on Build GRCh38
rs369563060 Chr.5: 138152830-138152830 on Build GRCh38
rs749485267 Chr.5: 138152682-138152682 on Build GRCh38
rs976743368 Chr.5: 138152746-138152746 on Build GRCh38
rs759239911 Chr.5: 138152750-138152750 on Build GRCh38
rs758978639 Chr.5: 138152488-138152488 on Build GRCh38
rs140870898 Chr.5: 138152557-138152557 on Build GRCh38
rs999844806 Chr.5: 138152850-138152850 on Build GRCh38
rs986681622 Chr.5: 138152627-138152627 on Build GRCh38
rs374458782 Chr.5: 138152767-138152767 on Build GRCh38
rs148605042 Chr.5: 138152751-138152751 on Build GRCh38
rs762397400 Chr.5: 138152753-138152753 on Build GRCh38
rs374460094 Chr.5: 138152588-138152588 on Build GRCh38
rs947720420 Chr.5: 138152544-138152544 on Build GRCh38
rs755421084 Chr.5: 138152625-138152626 on Build GRCh38
rs910140277 Chr.5: 138152799-138152799 on Build GRCh38
rs768523710 Chr.5: 138152464-138152464 on Build GRCh38
rs763123507 Chr.5: 138152605-138152605 on Build GRCh38
rs777634763 Chr.5: 138152711-138152711 on Build GRCh38
rs113105715 Chr.5: 138152790-138152790 on Build GRCh38
rs748015811 Chr.5: 138152577-138152577 on Build GRCh38
rs866573028 Chr.5: 138152610-138152610 on Build GRCh38
rs547001762 Chr.5: 138152822-138152822 on Build GRCh38
rs746729786 Chr.5: 138152713-138152713 on Build GRCh38
rs752319711 Chr.5: 138152528-138152528 on Build GRCh38
rs760376365 Chr.5: 138152597-138152597 on Build GRCh38
rs199793838 Chr.5: 138152591-138152591 on Build GRCh38
rs776463836 Chr.5: 138152736-138152736 on Build GRCh38

View Full Product Details